Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 58262292-58262834 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC113
Entrez Gene ID
29070
Gene Name
coiled-coil domain containing 113
Gene Aliases
HSPC065
UniGene
Hs.11614
Gene Chromosome Location
Chr.16: 58196863-58283836 on Build GRCh38
Transcripts
NM_001142302.1 NM_014157.3 XM_011523038.2 XM_011523039.2 XM_011523040.2 XM_011523041.2 XM_011523042.2 XM_017023176.1 XM_017023177.1 XM_017023178.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3937105 c.669G>T p.Q223H Chr.16: 58262426-58262426 on Build GRCh38
971966 c.697C>A p.L233I Chr.16: 58262454-58262454 on Build GRCh38
971967 c.709G>T p.E237* Chr.16: 58262466-58262466 on Build GRCh38
971965 c.655G>A p.D219N Chr.16: 58262412-58262412 on Build GRCh38
3932373 c.655G>T p.D219Y Chr.16: 58262412-58262412 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201907484 Chr.16: 58262759-58262760 on Build GRCh38
rs199740987 Chr.16: 58262764-58262771 on Build GRCh38
rs566275467 Chr.16: 58262800-58262800 on Build GRCh38
rs777889646 Chr.16: 58262585-58262585 on Build GRCh38
rs977846524 Chr.16: 58262443-58262443 on Build GRCh38
rs970317077 Chr.16: 58262561-58262561 on Build GRCh38
rs1053574547 Chr.16: 58262362-58262362 on Build GRCh38
rs748325662 Chr.16: 58262468-58262468 on Build GRCh38
rs759294574 Chr.16: 58262588-58262589 on Build GRCh38
rs9921710 Chr.16: 58262787-58262787 on Build GRCh38
rs1040717246 Chr.16: 58262653-58262653 on Build GRCh38
rs772076821 Chr.16: 58262382-58262382 on Build GRCh38
rs1006013949 Chr.16: 58262749-58262749 on Build GRCh38
rs758599504 Chr.16: 58262584-58262584 on Build GRCh38
rs773247826 Chr.16: 58262485-58262485 on Build GRCh38
rs766229061 Chr.16: 58262565-58262565 on Build GRCh38
rs746685604 Chr.16: 58262381-58262382 on Build GRCh38
rs1040966503 Chr.16: 58262598-58262598 on Build GRCh38
rs376693262 Chr.16: 58262413-58262413 on Build GRCh38
rs571843999 Chr.16: 58262758-58262758 on Build GRCh38
rs1034722462 Chr.16: 58262346-58262346 on Build GRCh38
rs183861532 Chr.16: 58262359-58262359 on Build GRCh38
rs778750262 Chr.16: 58262700-58262700 on Build GRCh38
rs369732429 Chr.16: 58262506-58262506 on Build GRCh38
rs71155255 Chr.16: 58262806-58262807 on Build GRCh38
rs563168078 Chr.16: 58262637-58262637 on Build GRCh38
rs761049442 Chr.16: 58262388-58262388 on Build GRCh38
rs1014315287 Chr.16: 58262805-58262808 on Build GRCh38
rs766179592 Chr.16: 58262587-58262587 on Build GRCh38
rs57582823 Chr.16: 58262800-58262807 on Build GRCh38
rs201764015 Chr.16: 58262556-58262556 on Build GRCh38
rs373005747 Chr.16: 58262530-58262530 on Build GRCh38
rs768476365 Chr.16: 58262582-58262583 on Build GRCh38
rs980789162 Chr.16: 58262574-58262574 on Build GRCh38
rs192239074 Chr.16: 58262353-58262353 on Build GRCh38
rs897262183 Chr.16: 58262772-58262772 on Build GRCh38
rs375707775 Chr.16: 58262743-58262746 on Build GRCh38
rs57284997 Chr.16: 58262807-58262808 on Build GRCh38
rs879068388 Chr.16: 58262515-58262516 on Build GRCh38
rs761525864 Chr.16: 58262548-58262548 on Build GRCh38
rs756370138 Chr.16: 58262801-58262808 on Build GRCh38
rs557683673 Chr.16: 58262734-58262737 on Build GRCh38
rs1015632578 Chr.16: 58262540-58262540 on Build GRCh38
rs762833992 Chr.16: 58262585-58262586 on Build GRCh38
rs374584564 Chr.16: 58262418-58262418 on Build GRCh38
rs757974268 Chr.16: 58262760-58262763 on Build GRCh38
rs143745173 Chr.16: 58262500-58262500 on Build GRCh38
rs762718354 Chr.16: 58262803-58262804 on Build GRCh38
rs771331010 Chr.16: 58262501-58262501 on Build GRCh38
rs945000805 Chr.16: 58262652-58262652 on Build GRCh38
rs1033561510 Chr.16: 58262791-58262791 on Build GRCh38
rs9931492 Chr.16: 58262808-58262808 on Build GRCh38
rs141343672 Chr.16: 58262787-58262788 on Build GRCh38
rs764442730 Chr.16: 58262589-58262590 on Build GRCh38
rs749885935 Chr.16: 58262560-58262560 on Build GRCh38
rs751190954 Chr.16: 58262412-58262412 on Build GRCh38
rs573698132 Chr.16: 58262445-58262445 on Build GRCh38
rs894442347 Chr.16: 58262765-58262765 on Build GRCh38
rs376207984 Chr.16: 58262355-58262355 on Build GRCh38
rs1005348435 Chr.16: 58262757-58262763 on Build GRCh38
rs546840172 Chr.16: 58262781-58262781 on Build GRCh38
rs779974395 Chr.16: 58262636-58262636 on Build GRCh38
rs769660685 Chr.16: 58262350-58262350 on Build GRCh38
rs536316164 Chr.16: 58262803-58262803 on Build GRCh38
rs369843578 Chr.16: 58262527-58262527 on Build GRCh38
rs28479863 Chr.16: 58262763-58262763 on Build GRCh38
rs923711571 Chr.16: 58262465-58262465 on Build GRCh38
rs942614846 Chr.16: 58262562-58262562 on Build GRCh38
rs1024498324 Chr.16: 58262771-58262771 on Build GRCh38
rs9921709 Chr.16: 58262783-58262783 on Build GRCh38
rs753747229 Chr.16: 58262571-58262571 on Build GRCh38
rs901109770 Chr.16: 58262421-58262421 on Build GRCh38
rs777122503 Chr.16: 58262504-58262504 on Build GRCh38
rs764277741 Chr.16: 58262763-58262764 on Build GRCh38
rs774180020 Chr.16: 58262586-58262587 on Build GRCh38
rs551025365 Chr.16: 58262430-58262430 on Build GRCh38
rs1015601305 Chr.16: 58262758-58262769 on Build GRCh38
rs754131934 Chr.16: 58262398-58262398 on Build GRCh38
rs748413468 Chr.16: 58262583-58262583 on Build GRCh38
rs371899294 Chr.16: 58262351-58262351 on Build GRCh38
rs201620741 Chr.16: 58262764-58262775 on Build GRCh38
rs773734746 Chr.16: 58262385-58262385 on Build GRCh38
rs957827619 Chr.16: 58262807-58262807 on Build GRCh38
rs869267898 Chr.16: 58262758-58262759 on Build GRCh38
rs539346992 Chr.16: 58262767-58262767 on Build GRCh38
rs779954350 Chr.16: 58262444-58262444 on Build GRCh38
rs558975207 Chr.16: 58262523-58262523 on Build GRCh38
rs541233791 Chr.16: 58262456-58262456 on Build GRCh38
rs989613773 Chr.16: 58262811-58262811 on Build GRCh38
rs554645771 Chr.16: 58262804-58262804 on Build GRCh38
rs188553365 Chr.16: 58262411-58262411 on Build GRCh38
rs776589145 Chr.16: 58262543-58262543 on Build GRCh38
rs766810491 Chr.16: 58262393-58262393 on Build GRCh38
rs750972665 Chr.16: 58262592-58262593 on Build GRCh38
rs768427983 Chr.16: 58262458-58262458 on Build GRCh38
rs772174543 Chr.16: 58262472-58262472 on Build GRCh38
rs200306973 Chr.16: 58262549-58262549 on Build GRCh38
rs547659732 Chr.16: 58262799-58262799 on Build GRCh38
rs892098932 Chr.16: 58262365-58262365 on Build GRCh38
rs754747181 Chr.16: 58262575-58262575 on Build GRCh38
rs750424211 Chr.16: 58262428-58262428 on Build GRCh38
rs900866689 Chr.16: 58262665-58262665 on Build GRCh38
rs3815802 Chr.16: 58262641-58262641 on Build GRCh38
rs778644250 Chr.16: 58262576-58262576 on Build GRCh38
rs1050148191 Chr.16: 58262730-58262730 on Build GRCh38
rs547748302 Chr.16: 58262619-58262619 on Build GRCh38
rs936379015 Chr.16: 58262490-58262490 on Build GRCh38
rs542580167 Chr.16: 58262488-58262488 on Build GRCh38
rs780808620 Chr.16: 58262424-58262424 on Build GRCh38
rs961322826 Chr.16: 58262547-58262547 on Build GRCh38
rs779116738 Chr.16: 58262463-58262463 on Build GRCh38
rs570736086 Chr.16: 58262698-58262698 on Build GRCh38
rs9923694 Chr.16: 58262638-58262638 on Build GRCh38

View Full Product Details