Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 58267401-58267903 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC113
Entrez Gene ID
29070
Gene Name
coiled-coil domain containing 113
Gene Aliases
HSPC065
UniGene
Hs.11614
Gene Chromosome Location
Chr.16: 58196863-58283836 on Build GRCh38
Transcripts
NM_001142302.1 NM_014157.3 XM_011523038.2 XM_011523039.2 XM_011523040.2 XM_011523041.2 XM_011523042.2 XM_017023176.1 XM_017023177.1 XM_017023178.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
353924 c.886G>T p.E296* Chr.16: 58267581-58267581 on Build GRCh38
1493651 c.828G>T p.K276N Chr.16: 58267523-58267523 on Build GRCh38
1749673 c.856G>A p.E286K Chr.16: 58267551-58267551 on Build GRCh38
3510607 c.796A>G p.K266E Chr.16: 58267491-58267491 on Build GRCh38
4612753 c.856_857insA p.I288fs*2 Chr.16: 58267551-58267552 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs979434258 Chr.16: 58267766-58267766 on Build GRCh38
rs767272063 Chr.16: 58267472-58267472 on Build GRCh38
rs746410924 Chr.16: 58267574-58267574 on Build GRCh38
rs1039836206 Chr.16: 58267580-58267580 on Build GRCh38
rs1014123100 Chr.16: 58267724-58267724 on Build GRCh38
rs747056937 Chr.16: 58267428-58267428 on Build GRCh38
rs772851671 Chr.16: 58267474-58267474 on Build GRCh38
rs960641729 Chr.16: 58267825-58267825 on Build GRCh38
rs967156158 Chr.16: 58267760-58267760 on Build GRCh38
rs541466393 Chr.16: 58267526-58267526 on Build GRCh38
rs777863353 Chr.16: 58267541-58267541 on Build GRCh38
rs757307503 Chr.16: 58267558-58267558 on Build GRCh38
rs370621578 Chr.16: 58267503-58267503 on Build GRCh38
rs746346498 Chr.16: 58267453-58267453 on Build GRCh38
rs1023468682 Chr.16: 58267838-58267838 on Build GRCh38
rs969209986 Chr.16: 58267854-58267854 on Build GRCh38
rs35986384 Chr.16: 58267481-58267481 on Build GRCh38
rs995879499 Chr.16: 58267677-58267677 on Build GRCh38
rs755392432 Chr.16: 58267563-58267564 on Build GRCh38
rs1021033634 Chr.16: 58267743-58267743 on Build GRCh38
rs753754241 Chr.16: 58267505-58267505 on Build GRCh38
rs747197430 Chr.16: 58267547-58267547 on Build GRCh38
rs754881650 Chr.16: 58267508-58267508 on Build GRCh38
rs796341324 Chr.16: 58267456-58267457 on Build GRCh38
rs749672037 Chr.16: 58267578-58267578 on Build GRCh38
rs547011366 Chr.16: 58267436-58267436 on Build GRCh38
rs900045694 Chr.16: 58267593-58267593 on Build GRCh38
rs538325570 Chr.16: 58267708-58267708 on Build GRCh38
rs750724854 Chr.16: 58267456-58267457 on Build GRCh38
rs372831969 Chr.16: 58267577-58267577 on Build GRCh38
rs1015026416 Chr.16: 58267814-58267814 on Build GRCh38
rs758407001 Chr.16: 58267670-58267670 on Build GRCh38
rs944212499 Chr.16: 58267881-58267881 on Build GRCh38
rs760331227 Chr.16: 58267624-58267624 on Build GRCh38
rs775973873 Chr.16: 58267461-58267461 on Build GRCh38
rs770322176 Chr.16: 58267459-58267459 on Build GRCh38
rs186061173 Chr.16: 58267692-58267692 on Build GRCh38
rs181472780 Chr.16: 58267641-58267641 on Build GRCh38
rs985691180 Chr.16: 58267827-58267827 on Build GRCh38
rs925243987 Chr.16: 58267786-58267786 on Build GRCh38
rs770410186 Chr.16: 58267575-58267575 on Build GRCh38
rs1002054827 Chr.16: 58267534-58267534 on Build GRCh38
rs1004570183 Chr.16: 58267790-58267790 on Build GRCh38
rs752478299 Chr.16: 58267521-58267521 on Build GRCh38
rs765068408 Chr.16: 58267513-58267513 on Build GRCh38
rs3833047 Chr.16: 58267457-58267458 on Build GRCh38
rs1048805503 Chr.16: 58267714-58267714 on Build GRCh38
rs781301504 Chr.16: 58267563-58267563 on Build GRCh38
rs556650835 Chr.16: 58267796-58267796 on Build GRCh38
rs777720217 Chr.16: 58267674-58267674 on Build GRCh38
rs772758673 Chr.16: 58267601-58267601 on Build GRCh38
rs769081208 Chr.16: 58267592-58267592 on Build GRCh38
rs578146049 Chr.16: 58267828-58267828 on Build GRCh38
rs747423196 Chr.16: 58267551-58267552 on Build GRCh38

View Full Product Details