Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 58516729-58517228 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SETD6
Entrez Gene ID
79918
Gene Name
SET domain containing 6
Gene Aliases
-
UniGene
Hs.731691
Gene Chromosome Location
Chr.16: 58515479-58521961 on Build GRCh38
Transcripts
NM_001160305.2 NM_024860.3 NR_134583.1 XM_011523336.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6775958 c.680A>G p.N227S Chr.16: 58516888-58516888 on Build GRCh38
6775948 c.771C>T p.N257N Chr.16: 58516907-58516907 on Build GRCh38
3387514 c.604C>T p.Q202* Chr.16: 58516812-58516812 on Build GRCh38
471876 c.647C>G p.S216C Chr.16: 58516855-58516855 on Build GRCh38
6775945 c.605A>G p.Q202R Chr.16: 58516813-58516813 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753760336 Chr.16: 58517022-58517022 on Build GRCh38
rs143850199 Chr.16: 58516906-58516906 on Build GRCh38
rs772468371 Chr.16: 58516902-58516902 on Build GRCh38
rs754472423 Chr.16: 58517029-58517029 on Build GRCh38
rs746380318 Chr.16: 58517065-58517065 on Build GRCh38
rs753947008 Chr.16: 58516910-58516910 on Build GRCh38
rs771456150 Chr.16: 58516968-58516968 on Build GRCh38
rs773669405 Chr.16: 58516905-58516905 on Build GRCh38
rs927261678 Chr.16: 58517166-58517166 on Build GRCh38
rs1019791988 Chr.16: 58517084-58517084 on Build GRCh38
rs1053381932 Chr.16: 58517064-58517064 on Build GRCh38
rs781152134 Chr.16: 58516924-58516924 on Build GRCh38
rs769467241 Chr.16: 58516793-58516793 on Build GRCh38
rs994509596 Chr.16: 58517104-58517107 on Build GRCh38
rs550313178 Chr.16: 58517138-58517138 on Build GRCh38
rs369607433 Chr.16: 58516988-58516988 on Build GRCh38
rs767351663 Chr.16: 58516992-58516992 on Build GRCh38
rs768103500 Chr.16: 58516944-58516944 on Build GRCh38
rs761093341 Chr.16: 58516963-58516963 on Build GRCh38
rs773115193 Chr.16: 58517044-58517045 on Build GRCh38
rs199982414 Chr.16: 58516945-58516945 on Build GRCh38
rs367708763 Chr.16: 58516823-58516823 on Build GRCh38
rs758040051 Chr.16: 58517046-58517046 on Build GRCh38
rs762217310 Chr.16: 58516892-58516892 on Build GRCh38
rs202031310 Chr.16: 58516808-58516808 on Build GRCh38
rs984214532 Chr.16: 58516927-58516927 on Build GRCh38
rs778311601 Chr.16: 58517035-58517035 on Build GRCh38
rs149833434 Chr.16: 58517156-58517156 on Build GRCh38
rs112854734 Chr.16: 58516891-58516891 on Build GRCh38
rs537112332 Chr.16: 58517195-58517195 on Build GRCh38
rs750242019 Chr.16: 58517001-58517001 on Build GRCh38
rs867106592 Chr.16: 58517020-58517020 on Build GRCh38
rs777153193 Chr.16: 58517063-58517063 on Build GRCh38
rs750324018 Chr.16: 58516928-58516928 on Build GRCh38
rs779070359 Chr.16: 58516848-58516848 on Build GRCh38
rs377260676 Chr.16: 58516876-58516876 on Build GRCh38
rs371517288 Chr.16: 58516852-58516852 on Build GRCh38
rs752900100 Chr.16: 58516917-58516917 on Build GRCh38
rs756157885 Chr.16: 58516830-58516830 on Build GRCh38
rs753959451 Chr.16: 58516839-58516839 on Build GRCh38
rs1025519307 Chr.16: 58517144-58517144 on Build GRCh38
rs545831603 Chr.16: 58516883-58516883 on Build GRCh38
rs765507593 Chr.16: 58516768-58516768 on Build GRCh38
rs768001927 Chr.16: 58516818-58516818 on Build GRCh38
rs770986535 Chr.16: 58516771-58516771 on Build GRCh38
rs971230335 Chr.16: 58516859-58516859 on Build GRCh38
rs755231669 Chr.16: 58516847-58516847 on Build GRCh38
rs768248041 Chr.16: 58516879-58516887 on Build GRCh38
rs984161812 Chr.16: 58517160-58517160 on Build GRCh38
rs766476778 Chr.16: 58516833-58516833 on Build GRCh38
rs765474032 Chr.16: 58516916-58516916 on Build GRCh38
rs751996802 Chr.16: 58517039-58517039 on Build GRCh38
rs35425008 Chr.16: 58516884-58516884 on Build GRCh38
rs917730957 Chr.16: 58516990-58516990 on Build GRCh38
rs965903407 Chr.16: 58517108-58517108 on Build GRCh38
rs771239199 Chr.16: 58517059-58517059 on Build GRCh38
rs1025870489 Chr.16: 58517116-58517116 on Build GRCh38
rs762748078 Chr.16: 58517059-58517060 on Build GRCh38
rs994429337 Chr.16: 58517109-58517109 on Build GRCh38
rs115215180 Chr.16: 58516952-58516952 on Build GRCh38
rs369202486 Chr.16: 58516971-58516971 on Build GRCh38
rs769851763 Chr.16: 58517068-58517068 on Build GRCh38
rs764312293 Chr.16: 58516800-58516800 on Build GRCh38
rs949345380 Chr.16: 58516999-58516999 on Build GRCh38
rs779430968 Chr.16: 58517100-58517100 on Build GRCh38
rs757724531 Chr.16: 58517073-58517073 on Build GRCh38
rs867247558 Chr.16: 58516801-58516801 on Build GRCh38
rs746804612 Chr.16: 58516851-58516851 on Build GRCh38
rs542442212 Chr.16: 58516758-58516758 on Build GRCh38
rs779926228 Chr.16: 58516935-58516935 on Build GRCh38
rs3743568 Chr.16: 58516756-58516756 on Build GRCh38
rs200494659 Chr.16: 58516783-58516783 on Build GRCh38
rs889566394 Chr.16: 58517066-58517066 on Build GRCh38
rs530582932 Chr.16: 58516860-58516860 on Build GRCh38
rs773581375 Chr.16: 58516962-58516962 on Build GRCh38
rs777458084 Chr.16: 58517047-58517047 on Build GRCh38
rs745829879 Chr.16: 58516858-58516858 on Build GRCh38
rs1037986050 Chr.16: 58517079-58517079 on Build GRCh38
rs745346231 Chr.16: 58516792-58516792 on Build GRCh38
rs551926500 Chr.16: 58516760-58516760 on Build GRCh38
rs747830878 Chr.16: 58516948-58516948 on Build GRCh38
rs769785510 Chr.16: 58517017-58517019 on Build GRCh38
rs780522684 Chr.16: 58516903-58516904 on Build GRCh38
rs775239643 Chr.16: 58516795-58516795 on Build GRCh38
rs758089903 Chr.16: 58516764-58516764 on Build GRCh38
rs753010630 Chr.16: 58516977-58516978 on Build GRCh38
rs141078613 Chr.16: 58516983-58516983 on Build GRCh38
rs772379893 Chr.16: 58516955-58516955 on Build GRCh38
rs763278048 Chr.16: 58516986-58516986 on Build GRCh38
rs181310677 Chr.16: 58517124-58517124 on Build GRCh38
rs769375638 Chr.16: 58516870-58516870 on Build GRCh38
rs748820100 Chr.16: 58516937-58516937 on Build GRCh38
rs757075683 Chr.16: 58516921-58516921 on Build GRCh38
rs994042371 Chr.16: 58516850-58516850 on Build GRCh38
rs747677108 Chr.16: 58516953-58516954 on Build GRCh38
rs756129574 Chr.16: 58516933-58516933 on Build GRCh38
rs774787085 Chr.16: 58516807-58516807 on Build GRCh38
rs766378092 Chr.16: 58516909-58516909 on Build GRCh38
rs1000521128 Chr.16: 58517040-58517040 on Build GRCh38
rs759590325 Chr.16: 58516912-58516912 on Build GRCh38
rs898136178 Chr.16: 58517104-58517104 on Build GRCh38
rs748829120 Chr.16: 58516881-58516881 on Build GRCh38
rs1051298451 Chr.16: 58517037-58517037 on Build GRCh38
rs777052098 Chr.16: 58516969-58516969 on Build GRCh38
rs781242172 Chr.16: 58516853-58516853 on Build GRCh38
rs188644486 Chr.16: 58517017-58517017 on Build GRCh38
rs1016843893 Chr.16: 58516770-58516770 on Build GRCh38
rs750979736 Chr.16: 58516821-58516821 on Build GRCh38
rs775714163 Chr.16: 58517070-58517070 on Build GRCh38

View Full Product Details