Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 75228908-75229405 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCAR1
Entrez Gene ID
9564
Gene Name
BCAR1, Cas family scaffolding protein
Gene Aliases
CAS, CAS1, CASS1, CRKAS, P130Cas
UniGene
Hs.479747
Gene Chromosome Location
Chr.16: 75229030-75268053 on Build GRCh38
Transcripts
NM_001170714.1 NM_001170715.1 NM_001170716.1 NM_001170717.1 NM_001170718.1 NM_001170719.1 NM_001170720.1 NM_001170721.1 NM_014567.3 XM_005256258.4 XM_005256259.4 XM_005256260.4 XM_011523475.2 XM_017023897.1 XM_017023898.1 XM_017023899.1

SNP Detail

SNP ID Chromosome Location
rs912385961 Chr.16: 75229317-75229333 on Build GRCh38
rs955673565 Chr.16: 75229251-75229251 on Build GRCh38
rs989418062 Chr.16: 75228934-75228934 on Build GRCh38
rs773465805 Chr.16: 75228971-75228971 on Build GRCh38
rs1006122199 Chr.16: 75229073-75229073 on Build GRCh38
rs750206331 Chr.16: 75229170-75229170 on Build GRCh38
rs1029878318 Chr.16: 75229117-75229117 on Build GRCh38
rs761660157 Chr.16: 75229132-75229132 on Build GRCh38
rs72802389 Chr.16: 75229303-75229303 on Build GRCh38
rs565591821 Chr.16: 75229248-75229248 on Build GRCh38
rs531102613 Chr.16: 75229194-75229194 on Build GRCh38
rs999156332 Chr.16: 75229366-75229366 on Build GRCh38
rs973834507 Chr.16: 75229302-75229302 on Build GRCh38
rs924337636 Chr.16: 75229130-75229130 on Build GRCh38
rs1032342061 Chr.16: 75229368-75229368 on Build GRCh38
rs571366646 Chr.16: 75229123-75229123 on Build GRCh38
rs998913431 Chr.16: 75229252-75229252 on Build GRCh38
rs780857412 Chr.16: 75229326-75229326 on Build GRCh38
rs977542723 Chr.16: 75228968-75228968 on Build GRCh38
rs151033775 Chr.16: 75229273-75229273 on Build GRCh38
rs1029847154 Chr.16: 75229222-75229222 on Build GRCh38
rs929275817 Chr.16: 75229312-75229312 on Build GRCh38
rs143727544 Chr.16: 75229211-75229211 on Build GRCh38
rs927650466 Chr.16: 75228956-75228956 on Build GRCh38
rs990404963 Chr.16: 75229266-75229266 on Build GRCh38
rs565691316 Chr.16: 75228997-75228997 on Build GRCh38
rs915165276 Chr.16: 75229107-75229107 on Build GRCh38
rs1026766172 Chr.16: 75229029-75229029 on Build GRCh38
rs1047079926 Chr.16: 75229192-75229192 on Build GRCh38
rs183257480 Chr.16: 75229159-75229159 on Build GRCh38
rs550757255 Chr.16: 75228945-75228945 on Build GRCh38
rs1022583548 Chr.16: 75229224-75229224 on Build GRCh38
rs770417481 Chr.16: 75229173-75229173 on Build GRCh38
rs1045641633 Chr.16: 75229165-75229165 on Build GRCh38
rs571563343 Chr.16: 75229034-75229034 on Build GRCh38
rs748520832 Chr.16: 75228946-75228946 on Build GRCh38
rs11640206 Chr.16: 75229031-75229031 on Build GRCh38
rs190712814 Chr.16: 75229147-75229147 on Build GRCh38
rs969849014 Chr.16: 75229276-75229276 on Build GRCh38
rs751103019 Chr.16: 75229277-75229277 on Build GRCh38
rs1013008567 Chr.16: 75228998-75228998 on Build GRCh38
rs183438034 Chr.16: 75229059-75229059 on Build GRCh38
rs997016957 Chr.16: 75229095-75229095 on Build GRCh38
rs1031370279 Chr.16: 75229261-75229261 on Build GRCh38
rs557179447 Chr.16: 75229076-75229076 on Build GRCh38
rs983188887 Chr.16: 75229316-75229316 on Build GRCh38
rs964555655 Chr.16: 75229104-75229104 on Build GRCh38
rs549078097 Chr.16: 75229247-75229247 on Build GRCh38
rs536015678 Chr.16: 75229356-75229356 on Build GRCh38
rs5842 Chr.16: 75229126-75229126 on Build GRCh38
rs182556327 Chr.16: 75229272-75229272 on Build GRCh38
rs915670036 Chr.16: 75229275-75229275 on Build GRCh38
rs574215766 Chr.16: 75229080-75229080 on Build GRCh38
rs1011258034 Chr.16: 75229384-75229384 on Build GRCh38
rs192557216 Chr.16: 75229362-75229362 on Build GRCh38
rs1038390362 Chr.16: 75229330-75229330 on Build GRCh38
rs957344732 Chr.16: 75229268-75229268 on Build GRCh38
rs957171496 Chr.16: 75229140-75229140 on Build GRCh38
rs187825747 Chr.16: 75229212-75229212 on Build GRCh38
rs35971393 Chr.16: 75229216-75229216 on Build GRCh38
rs145746484 Chr.16: 75228999-75228999 on Build GRCh38
rs548730164 Chr.16: 75228991-75228991 on Build GRCh38
rs764003757 Chr.16: 75229106-75229106 on Build GRCh38
rs993760462 Chr.16: 75229011-75229011 on Build GRCh38
rs899371426 Chr.16: 75229000-75229000 on Build GRCh38
rs188022096 Chr.16: 75229350-75229350 on Build GRCh38
rs966557419 Chr.16: 75229230-75229230 on Build GRCh38
rs892767667 Chr.16: 75229373-75229373 on Build GRCh38
rs1005676495 Chr.16: 75229200-75229200 on Build GRCh38
rs975043204 Chr.16: 75228953-75228953 on Build GRCh38
rs887216600 Chr.16: 75229069-75229069 on Build GRCh38
rs1051234223 Chr.16: 75229351-75229351 on Build GRCh38
rs948537205 Chr.16: 75229162-75229162 on Build GRCh38
rs967069553 Chr.16: 75229114-75229116 on Build GRCh38
rs1042407598 Chr.16: 75228951-75228951 on Build GRCh38
rs1017712620 Chr.16: 75229202-75229202 on Build GRCh38
rs899947591 Chr.16: 75229337-75229337 on Build GRCh38
rs939132329 Chr.16: 75228969-75228969 on Build GRCh38
rs900496322 Chr.16: 75229348-75229348 on Build GRCh38
rs922312830 Chr.16: 75228963-75228963 on Build GRCh38
rs191176577 Chr.16: 75228970-75228970 on Build GRCh38
rs1017680104 Chr.16: 75229084-75229084 on Build GRCh38
rs371759406 Chr.16: 75228977-75228977 on Build GRCh38
rs906492062 Chr.16: 75229060-75229060 on Build GRCh38
rs1047558808 Chr.16: 75229066-75229066 on Build GRCh38
rs779902357 Chr.16: 75229267-75229267 on Build GRCh38
rs939322378 Chr.16: 75229172-75229172 on Build GRCh38
rs192598389 Chr.16: 75229250-75229250 on Build GRCh38
rs770131945 Chr.16: 75228967-75228967 on Build GRCh38
rs1034903189 Chr.16: 75229372-75229372 on Build GRCh38
rs148949337 Chr.16: 75229157-75229157 on Build GRCh38
rs34078065 Chr.16: 75229192-75229193 on Build GRCh38
rs186588022 Chr.16: 75229065-75229065 on Build GRCh38
rs781779698 Chr.16: 75228939-75228939 on Build GRCh38
rs544438819 Chr.16: 75229201-75229201 on Build GRCh38
rs75627144 Chr.16: 75229319-75229319 on Build GRCh38

View Full Product Details