Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 82070857-82071365 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSD17B2
Entrez Gene ID
3294
Gene Name
hydroxysteroid 17-beta dehydrogenase 2
Gene Aliases
EDH17B2, HSD17, SDR9C2
UniGene
Hs.162795
Gene Chromosome Location
Chr.16: 82035237-82098534 on Build GRCh38
Transcripts
NM_002153.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4579320 c.560A>G p.Q187R Chr.16: 82071023-82071023 on Build GRCh38
3818629 c.653G>A p.S218N Chr.16: 82071116-82071116 on Build GRCh38
1609709 c.552C>G p.D184E Chr.16: 82071015-82071015 on Build GRCh38
974263 c.657C>A p.S219R Chr.16: 82071120-82071120 on Build GRCh38
6682357 c.602A>C p.K201T Chr.16: 82071065-82071065 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372223061 Chr.16: 82070998-82070998 on Build GRCh38
rs919490086 Chr.16: 82071162-82071162 on Build GRCh38
rs758568628 Chr.16: 82070894-82070894 on Build GRCh38
rs763169865 Chr.16: 82071336-82071336 on Build GRCh38
rs767788961 Chr.16: 82071033-82071033 on Build GRCh38
rs766429944 Chr.16: 82070998-82071000 on Build GRCh38
rs113549384 Chr.16: 82071118-82071118 on Build GRCh38
rs145206359 Chr.16: 82070944-82070944 on Build GRCh38
rs747674695 Chr.16: 82071060-82071060 on Build GRCh38
rs142772496 Chr.16: 82071294-82071294 on Build GRCh38
rs199677279 Chr.16: 82071055-82071055 on Build GRCh38
rs753577590 Chr.16: 82071101-82071101 on Build GRCh38
rs772574417 Chr.16: 82071251-82071251 on Build GRCh38
rs754885156 Chr.16: 82071106-82071106 on Build GRCh38
rs766522016 Chr.16: 82071187-82071187 on Build GRCh38
rs765071943 Chr.16: 82071267-82071267 on Build GRCh38
rs865809031 Chr.16: 82071058-82071058 on Build GRCh38
rs527491978 Chr.16: 82071199-82071199 on Build GRCh38
rs757416170 Chr.16: 82071031-82071031 on Build GRCh38
rs774022930 Chr.16: 82070986-82070986 on Build GRCh38
rs137973619 Chr.16: 82071044-82071044 on Build GRCh38
rs779272545 Chr.16: 82071242-82071242 on Build GRCh38
rs201293323 Chr.16: 82071059-82071059 on Build GRCh38
rs118058400 Chr.16: 82071279-82071279 on Build GRCh38
rs749676423 Chr.16: 82071159-82071159 on Build GRCh38
rs997342495 Chr.16: 82071062-82071062 on Build GRCh38
rs563061154 Chr.16: 82071194-82071194 on Build GRCh38
rs777824285 Chr.16: 82070898-82070898 on Build GRCh38
rs774878610 Chr.16: 82071167-82071167 on Build GRCh38
rs562666235 Chr.16: 82071143-82071143 on Build GRCh38
rs777376034 Chr.16: 82071200-82071200 on Build GRCh38
rs777541430 Chr.16: 82071131-82071131 on Build GRCh38
rs762917224 Chr.16: 82071008-82071008 on Build GRCh38
rs8191169 Chr.16: 82071172-82071172 on Build GRCh38
rs571368802 Chr.16: 82071305-82071305 on Build GRCh38
rs947791305 Chr.16: 82071192-82071192 on Build GRCh38
rs376828783 Chr.16: 82071166-82071166 on Build GRCh38
rs367647331 Chr.16: 82071150-82071150 on Build GRCh38
rs752570472 Chr.16: 82071274-82071274 on Build GRCh38
rs896234001 Chr.16: 82070942-82070942 on Build GRCh38
rs748616008 Chr.16: 82071245-82071245 on Build GRCh38
rs762488702 Chr.16: 82070921-82070923 on Build GRCh38
rs916196248 Chr.16: 82071039-82071039 on Build GRCh38
rs759378846 Chr.16: 82071262-82071262 on Build GRCh38
rs753283261 Chr.16: 82071000-82071000 on Build GRCh38
rs553574089 Chr.16: 82071329-82071329 on Build GRCh38
rs763997281 Chr.16: 82071288-82071288 on Build GRCh38
rs770519395 Chr.16: 82071149-82071149 on Build GRCh38
rs144659659 Chr.16: 82071085-82071085 on Build GRCh38
rs766160176 Chr.16: 82071092-82071092 on Build GRCh38
rs756924298 Chr.16: 82071229-82071229 on Build GRCh38
rs972652153 Chr.16: 82070945-82070945 on Build GRCh38
rs368537495 Chr.16: 82070938-82070938 on Build GRCh38
rs537855379 Chr.16: 82071292-82071292 on Build GRCh38
rs750214956 Chr.16: 82071034-82071034 on Build GRCh38
rs751825809 Chr.16: 82070905-82070905 on Build GRCh38
rs115818408 Chr.16: 82071230-82071230 on Build GRCh38
rs749998417 Chr.16: 82071315-82071315 on Build GRCh38
rs749202717 Chr.16: 82071051-82071051 on Build GRCh38
rs543173162 Chr.16: 82071082-82071082 on Build GRCh38
rs777077245 Chr.16: 82070993-82070993 on Build GRCh38
rs770240144 Chr.16: 82071074-82071074 on Build GRCh38
rs777171964 Chr.16: 82071069-82071069 on Build GRCh38
rs577777799 Chr.16: 82070974-82070974 on Build GRCh38
rs756314669 Chr.16: 82071088-82071088 on Build GRCh38
rs767537488 Chr.16: 82071171-82071171 on Build GRCh38
rs769057335 Chr.16: 82071238-82071238 on Build GRCh38
rs1016342875 Chr.16: 82071314-82071314 on Build GRCh38
rs760035512 Chr.16: 82070994-82070994 on Build GRCh38
rs199780292 Chr.16: 82071075-82071075 on Build GRCh38
rs1053255620 Chr.16: 82071139-82071139 on Build GRCh38
rs769608701 Chr.16: 82070955-82070955 on Build GRCh38
rs751047349 Chr.16: 82071203-82071203 on Build GRCh38
rs764183038 Chr.16: 82071025-82071025 on Build GRCh38
rs972335755 Chr.16: 82071161-82071161 on Build GRCh38
rs532051207 Chr.16: 82071260-82071260 on Build GRCh38
rs760882182 Chr.16: 82071178-82071178 on Build GRCh38
rs773283154 Chr.16: 82071253-82071253 on Build GRCh38
rs142605504 Chr.16: 82071068-82071068 on Build GRCh38
rs867841191 Chr.16: 82070995-82070995 on Build GRCh38
rs749637589 Chr.16: 82071233-82071233 on Build GRCh38
rs751645890 Chr.16: 82071029-82071029 on Build GRCh38
rs759567766 Chr.16: 82071079-82071079 on Build GRCh38
rs779184711 Chr.16: 82071325-82071325 on Build GRCh38
rs148893933 Chr.16: 82070987-82070987 on Build GRCh38
rs757504045 Chr.16: 82070912-82070912 on Build GRCh38
rs759277446 Chr.16: 82071290-82071290 on Build GRCh38
rs965174291 Chr.16: 82070943-82070943 on Build GRCh38
rs538683936 Chr.16: 82071323-82071323 on Build GRCh38
rs751491299 Chr.16: 82071288-82071292 on Build GRCh38
rs201392108 Chr.16: 82071155-82071155 on Build GRCh38
rs113246649 Chr.16: 82071077-82071077 on Build GRCh38
rs201400632 Chr.16: 82071127-82071127 on Build GRCh38
rs759316298 Chr.16: 82071190-82071190 on Build GRCh38
rs761859772 Chr.16: 82071169-82071169 on Build GRCh38
rs918523627 Chr.16: 82070964-82070964 on Build GRCh38
rs143607337 Chr.16: 82070997-82070997 on Build GRCh38
rs747656471 Chr.16: 82071121-82071121 on Build GRCh38
rs780901902 Chr.16: 82070926-82070926 on Build GRCh38
rs779050764 Chr.16: 82071117-82071117 on Build GRCh38
rs919394756 Chr.16: 82070908-82070908 on Build GRCh38
rs760723565 Chr.16: 82071258-82071258 on Build GRCh38
rs780106425 Chr.16: 82070971-82070971 on Build GRCh38

View Full Product Details