Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 84294629-84295126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC105376777
Entrez Gene ID
105376777
Gene Name
uncharacterized LOC105376777
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.16: 84286551-84294838 on Build GRCh38
Transcripts
XM_011523512.2 NM_001282466.1 NM_001282467.1 NM_021197.3 XM_005256079.3

Gene Symbol
WFDC1
Entrez Gene ID
58189
Gene Name
WAP four-disulfide core domain 1
Gene Aliases
PS20
UniGene
Hs.36688
Gene Chromosome Location
Chr.16: 84294706-84329851 on Build GRCh38
Transcripts
XM_011523512.2 NM_001282466.1 NM_001282467.1 NM_021197.3 XM_005256079.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5955305 c.133G>A p.E45K Chr.16: 84295104-84295104 on Build GRCh38
1679310 c.56T>G p.L19R Chr.16: 84295027-84295027 on Build GRCh38
7397769 c.107A>T p.K36I Chr.16: 84295078-84295078 on Build GRCh38
4503441 c.63C>G p.L21L Chr.16: 84295034-84295034 on Build GRCh38
974383 c.73_75delCTC p.L26delL Chr.16: 84295044-84295046 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774277987 Chr.16: 84295087-84295087 on Build GRCh38
rs759423826 Chr.16: 84294955-84294955 on Build GRCh38
rs965372199 Chr.16: 84294714-84294714 on Build GRCh38
rs751406965 Chr.16: 84295029-84295029 on Build GRCh38
rs762315378 Chr.16: 84294932-84294932 on Build GRCh38
rs774211287 Chr.16: 84294858-84294858 on Build GRCh38
rs375427 Chr.16: 84294741-84294741 on Build GRCh38
rs79769516 Chr.16: 84295020-84295020 on Build GRCh38
rs757702328 Chr.16: 84294975-84294975 on Build GRCh38
rs747582762 Chr.16: 84295104-84295104 on Build GRCh38
rs147368448 Chr.16: 84294716-84294716 on Build GRCh38
rs762841944 Chr.16: 84295022-84295022 on Build GRCh38
rs960569854 Chr.16: 84294764-84294764 on Build GRCh38
rs372403601 Chr.16: 84294944-84294944 on Build GRCh38
rs945888189 Chr.16: 84294816-84294816 on Build GRCh38
rs80068047 Chr.16: 84294785-84294785 on Build GRCh38
rs942451516 Chr.16: 84294827-84294827 on Build GRCh38
rs1042838738 Chr.16: 84294818-84294818 on Build GRCh38
rs867864611 Chr.16: 84294670-84294670 on Build GRCh38
rs886650975 Chr.16: 84294848-84294848 on Build GRCh38
rs985258634 Chr.16: 84294728-84294728 on Build GRCh38
rs556809121 Chr.16: 84294865-84294865 on Build GRCh38
rs904243545 Chr.16: 84294688-84294688 on Build GRCh38
rs400345 Chr.16: 84294888-84294888 on Build GRCh38
rs761696966 Chr.16: 84294691-84294691 on Build GRCh38
rs771082930 Chr.16: 84295004-84295004 on Build GRCh38
rs891606074 Chr.16: 84294666-84294666 on Build GRCh38
rs1016792654 Chr.16: 84294732-84294732 on Build GRCh38
rs950792741 Chr.16: 84294724-84294724 on Build GRCh38
rs1026286460 Chr.16: 84294706-84294706 on Build GRCh38
rs549482902 Chr.16: 84294835-84294835 on Build GRCh38
rs1009998305 Chr.16: 84294675-84294675 on Build GRCh38
rs1056179401 Chr.16: 84294653-84294653 on Build GRCh38
rs768962826 Chr.16: 84294931-84294931 on Build GRCh38
rs144405726 Chr.16: 84294994-84294994 on Build GRCh38
rs775538104 Chr.16: 84295072-84295072 on Build GRCh38
rs575287609 Chr.16: 84294952-84294952 on Build GRCh38
rs750143557 Chr.16: 84295038-84295038 on Build GRCh38
rs746145451 Chr.16: 84295066-84295066 on Build GRCh38
rs901884627 Chr.16: 84294702-84294702 on Build GRCh38
rs573023883 Chr.16: 84294915-84294915 on Build GRCh38
rs760867274 Chr.16: 84295000-84295000 on Build GRCh38
rs764126062 Chr.16: 84295023-84295023 on Build GRCh38
rs902105611 Chr.16: 84294842-84294842 on Build GRCh38
rs1030945235 Chr.16: 84294721-84294721 on Build GRCh38
rs998231311 Chr.16: 84294830-84294830 on Build GRCh38
rs568894766 Chr.16: 84294849-84294849 on Build GRCh38
rs376201588 Chr.16: 84294826-84294826 on Build GRCh38
rs570423660 Chr.16: 84294880-84294880 on Build GRCh38
rs372307290 Chr.16: 84294711-84294711 on Build GRCh38
rs770029658 Chr.16: 84295067-84295067 on Build GRCh38
rs776842177 Chr.16: 84295065-84295065 on Build GRCh38
rs899316588 Chr.16: 84295046-84295046 on Build GRCh38
rs913119867 Chr.16: 84294815-84294815 on Build GRCh38
rs570586050 Chr.16: 84295094-84295094 on Build GRCh38
rs143040901 Chr.16: 84294987-84294987 on Build GRCh38
rs147196840 Chr.16: 84295017-84295017 on Build GRCh38
rs779142020 Chr.16: 84295044-84295046 on Build GRCh38
rs756774832 Chr.16: 84295030-84295030 on Build GRCh38
rs925503521 Chr.16: 84294824-84294824 on Build GRCh38
rs371335943 Chr.16: 84295055-84295055 on Build GRCh38
rs762831200 Chr.16: 84295080-84295080 on Build GRCh38
rs796912665 Chr.16: 84294985-84294985 on Build GRCh38
rs1039800931 Chr.16: 84294828-84294828 on Build GRCh38
rs980132035 Chr.16: 84294751-84294751 on Build GRCh38
rs758800904 Chr.16: 84295102-84295102 on Build GRCh38
rs531551784 Chr.16: 84295093-84295093 on Build GRCh38
rs148802894 Chr.16: 84294995-84294995 on Build GRCh38
rs1039163923 Chr.16: 84294840-84294840 on Build GRCh38
rs748513259 Chr.16: 84295048-84295048 on Build GRCh38
rs960509910 Chr.16: 84295010-84295010 on Build GRCh38
rs886478501 Chr.16: 84294963-84294963 on Build GRCh38
rs552890523 Chr.16: 84294889-84294889 on Build GRCh38
rs145499544 Chr.16: 84295103-84295103 on Build GRCh38
rs147313495 Chr.16: 84294921-84294922 on Build GRCh38
rs137903168 Chr.16: 84294984-84294984 on Build GRCh38
rs1047780659 Chr.16: 84294859-84294859 on Build GRCh38
rs1007833016 Chr.16: 84294681-84294681 on Build GRCh38
rs1002446551 Chr.16: 84294891-84294891 on Build GRCh38
rs200771721 Chr.16: 84295021-84295021 on Build GRCh38
rs960623379 Chr.16: 84294906-84294906 on Build GRCh38
rs960078577 Chr.16: 84294914-84294914 on Build GRCh38
rs200391042 Chr.16: 84295056-84295056 on Build GRCh38
rs114517313 Chr.16: 84295053-84295053 on Build GRCh38
rs933673781 Chr.16: 84295003-84295003 on Build GRCh38
rs766149943 Chr.16: 84295083-84295083 on Build GRCh38
rs1034791165 Chr.16: 84294911-84294911 on Build GRCh38
rs1020702985 Chr.16: 84295078-84295078 on Build GRCh38
rs763741073 Chr.16: 84295085-84295085 on Build GRCh38
rs777201368 Chr.16: 84294964-84294964 on Build GRCh38
rs752577401 Chr.16: 84294958-84294958 on Build GRCh38
rs185771367 Chr.16: 84294966-84294966 on Build GRCh38
rs927311875 Chr.16: 84294758-84294758 on Build GRCh38
rs1006521628 Chr.16: 84294707-84294707 on Build GRCh38
rs746701534 Chr.16: 84294923-84294923 on Build GRCh38
rs559359532 Chr.16: 84294703-84294703 on Build GRCh38
rs541956034 Chr.16: 84294733-84294733 on Build GRCh38
rs867137663 Chr.16: 84294747-84294747 on Build GRCh38
rs756718228 Chr.16: 84294975-84294975 on Build GRCh38
rs545700559 Chr.16: 84294660-84294660 on Build GRCh38
rs374721328 Chr.16: 84294999-84294999 on Build GRCh38
rs1057393475 Chr.16: 84294661-84294661 on Build GRCh38
rs11547593 Chr.16: 84295005-84295005 on Build GRCh38
rs541546535 Chr.16: 84294920-84294920 on Build GRCh38
rs777003630 Chr.16: 84295012-84295012 on Build GRCh38
rs769014182 Chr.16: 84294986-84294986 on Build GRCh38
rs768425473 Chr.16: 84294805-84294805 on Build GRCh38
rs758347868 Chr.16: 84294961-84294961 on Build GRCh38
rs879325034 Chr.16: 84294742-84294742 on Build GRCh38
rs200259458 Chr.16: 84294937-84294937 on Build GRCh38
rs980163028 Chr.16: 84294900-84294900 on Build GRCh38
rs755750737 Chr.16: 84295042-84295042 on Build GRCh38
rs567092432 Chr.16: 84294829-84294829 on Build GRCh38
rs886423280 Chr.16: 84294877-84294877 on Build GRCh38
rs145207818 Chr.16: 84294941-84294941 on Build GRCh38
rs765063540 Chr.16: 84294956-84294956 on Build GRCh38
rs1030600389 Chr.16: 84294846-84294846 on Build GRCh38
rs3743643 Chr.16: 84294744-84294744 on Build GRCh38
rs779195118 Chr.16: 84295047-84295047 on Build GRCh38
rs191461582 Chr.16: 84294983-84294983 on Build GRCh38
rs892217462 Chr.16: 84294659-84294659 on Build GRCh38
rs150880096 Chr.16: 84295052-84295052 on Build GRCh38
rs1042808216 Chr.16: 84294680-84294680 on Build GRCh38
rs139563386 Chr.16: 84294804-84294804 on Build GRCh38
rs770551854 Chr.16: 84294925-84294925 on Build GRCh38
rs537864631 Chr.16: 84294861-84294861 on Build GRCh38
rs766948731 Chr.16: 84295032-84295032 on Build GRCh38
rs780440065 Chr.16: 84294928-84294928 on Build GRCh38
rs564793681 Chr.16: 84294801-84294801 on Build GRCh38
rs529605456 Chr.16: 84294736-84294736 on Build GRCh38
rs1016755659 Chr.16: 84294883-84294883 on Build GRCh38
rs546885843 Chr.16: 84294821-84294821 on Build GRCh38
rs886724233 Chr.16: 84295019-84295019 on Build GRCh38
rs772109998 Chr.16: 84294933-84294933 on Build GRCh38
rs753380851 Chr.16: 84295098-84295098 on Build GRCh38
rs993200821 Chr.16: 84294919-84294919 on Build GRCh38
rs950926031 Chr.16: 84294730-84294730 on Build GRCh38
rs767071582 Chr.16: 84295092-84295092 on Build GRCh38
rs374867260 Chr.16: 84295090-84295090 on Build GRCh38

View Full Product Details