Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 3577045-3577556 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRPV1
Entrez Gene ID
7442
Gene Name
transient receptor potential cation channel subfamily V member 1
Gene Aliases
VR1
UniGene
Hs.579217
Gene Chromosome Location
Chr.17: 3565446-3609411 on Build GRCh38
Transcripts
NM_018727.5 NM_080704.3 NM_080705.3 NM_080706.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1189361 c.1741A>T p.M581L Chr.17: 3577165-3577165 on Build GRCh38
3766069 c.1714-1G>A p.? Chr.17: 3577193-3577193 on Build GRCh38
5364221 c.1800C>G p.F600L Chr.17: 3577139-3577139 on Build GRCh38
3766066 c.1753A>G p.I585V Chr.17: 3577153-3577153 on Build GRCh38
3766067 c.1786A>G p.I596V Chr.17: 3577153-3577153 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201889882 Chr.17: 3577151-3577151 on Build GRCh38
rs1048121616 Chr.17: 3577073-3577073 on Build GRCh38
rs749356670 Chr.17: 3577100-3577100 on Build GRCh38
rs917324463 Chr.17: 3577399-3577399 on Build GRCh38
rs756322247 Chr.17: 3577092-3577092 on Build GRCh38
rs538465503 Chr.17: 3577397-3577398 on Build GRCh38
rs948794639 Chr.17: 3577276-3577276 on Build GRCh38
rs531031994 Chr.17: 3577172-3577172 on Build GRCh38
rs748172067 Chr.17: 3577217-3577217 on Build GRCh38
rs201835124 Chr.17: 3577221-3577221 on Build GRCh38
rs9912962 Chr.17: 3577070-3577070 on Build GRCh38
rs200664439 Chr.17: 3577215-3577215 on Build GRCh38
rs749661523 Chr.17: 3577271-3577271 on Build GRCh38
rs750186634 Chr.17: 3577093-3577093 on Build GRCh38
rs935979665 Chr.17: 3577260-3577260 on Build GRCh38
rs200129356 Chr.17: 3577104-3577104 on Build GRCh38
rs1019367657 Chr.17: 3577380-3577380 on Build GRCh38
rs549533704 Chr.17: 3577263-3577263 on Build GRCh38
rs928756823 Chr.17: 3577496-3577496 on Build GRCh38
rs751335667 Chr.17: 3577177-3577177 on Build GRCh38
rs183832159 Chr.17: 3577117-3577117 on Build GRCh38
rs370111156 Chr.17: 3577341-3577341 on Build GRCh38
rs1045851908 Chr.17: 3577279-3577279 on Build GRCh38
rs2277679 Chr.17: 3577481-3577481 on Build GRCh38
rs756855799 Chr.17: 3577266-3577266 on Build GRCh38
rs748177038 Chr.17: 3577532-3577532 on Build GRCh38
rs375321968 Chr.17: 3577170-3577170 on Build GRCh38
rs915892234 Chr.17: 3577446-3577446 on Build GRCh38
rs898430957 Chr.17: 3577314-3577314 on Build GRCh38
rs557326176 Chr.17: 3577377-3577377 on Build GRCh38
rs771110693 Chr.17: 3577166-3577167 on Build GRCh38
rs565947084 Chr.17: 3577286-3577286 on Build GRCh38
rs549039078 Chr.17: 3577277-3577277 on Build GRCh38
rs1034540420 Chr.17: 3577391-3577391 on Build GRCh38
rs769427434 Chr.17: 3577111-3577111 on Build GRCh38
rs1030506578 Chr.17: 3577292-3577292 on Build GRCh38
rs931814776 Chr.17: 3577460-3577460 on Build GRCh38
rs1019958591 Chr.17: 3577348-3577348 on Build GRCh38
rs200893310 Chr.17: 3577234-3577234 on Build GRCh38
rs200562983 Chr.17: 3577150-3577150 on Build GRCh38
rs754907484 Chr.17: 3577202-3577202 on Build GRCh38
rs866889408 Chr.17: 3577358-3577358 on Build GRCh38
rs551055986 Chr.17: 3577255-3577255 on Build GRCh38
rs201445706 Chr.17: 3577097-3577097 on Build GRCh38
rs931216946 Chr.17: 3577515-3577515 on Build GRCh38
rs200364975 Chr.17: 3577084-3577084 on Build GRCh38
rs376378147 Chr.17: 3577144-3577144 on Build GRCh38
rs9913209 Chr.17: 3577139-3577139 on Build GRCh38
rs771788816 Chr.17: 3577200-3577200 on Build GRCh38
rs8065080 Chr.17: 3577153-3577153 on Build GRCh38
rs913492076 Chr.17: 3577208-3577208 on Build GRCh38
rs536757593 Chr.17: 3577406-3577406 on Build GRCh38
rs1049700826 Chr.17: 3577520-3577520 on Build GRCh38
rs878903061 Chr.17: 3577274-3577274 on Build GRCh38
rs758254963 Chr.17: 3577096-3577096 on Build GRCh38
rs1054396511 Chr.17: 3577261-3577261 on Build GRCh38
rs890177095 Chr.17: 3577293-3577293 on Build GRCh38
rs1037602271 Chr.17: 3577447-3577447 on Build GRCh38
rs966270482 Chr.17: 3577244-3577244 on Build GRCh38
rs202223994 Chr.17: 3577083-3577083 on Build GRCh38
rs977576629 Chr.17: 3577403-3577403 on Build GRCh38
rs369312772 Chr.17: 3577259-3577259 on Build GRCh38
rs8078936 Chr.17: 3577102-3577102 on Build GRCh38
rs1004054188 Chr.17: 3577289-3577289 on Build GRCh38
rs889499762 Chr.17: 3577371-3577371 on Build GRCh38
rs966570947 Chr.17: 3577402-3577402 on Build GRCh38
rs968992293 Chr.17: 3577378-3577379 on Build GRCh38
rs1036927997 Chr.17: 3577299-3577299 on Build GRCh38
rs78613629 Chr.17: 3577422-3577422 on Build GRCh38
rs1012722023 Chr.17: 3577085-3577085 on Build GRCh38
rs116450217 Chr.17: 3577404-3577404 on Build GRCh38
rs756584547 Chr.17: 3577125-3577125 on Build GRCh38
rs904170479 Chr.17: 3577521-3577521 on Build GRCh38
rs988464783 Chr.17: 3577166-3577166 on Build GRCh38
rs898355960 Chr.17: 3577506-3577506 on Build GRCh38
rs1036855343 Chr.17: 3577505-3577505 on Build GRCh38
rs149633009 Chr.17: 3577451-3577451 on Build GRCh38
rs767938216 Chr.17: 3577145-3577145 on Build GRCh38
rs188941003 Chr.17: 3577135-3577135 on Build GRCh38
rs745796927 Chr.17: 3577398-3577398 on Build GRCh38
rs752740898 Chr.17: 3577091-3577091 on Build GRCh38
rs200101801 Chr.17: 3577222-3577222 on Build GRCh38
rs201320275 Chr.17: 3577108-3577108 on Build GRCh38
rs993302718 Chr.17: 3577244-3577245 on Build GRCh38
rs534379633 Chr.17: 3577470-3577470 on Build GRCh38
rs939808011 Chr.17: 3577504-3577504 on Build GRCh38
rs192892777 Chr.17: 3577265-3577265 on Build GRCh38
rs200330186 Chr.17: 3577107-3577107 on Build GRCh38

View Full Product Details