Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4149894-4150397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYB5D2
Entrez Gene ID
124936
Gene Name
cytochrome b5 domain containing 2
Gene Aliases
-
UniGene
Hs.513871
Gene Chromosome Location
Chr.17: 4143117-4157701 on Build GRCh38
Transcripts
NM_001254755.1 NM_001254756.1 NM_144611.3 XM_006721447.2 XM_011523649.2 XM_017024180.1 XM_017024181.1 XM_017024182.1 XM_017024183.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5926954 c.310G>A p.D104N Chr.17: 4149950-4149950 on Build GRCh38
5470679 c.313G>A p.V105I Chr.17: 4149953-4149953 on Build GRCh38
979482 c.336G>T p.E112D Chr.17: 4149976-4149976 on Build GRCh38
979481 c.259G>A p.A87T Chr.17: 4149899-4149899 on Build GRCh38
1202772 c.341T>C p.L114P Chr.17: 4149981-4149981 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1037520830 Chr.17: 4150268-4150268 on Build GRCh38
rs111697189 Chr.17: 4150185-4150185 on Build GRCh38
rs980064513 Chr.17: 4150166-4150166 on Build GRCh38
rs891876903 Chr.17: 4150137-4150137 on Build GRCh38
rs376171076 Chr.17: 4149919-4149919 on Build GRCh38
rs201923584 Chr.17: 4149956-4149956 on Build GRCh38
rs1026656665 Chr.17: 4149966-4149966 on Build GRCh38
rs750718885 Chr.17: 4149970-4149970 on Build GRCh38
rs777828485 Chr.17: 4150341-4150342 on Build GRCh38
rs959908920 Chr.17: 4150237-4150237 on Build GRCh38
rs886368580 Chr.17: 4150249-4150249 on Build GRCh38
rs1039670665 Chr.17: 4150093-4150093 on Build GRCh38
rs775945009 Chr.17: 4150067-4150067 on Build GRCh38
rs747485700 Chr.17: 4149989-4149989 on Build GRCh38
rs1010225922 Chr.17: 4150141-4150141 on Build GRCh38
rs1030375025 Chr.17: 4150135-4150135 on Build GRCh38
rs180873288 Chr.17: 4149960-4149960 on Build GRCh38
rs373687592 Chr.17: 4150369-4150369 on Build GRCh38
rs866040883 Chr.17: 4150007-4150008 on Build GRCh38
rs758750553 Chr.17: 4149971-4149971 on Build GRCh38
rs763682141 Chr.17: 4150061-4150061 on Build GRCh38
rs748419098 Chr.17: 4150022-4150022 on Build GRCh38
rs753140324 Chr.17: 4150052-4150052 on Build GRCh38
rs866656345 Chr.17: 4150051-4150051 on Build GRCh38
rs937769230 Chr.17: 4150208-4150208 on Build GRCh38
rs769846026 Chr.17: 4150026-4150026 on Build GRCh38
rs370795820 Chr.17: 4150347-4150347 on Build GRCh38
rs1048089677 Chr.17: 4150028-4150028 on Build GRCh38
rs750056921 Chr.17: 4150075-4150075 on Build GRCh38
rs753557822 Chr.17: 4149961-4149961 on Build GRCh38
rs372881821 Chr.17: 4149953-4149953 on Build GRCh38
rs147852286 Chr.17: 4150184-4150184 on Build GRCh38
rs1016942736 Chr.17: 4150011-4150011 on Build GRCh38
rs896117776 Chr.17: 4150229-4150229 on Build GRCh38
rs918380446 Chr.17: 4150271-4150271 on Build GRCh38
rs756809009 Chr.17: 4149964-4149964 on Build GRCh38
rs1021565890 Chr.17: 4150163-4150163 on Build GRCh38
rs949073432 Chr.17: 4150236-4150236 on Build GRCh38
rs201614194 Chr.17: 4150015-4150015 on Build GRCh38
rs938492723 Chr.17: 4149963-4149963 on Build GRCh38
rs1047614729 Chr.17: 4150242-4150242 on Build GRCh38
rs968655010 Chr.17: 4149943-4149943 on Build GRCh38
rs754209157 Chr.17: 4150251-4150251 on Build GRCh38
rs568915047 Chr.17: 4150328-4150328 on Build GRCh38
rs764928105 Chr.17: 4150071-4150071 on Build GRCh38
rs144481974 Chr.17: 4150104-4150104 on Build GRCh38
rs375882168 Chr.17: 4150038-4150038 on Build GRCh38
rs998057889 Chr.17: 4150114-4150114 on Build GRCh38
rs772508937 Chr.17: 4149914-4149914 on Build GRCh38
rs765648102 Chr.17: 4149969-4149969 on Build GRCh38
rs761168731 Chr.17: 4150129-4150129 on Build GRCh38
rs757603788 Chr.17: 4150272-4150272 on Build GRCh38
rs147460942 Chr.17: 4150027-4150027 on Build GRCh38
rs186508057 Chr.17: 4150091-4150091 on Build GRCh38
rs771500743 Chr.17: 4150047-4150048 on Build GRCh38
rs776953184 Chr.17: 4149922-4149922 on Build GRCh38
rs371086906 Chr.17: 4150039-4150039 on Build GRCh38
rs532484130 Chr.17: 4150226-4150226 on Build GRCh38
rs942331280 Chr.17: 4150352-4150352 on Build GRCh38
rs775550947 Chr.17: 4149958-4149958 on Build GRCh38
rs761531164 Chr.17: 4150068-4150068 on Build GRCh38
rs537846111 Chr.17: 4150341-4150341 on Build GRCh38
rs567429780 Chr.17: 4150348-4150348 on Build GRCh38
rs755118905 Chr.17: 4149999-4149999 on Build GRCh38
rs370012754 Chr.17: 4149920-4149920 on Build GRCh38
rs770784001 Chr.17: 4149944-4149944 on Build GRCh38
rs775606148 Chr.17: 4150047-4150047 on Build GRCh38
rs1055198288 Chr.17: 4150219-4150219 on Build GRCh38
rs1028834979 Chr.17: 4150311-4150311 on Build GRCh38
rs189292607 Chr.17: 4149930-4149930 on Build GRCh38
rs780587456 Chr.17: 4149972-4149972 on Build GRCh38
rs781086989 Chr.17: 4150003-4150003 on Build GRCh38
rs552210781 Chr.17: 4150262-4150262 on Build GRCh38
rs1039294477 Chr.17: 4150358-4150358 on Build GRCh38
rs546865343 Chr.17: 4150165-4150165 on Build GRCh38
rs774234123 Chr.17: 4149952-4149952 on Build GRCh38
rs879740482 Chr.17: 4149974-4149974 on Build GRCh38
rs749069877 Chr.17: 4149997-4149997 on Build GRCh38
rs368344135 Chr.17: 4149959-4149959 on Build GRCh38

View Full Product Details