Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4950997-4951502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENO3
Entrez Gene ID
2027
Gene Name
enolase 3
Gene Aliases
GSD13, MSE
UniGene
Hs.224171
Gene Chromosome Location
Chr.17: 4949182-4957131 on Build GRCh38
Transcripts
NM_001193503.1 NM_001976.4 NM_053013.3 XM_005256521.2 XM_011523729.1 XM_017024346.1

SNP Detail

SNP ID Chromosome Location
rs527787748 Chr.17: 4951294-4951294 on Build GRCh38
rs964075731 Chr.17: 4951267-4951267 on Build GRCh38
rs570974547 Chr.17: 4951368-4951368 on Build GRCh38
rs961074425 Chr.17: 4951051-4951052 on Build GRCh38
rs919603678 Chr.17: 4951258-4951258 on Build GRCh38
rs939295289 Chr.17: 4951086-4951086 on Build GRCh38
rs955786886 Chr.17: 4951212-4951212 on Build GRCh38
rs973793543 Chr.17: 4951256-4951256 on Build GRCh38
rs555076147 Chr.17: 4951424-4951424 on Build GRCh38
rs903591293 Chr.17: 4951435-4951435 on Build GRCh38
rs998902912 Chr.17: 4951108-4951108 on Build GRCh38
rs999246590 Chr.17: 4951459-4951459 on Build GRCh38
rs550525518 Chr.17: 4951371-4951371 on Build GRCh38
rs541078523 Chr.17: 4951085-4951085 on Build GRCh38
rs974104773 Chr.17: 4951273-4951273 on Build GRCh38
rs752319724 Chr.17: 4951182-4951182 on Build GRCh38
rs555482993 Chr.17: 4951062-4951062 on Build GRCh38
rs948689419 Chr.17: 4951480-4951480 on Build GRCh38
rs907708758 Chr.17: 4951343-4951343 on Build GRCh38
rs188510021 Chr.17: 4951197-4951197 on Build GRCh38
rs563227031 Chr.17: 4951193-4951193 on Build GRCh38
rs562074451 Chr.17: 4951206-4951206 on Build GRCh38
rs183617200 Chr.17: 4951046-4951046 on Build GRCh38
rs1008490460 Chr.17: 4951231-4951231 on Build GRCh38
rs879184006 Chr.17: 4951063-4951063 on Build GRCh38
rs370707541 Chr.17: 4951058-4951059 on Build GRCh38
rs908254620 Chr.17: 4951048-4951048 on Build GRCh38
rs974142543 Chr.17: 4951060-4951060 on Build GRCh38
rs537867835 Chr.17: 4951144-4951144 on Build GRCh38
rs1056500966 Chr.17: 4951094-4951094 on Build GRCh38
rs1018652995 Chr.17: 4951246-4951246 on Build GRCh38
rs920048346 Chr.17: 4951059-4951059 on Build GRCh38
rs1043395161 Chr.17: 4951396-4951396 on Build GRCh38
rs1052521408 Chr.17: 4951461-4951461 on Build GRCh38
rs919996196 Chr.17: 4951303-4951303 on Build GRCh38
rs767888492 Chr.17: 4951131-4951131 on Build GRCh38
rs1012805249 Chr.17: 4951160-4951160 on Build GRCh38
rs572034415 Chr.17: 4951079-4951079 on Build GRCh38
rs565622777 Chr.17: 4951441-4951441 on Build GRCh38
rs1015382669 Chr.17: 4951248-4951248 on Build GRCh38
rs533552675 Chr.17: 4951370-4951370 on Build GRCh38
rs951460334 Chr.17: 4951304-4951304 on Build GRCh38
rs916517097 Chr.17: 4951387-4951387 on Build GRCh38
rs151277621 Chr.17: 4951202-4951202 on Build GRCh38
rs1026631527 Chr.17: 4951276-4951276 on Build GRCh38
rs1047618657 Chr.17: 4951071-4951071 on Build GRCh38
rs974157754 Chr.17: 4951033-4951033 on Build GRCh38
rs1043446151 Chr.17: 4951156-4951156 on Build GRCh38
rs73343373 Chr.17: 4951097-4951097 on Build GRCh38
rs910740705 Chr.17: 4951075-4951075 on Build GRCh38
rs366577 Chr.17: 4951185-4951185 on Build GRCh38
rs939226003 Chr.17: 4951349-4951349 on Build GRCh38
rs374618385 Chr.17: 4951356-4951356 on Build GRCh38
rs371775211 Chr.17: 4951335-4951335 on Build GRCh38
rs753330617 Chr.17: 4951308-4951308 on Build GRCh38
rs961410790 Chr.17: 4951253-4951253 on Build GRCh38
rs577387646 Chr.17: 4951147-4951147 on Build GRCh38
rs1054374870 Chr.17: 4951145-4951145 on Build GRCh38
rs535481050 Chr.17: 4951378-4951378 on Build GRCh38
rs773023451 Chr.17: 4951394-4951394 on Build GRCh38
rs944839837 Chr.17: 4951078-4951078 on Build GRCh38
rs1022809235 Chr.17: 4951175-4951175 on Build GRCh38
rs976650457 Chr.17: 4951374-4951374 on Build GRCh38
rs763707858 Chr.17: 4951211-4951211 on Build GRCh38
rs930868366 Chr.17: 4951066-4951066 on Build GRCh38
rs570358095 Chr.17: 4951372-4951372 on Build GRCh38

View Full Product Details