Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 13600375-13600872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HS3ST3A1
Entrez Gene ID
9955
Gene Name
heparan sulfate-glucosamine 3-sulfotransferase 3A1
Gene Aliases
3-OST-3A, 3OST3A1
UniGene
Hs.462270
Gene Chromosome Location
Chr.17: 13494032-13601942 on Build GRCh38
Transcripts
NM_006042.2 XM_017025480.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4642072 c.577G>A p.D193N Chr.17: 13600553-13600553 on Build GRCh38
4638976 c.588C>T p.L196L Chr.17: 13600542-13600542 on Build GRCh38
4617720 c.531C>T p.D177D Chr.17: 13600599-13600599 on Build GRCh38
6046309 c.325G>A p.E109K Chr.17: 13600805-13600805 on Build GRCh38
4691196 c.547G>A p.A183T Chr.17: 13600583-13600583 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758817855 Chr.17: 13600686-13600686 on Build GRCh38
rs778325635 Chr.17: 13600630-13600631 on Build GRCh38
rs949091637 Chr.17: 13600811-13600811 on Build GRCh38
rs1024554136 Chr.17: 13600533-13600533 on Build GRCh38
rs1031246644 Chr.17: 13600447-13600447 on Build GRCh38
rs767211034 Chr.17: 13600661-13600661 on Build GRCh38
rs748487626 Chr.17: 13600499-13600499 on Build GRCh38
rs374947064 Chr.17: 13600636-13600636 on Build GRCh38
rs191813363 Chr.17: 13600514-13600514 on Build GRCh38
rs769132469 Chr.17: 13600658-13600658 on Build GRCh38
rs760778867 Chr.17: 13600560-13600560 on Build GRCh38
rs775330288 Chr.17: 13600764-13600764 on Build GRCh38
rs750697453 Chr.17: 13600784-13600784 on Build GRCh38
rs939559948 Chr.17: 13600758-13600758 on Build GRCh38
rs184051619 Chr.17: 13600551-13600551 on Build GRCh38
rs1009926960 Chr.17: 13600494-13600494 on Build GRCh38
rs377607920 Chr.17: 13600483-13600483 on Build GRCh38
rs758949700 Chr.17: 13600796-13600796 on Build GRCh38
rs776419858 Chr.17: 13600617-13600617 on Build GRCh38
rs773253555 Chr.17: 13600630-13600630 on Build GRCh38
rs779719304 Chr.17: 13600482-13600482 on Build GRCh38
rs749935123 Chr.17: 13600584-13600584 on Build GRCh38
rs765096766 Chr.17: 13600581-13600581 on Build GRCh38
rs954538422 Chr.17: 13600611-13600611 on Build GRCh38
rs761560994 Chr.17: 13600622-13600622 on Build GRCh38
rs866928905 Chr.17: 13600722-13600722 on Build GRCh38
rs775731646 Chr.17: 13600554-13600554 on Build GRCh38
rs754668033 Chr.17: 13600594-13600594 on Build GRCh38
rs974917991 Chr.17: 13600595-13600595 on Build GRCh38
rs774232805 Chr.17: 13600710-13600710 on Build GRCh38
rs781347247 Chr.17: 13600711-13600711 on Build GRCh38
rs779995510 Chr.17: 13600607-13600607 on Build GRCh38
rs759075191 Chr.17: 13600754-13600754 on Build GRCh38
rs762765643 Chr.17: 13600633-13600633 on Build GRCh38
rs78372491 Chr.17: 13600562-13600562 on Build GRCh38
rs757645775 Chr.17: 13600676-13600676 on Build GRCh38
rs371109356 Chr.17: 13600525-13600525 on Build GRCh38
rs756593603 Chr.17: 13600511-13600511 on Build GRCh38
rs770713623 Chr.17: 13600727-13600727 on Build GRCh38
rs765781515 Chr.17: 13600585-13600585 on Build GRCh38
rs906713195 Chr.17: 13600403-13600403 on Build GRCh38
rs574327133 Chr.17: 13600463-13600463 on Build GRCh38
rs768809737 Chr.17: 13600613-13600613 on Build GRCh38
rs967395434 Chr.17: 13600653-13600653 on Build GRCh38
rs542119757 Chr.17: 13600820-13600820 on Build GRCh38
rs767300684 Chr.17: 13600755-13600755 on Build GRCh38
rs368514718 Chr.17: 13600517-13600517 on Build GRCh38
rs769666902 Chr.17: 13600723-13600723 on Build GRCh38
rs745893140 Chr.17: 13600540-13600540 on Build GRCh38
rs772213817 Chr.17: 13600553-13600553 on Build GRCh38
rs774000727 Chr.17: 13600745-13600745 on Build GRCh38
rs868444093 Chr.17: 13600700-13600700 on Build GRCh38
rs749302965 Chr.17: 13600726-13600726 on Build GRCh38
rs765691290 Chr.17: 13600776-13600776 on Build GRCh38
rs769643418 Chr.17: 13600629-13600629 on Build GRCh38
rs941786022 Chr.17: 13600734-13600734 on Build GRCh38
rs536651320 Chr.17: 13600477-13600477 on Build GRCh38
rs973458216 Chr.17: 13600705-13600705 on Build GRCh38
rs751663985 Chr.17: 13600815-13600815 on Build GRCh38
rs778337155 Chr.17: 13600598-13600598 on Build GRCh38
rs778871842 Chr.17: 13600532-13600532 on Build GRCh38
rs749924171 Chr.17: 13600656-13600658 on Build GRCh38
rs747027495 Chr.17: 13600694-13600694 on Build GRCh38
rs780198203 Chr.17: 13600691-13600691 on Build GRCh38
rs906618630 Chr.17: 13600748-13600748 on Build GRCh38
rs367571505 Chr.17: 13600679-13600679 on Build GRCh38
rs563386534 Chr.17: 13600610-13600610 on Build GRCh38
rs772875081 Chr.17: 13600725-13600725 on Build GRCh38
rs545942332 Chr.17: 13600592-13600592 on Build GRCh38
rs754285700 Chr.17: 13600675-13600675 on Build GRCh38
rs1003632819 Chr.17: 13600415-13600415 on Build GRCh38
rs766759088 Chr.17: 13600801-13600801 on Build GRCh38
rs892744857 Chr.17: 13600802-13600802 on Build GRCh38
rs751016209 Chr.17: 13600644-13600644 on Build GRCh38
rs772235059 Chr.17: 13600603-13600603 on Build GRCh38
rs779434343 Chr.17: 13600600-13600600 on Build GRCh38
rs755227727 Chr.17: 13600708-13600708 on Build GRCh38
rs760240531 Chr.17: 13600772-13600772 on Build GRCh38
rs745954983 Chr.17: 13600602-13600602 on Build GRCh38
rs142627880 Chr.17: 13600456-13600456 on Build GRCh38
rs941664740 Chr.17: 13600481-13600481 on Build GRCh38
rs74954568 Chr.17: 13600565-13600565 on Build GRCh38
rs573090724 Chr.17: 13600782-13600782 on Build GRCh38
rs368104034 Chr.17: 13600599-13600599 on Build GRCh38
rs761755593 Chr.17: 13600580-13600580 on Build GRCh38
rs748352232 Chr.17: 13600715-13600715 on Build GRCh38
rs866889757 Chr.17: 13600688-13600688 on Build GRCh38
rs541122669 Chr.17: 13600641-13600641 on Build GRCh38
rs1045099767 Chr.17: 13600401-13600401 on Build GRCh38
rs887530845 Chr.17: 13600462-13600462 on Build GRCh38

View Full Product Details