Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 31162904-31163411 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NF1
Entrez Gene ID
4763
Gene Name
neurofibromin 1
Gene Aliases
NFNS, VRNF, WSS
UniGene
Hs.113577
Gene Chromosome Location
Chr.17: 31094927-31377677 on Build GRCh38
Transcripts
NM_000267.3 NM_001042492.2 NM_001128147.2 XM_005257983.2 XM_005257984.2 XM_006721922.2 XM_006721924.2 XM_006721925.2 XM_006721926.3 XM_011524852.2 XM_011524857.2 XM_017024689.1 XM_017024690.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6933179 c.354C>A p.C118* Chr.17: 31163251-31163251 on Build GRCh38
7404174 c.416C>G p.S139C Chr.17: 31163313-31163313 on Build GRCh38
5049795 c.451A>C p.N151H Chr.17: 31163348-31163348 on Build GRCh38
6923690 c.331A>T p.K111* Chr.17: 31163228-31163228 on Build GRCh38
5944636 c.474_475insT p.T159fs*15 Chr.17: 31163371-31163372 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs58470207 Chr.17: 31163160-31163160 on Build GRCh38
rs769391944 Chr.17: 31163348-31163348 on Build GRCh38
rs144624699 Chr.17: 31163088-31163088 on Build GRCh38
rs876658752 Chr.17: 31163379-31163379 on Build GRCh38
rs112860408 Chr.17: 31163233-31163233 on Build GRCh38
rs876659418 Chr.17: 31163270-31163270 on Build GRCh38
rs587781757 Chr.17: 31163264-31163264 on Build GRCh38
rs146691765 Chr.17: 31163266-31163266 on Build GRCh38
rs878853886 Chr.17: 31163265-31163265 on Build GRCh38
rs937049509 Chr.17: 31163030-31163030 on Build GRCh38
rs768777585 Chr.17: 31163251-31163251 on Build GRCh38
rs377481833 Chr.17: 31163353-31163353 on Build GRCh38
rs757900586 Chr.17: 31163162-31163162 on Build GRCh38
rs952927289 Chr.17: 31163132-31163132 on Build GRCh38
rs970736690 Chr.17: 31163047-31163047 on Build GRCh38
rs981248655 Chr.17: 31162967-31162967 on Build GRCh38
rs1000597075 Chr.17: 31162928-31162928 on Build GRCh38
rs786203460 Chr.17: 31163330-31163330 on Build GRCh38
rs869196514 Chr.17: 31163324-31163324 on Build GRCh38
rs897354511 Chr.17: 31163085-31163085 on Build GRCh38
rs780233935 Chr.17: 31163220-31163220 on Build GRCh38
rs199474731 Chr.17: 31163247-31163247 on Build GRCh38
rs876658428 Chr.17: 31163194-31163194 on Build GRCh38
rs775191883 Chr.17: 31163300-31163300 on Build GRCh38
rs774909376 Chr.17: 31163137-31163137 on Build GRCh38
rs150080742 Chr.17: 31163253-31163253 on Build GRCh38
rs755890242 Chr.17: 31163363-31163363 on Build GRCh38
rs199474744 Chr.17: 31163367-31163367 on Build GRCh38
rs941595623 Chr.17: 31163062-31163062 on Build GRCh38
rs199474734 Chr.17: 31163331-31163331 on Build GRCh38
rs750974024 Chr.17: 31163342-31163342 on Build GRCh38
rs1012141454 Chr.17: 31163044-31163044 on Build GRCh38
rs750268216 Chr.17: 31163359-31163359 on Build GRCh38
rs778398666 Chr.17: 31163383-31163383 on Build GRCh38
rs786201914 Chr.17: 31163275-31163275 on Build GRCh38
rs759286941 Chr.17: 31163149-31163149 on Build GRCh38
rs773657744 Chr.17: 31163146-31163146 on Build GRCh38
rs761086521 Chr.17: 31163290-31163290 on Build GRCh38
rs918556790 Chr.17: 31163100-31163100 on Build GRCh38
rs984194433 Chr.17: 31163152-31163152 on Build GRCh38
rs751506868 Chr.17: 31163179-31163179 on Build GRCh38
rs922854899 Chr.17: 31163227-31163227 on Build GRCh38
rs7207410 Chr.17: 31163237-31163237 on Build GRCh38
rs533527555 Chr.17: 31162961-31162961 on Build GRCh38
rs757074803 Chr.17: 31163180-31163180 on Build GRCh38
rs749115431 Chr.17: 31163007-31163007 on Build GRCh38
rs763991276 Chr.17: 31163166-31163166 on Build GRCh38
rs786201731 Chr.17: 31163218-31163218 on Build GRCh38
rs769615182 Chr.17: 31163291-31163291 on Build GRCh38
rs751482887 Chr.17: 31163039-31163039 on Build GRCh38
rs764946966 Chr.17: 31163157-31163157 on Build GRCh38
rs1038627862 Chr.17: 31163079-31163079 on Build GRCh38
rs754096545 Chr.17: 31163364-31163364 on Build GRCh38
rs1051210268 Chr.17: 31163100-31163101 on Build GRCh38
rs138952888 Chr.17: 31163263-31163263 on Build GRCh38
rs779074793 Chr.17: 31163365-31163365 on Build GRCh38
rs371192107 Chr.17: 31163372-31163372 on Build GRCh38
rs199474752 Chr.17: 31163376-31163376 on Build GRCh38
rs869287312 Chr.17: 31163280-31163280 on Build GRCh38
rs768002347 Chr.17: 31163143-31163143 on Build GRCh38
rs763663904 Chr.17: 31163337-31163337 on Build GRCh38
rs587782107 Chr.17: 31163261-31163261 on Build GRCh38
rs149003051 Chr.17: 31163271-31163271 on Build GRCh38
rs780979519 Chr.17: 31163190-31163190 on Build GRCh38
rs202142624 Chr.17: 31163347-31163347 on Build GRCh38
rs752292696 Chr.17: 31163158-31163158 on Build GRCh38
rs928360284 Chr.17: 31163029-31163029 on Build GRCh38
rs762152769 Chr.17: 31163138-31163138 on Build GRCh38
rs762434870 Chr.17: 31163314-31163314 on Build GRCh38
rs572990602 Chr.17: 31162947-31162947 on Build GRCh38
rs567828963 Chr.17: 31163134-31163134 on Build GRCh38
rs756370324 Chr.17: 31163201-31163201 on Build GRCh38
rs563422566 Chr.17: 31162978-31162978 on Build GRCh38
rs876659818 Chr.17: 31163332-31163332 on Build GRCh38
rs149027856 Chr.17: 31163045-31163045 on Build GRCh38
rs876660277 Chr.17: 31163286-31163286 on Build GRCh38
rs745611945 Chr.17: 31163193-31163193 on Build GRCh38
rs878853891 Chr.17: 31163294-31163294 on Build GRCh38

View Full Product Details