Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18977494-18977997 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM83G
Entrez Gene ID
644815
Gene Name
family with sequence similarity 83 member G
Gene Aliases
PAWS1
UniGene
Hs.710727
Gene Chromosome Location
Chr.17: 18968789-19006144 on Build GRCh38
Transcripts
NM_001039999.2 XM_017024953.1 NM_001042450.2 NM_001270648.1 NM_001270649.1 NM_001282417.1 NM_152351.4 XM_011523652.1 XM_011523653.2 XM_017024191.1

Gene Symbol
SLC5A10
Entrez Gene ID
125206
Gene Name
solute carrier family 5 member 10
Gene Aliases
SGLT-5, SGLT5
UniGene
Hs.462418
Gene Chromosome Location
Chr.17: 18950676-19020691 on Build GRCh38
Transcripts
NM_001039999.2 XM_017024953.1 NM_001042450.2 NM_001270648.1 NM_001270649.1 NM_001282417.1 NM_152351.4 XM_011523652.1 XM_011523653.2 XM_017024191.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5587781 c.1851G>A p.E617E Chr.17: 18977815-18977815 on Build GRCh38
1609930 c.1895G>A p.R632H Chr.17: 18977771-18977771 on Build GRCh38
6839297 c.2056C>T p.Q686* Chr.17: 18977610-18977610 on Build GRCh38
6335116 c.1917C>A p.N639K Chr.17: 18977749-18977749 on Build GRCh38
5620790 c.1883C>T p.P628L Chr.17: 18977783-18977783 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753077917 Chr.17: 18977846-18977847 on Build GRCh38
rs1049792809 Chr.17: 18977615-18977615 on Build GRCh38
rs752126627 Chr.17: 18977731-18977731 on Build GRCh38
rs1052398432 Chr.17: 18977785-18977785 on Build GRCh38
rs371125554 Chr.17: 18977749-18977749 on Build GRCh38
rs972068048 Chr.17: 18977941-18977941 on Build GRCh38
rs75425060 Chr.17: 18977673-18977673 on Build GRCh38
rs777664075 Chr.17: 18977946-18977946 on Build GRCh38
rs756232582 Chr.17: 18977912-18977914 on Build GRCh38
rs775468730 Chr.17: 18977964-18977964 on Build GRCh38
rs752034805 Chr.17: 18977805-18977805 on Build GRCh38
rs749383561 Chr.17: 18977927-18977935 on Build GRCh38
rs374398726 Chr.17: 18977651-18977651 on Build GRCh38
rs769892325 Chr.17: 18977960-18977960 on Build GRCh38
rs182384089 Chr.17: 18977630-18977630 on Build GRCh38
rs752937192 Chr.17: 18977782-18977782 on Build GRCh38
rs758976292 Chr.17: 18977660-18977660 on Build GRCh38
rs773273233 Chr.17: 18977764-18977764 on Build GRCh38
rs568000157 Chr.17: 18977665-18977665 on Build GRCh38
rs540858760 Chr.17: 18977911-18977911 on Build GRCh38
rs372612886 Chr.17: 18977617-18977617 on Build GRCh38
rs572824194 Chr.17: 18977748-18977748 on Build GRCh38
rs752365106 Chr.17: 18977942-18977942 on Build GRCh38
rs779496696 Chr.17: 18977806-18977806 on Build GRCh38
rs1004418613 Chr.17: 18977884-18977884 on Build GRCh38
rs201668455 Chr.17: 18977727-18977727 on Build GRCh38
rs905241832 Chr.17: 18977971-18977971 on Build GRCh38
rs1014037928 Chr.17: 18977796-18977796 on Build GRCh38
rs1009729866 Chr.17: 18977661-18977661 on Build GRCh38
rs1045483953 Chr.17: 18977962-18977962 on Build GRCh38
rs750648871 Chr.17: 18977556-18977556 on Build GRCh38
rs759629345 Chr.17: 18977777-18977777 on Build GRCh38
rs759663540 Chr.17: 18977883-18977883 on Build GRCh38
rs755450195 Chr.17: 18977584-18977584 on Build GRCh38
rs201172910 Chr.17: 18977722-18977722 on Build GRCh38
rs924496501 Chr.17: 18977779-18977779 on Build GRCh38
rs965059488 Chr.17: 18977888-18977888 on Build GRCh38
rs745395779 Chr.17: 18977667-18977667 on Build GRCh38
rs756438694 Chr.17: 18977557-18977557 on Build GRCh38
rs376972051 Chr.17: 18977623-18977623 on Build GRCh38
rs186697015 Chr.17: 18977959-18977959 on Build GRCh38
rs768311912 Chr.17: 18977699-18977699 on Build GRCh38
rs769831705 Chr.17: 18977885-18977885 on Build GRCh38
rs748573813 Chr.17: 18977658-18977658 on Build GRCh38
rs577449943 Chr.17: 18977861-18977861 on Build GRCh38
rs374490985 Chr.17: 18977771-18977771 on Build GRCh38
rs748195382 Chr.17: 18977552-18977552 on Build GRCh38
rs1027117583 Chr.17: 18977847-18977847 on Build GRCh38
rs772271859 Chr.17: 18977886-18977886 on Build GRCh38
rs775115642 Chr.17: 18977683-18977683 on Build GRCh38
rs200348726 Chr.17: 18977912-18977912 on Build GRCh38
rs746816131 Chr.17: 18977951-18977951 on Build GRCh38
rs368737975 Chr.17: 18977908-18977908 on Build GRCh38
rs201046878 Chr.17: 18977778-18977778 on Build GRCh38
rs972543410 Chr.17: 18977770-18977770 on Build GRCh38
rs753405420 Chr.17: 18977824-18977824 on Build GRCh38
rs766625194 Chr.17: 18977719-18977719 on Build GRCh38
rs771971752 Chr.17: 18977858-18977858 on Build GRCh38
rs747982022 Chr.17: 18977753-18977753 on Build GRCh38
rs749469051 Chr.17: 18977970-18977970 on Build GRCh38
rs371029426 Chr.17: 18977659-18977659 on Build GRCh38
rs551954434 Chr.17: 18977596-18977596 on Build GRCh38
rs761609450 Chr.17: 18977701-18977701 on Build GRCh38
rs780520275 Chr.17: 18977571-18977571 on Build GRCh38
rs761856215 Chr.17: 18977621-18977621 on Build GRCh38
rs201854204 Chr.17: 18977544-18977544 on Build GRCh38
rs532245695 Chr.17: 18977588-18977588 on Build GRCh38
rs775207503 Chr.17: 18977607-18977607 on Build GRCh38
rs968031888 Chr.17: 18977521-18977521 on Build GRCh38
rs911984177 Chr.17: 18977574-18977574 on Build GRCh38
rs1039903718 Chr.17: 18977575-18977575 on Build GRCh38
rs758772623 Chr.17: 18977729-18977729 on Build GRCh38
rs375307645 Chr.17: 18977610-18977610 on Build GRCh38
rs896136965 Chr.17: 18977851-18977851 on Build GRCh38
rs766692916 Chr.17: 18977741-18977741 on Build GRCh38
rs771017043 Chr.17: 18977866-18977866 on Build GRCh38
rs781389324 Chr.17: 18977555-18977555 on Build GRCh38
rs896530223 Chr.17: 18977648-18977648 on Build GRCh38
rs776514603 Chr.17: 18977876-18977876 on Build GRCh38
rs376311708 Chr.17: 18977899-18977899 on Build GRCh38
rs529981513 Chr.17: 18977916-18977916 on Build GRCh38
rs775766698 Chr.17: 18977896-18977896 on Build GRCh38
rs764496911 Chr.17: 18977794-18977794 on Build GRCh38
rs1050345347 Chr.17: 18977564-18977564 on Build GRCh38
rs779285096 Chr.17: 18977586-18977586 on Build GRCh38
rs755345347 Chr.17: 18977556-18977556 on Build GRCh38
rs377667225 Chr.17: 18977652-18977652 on Build GRCh38
rs905300005 Chr.17: 18977836-18977836 on Build GRCh38
rs968273750 Chr.17: 18977669-18977669 on Build GRCh38
rs369485861 Chr.17: 18977581-18977581 on Build GRCh38
rs34552844 Chr.17: 18977745-18977746 on Build GRCh38
rs368202632 Chr.17: 18977900-18977900 on Build GRCh38
rs751950262 Chr.17: 18977904-18977904 on Build GRCh38
rs372021645 Chr.17: 18977846-18977846 on Build GRCh38
rs1056183148 Chr.17: 18977847-18977848 on Build GRCh38
rs772063783 Chr.17: 18977754-18977754 on Build GRCh38
rs750961572 Chr.17: 18977914-18977914 on Build GRCh38
rs146085023 Chr.17: 18977551-18977551 on Build GRCh38
rs758156745 Chr.17: 18977945-18977945 on Build GRCh38
rs763298260 Chr.17: 18977784-18977784 on Build GRCh38
rs200038747 Chr.17: 18977726-18977726 on Build GRCh38
rs367677167 Chr.17: 18977736-18977736 on Build GRCh38
rs571835978 Chr.17: 18977629-18977629 on Build GRCh38
rs771103367 Chr.17: 18977772-18977772 on Build GRCh38
rs372199593 Chr.17: 18977606-18977606 on Build GRCh38
rs749024147 Chr.17: 18977692-18977692 on Build GRCh38
rs745481489 Chr.17: 18977603-18977603 on Build GRCh38
rs930936505 Chr.17: 18977614-18977614 on Build GRCh38
rs377259741 Chr.17: 18977958-18977958 on Build GRCh38
rs913718591 Chr.17: 18977518-18977518 on Build GRCh38
rs746904439 Chr.17: 18977865-18977865 on Build GRCh38
rs767578750 Chr.17: 18977704-18977704 on Build GRCh38
rs987981654 Chr.17: 18977515-18977515 on Build GRCh38
rs373827776 Chr.17: 18977732-18977732 on Build GRCh38
rs766717165 Chr.17: 18977645-18977645 on Build GRCh38
rs768167872 Chr.17: 18977708-18977709 on Build GRCh38
rs201669881 Chr.17: 18977730-18977730 on Build GRCh38
rs199864492 Chr.17: 18977537-18977537 on Build GRCh38
rs867328100 Chr.17: 18977966-18977966 on Build GRCh38
rs773363329 Chr.17: 18977714-18977714 on Build GRCh38
rs754522335 Chr.17: 18977843-18977843 on Build GRCh38
rs777773357 Chr.17: 18977918-18977920 on Build GRCh38
rs764706502 Chr.17: 18977918-18977918 on Build GRCh38
rs376662345 Chr.17: 18977905-18977905 on Build GRCh38
rs747816872 Chr.17: 18977849-18977849 on Build GRCh38
rs369334119 Chr.17: 18977611-18977611 on Build GRCh38
rs767444753 Chr.17: 18977700-18977700 on Build GRCh38

View Full Product Details