Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 21300256-21300754 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP2K3
Entrez Gene ID
5606
Gene Name
mitogen-activated protein kinase kinase 3
Gene Aliases
MAPKK3, MEK3, MKK3, PRKMK3, SAPKK-2, SAPKK2
UniGene
Hs.514012
Gene Chromosome Location
Chr.17: 21284656-21315240 on Build GRCh38
Transcripts
NM_001316332.1 NM_002756.4 NM_145109.2 XM_005256723.2 XM_011523958.2 XM_011523959.1 XM_017024857.1 XM_017024858.1 XM_017024859.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
976585 c.271G>A p.G91S Chr.17: 21300638-21300638 on Build GRCh38
7376209 c.169A>G p.S57G Chr.17: 21300635-21300635 on Build GRCh38
976587 c.272G>A p.G91D Chr.17: 21300639-21300639 on Build GRCh38
7294282 c.161A>G p.H54R Chr.17: 21300627-21300627 on Build GRCh38
384620 c.281T>A p.M94K Chr.17: 21300648-21300648 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762956297 Chr.17: 21300698-21300698 on Build GRCh38
rs886807903 Chr.17: 21300610-21300610 on Build GRCh38
rs928596251 Chr.17: 21300362-21300362 on Build GRCh38
rs904961483 Chr.17: 21300317-21300317 on Build GRCh38
rs922094933 Chr.17: 21300422-21300422 on Build GRCh38
rs983743374 Chr.17: 21300387-21300387 on Build GRCh38
rs200914153 Chr.17: 21300593-21300593 on Build GRCh38
rs751127330 Chr.17: 21300499-21300499 on Build GRCh38
rs62057673 Chr.17: 21300637-21300637 on Build GRCh38
rs771553046 Chr.17: 21300495-21300495 on Build GRCh38
rs1007548145 Chr.17: 21300553-21300553 on Build GRCh38
rs935323804 Chr.17: 21300473-21300473 on Build GRCh38
rs1026682173 Chr.17: 21300367-21300367 on Build GRCh38
rs67627398 Chr.17: 21300363-21300363 on Build GRCh38
rs78304829 Chr.17: 21300508-21300508 on Build GRCh38
rs62057672 Chr.17: 21300622-21300622 on Build GRCh38
rs540081640 Chr.17: 21300363-21300363 on Build GRCh38
rs1029598403 Chr.17: 21300635-21300635 on Build GRCh38
rs764171001 Chr.17: 21300701-21300701 on Build GRCh38
rs995228259 Chr.17: 21300350-21300350 on Build GRCh38
rs1023017873 Chr.17: 21300718-21300718 on Build GRCh38
rs775190639 Chr.17: 21300496-21300496 on Build GRCh38
rs772854520 Chr.17: 21300653-21300653 on Build GRCh38
rs753946415 Chr.17: 21300506-21300506 on Build GRCh38
rs770333840 Chr.17: 21300607-21300607 on Build GRCh38
rs548881323 Chr.17: 21300704-21300704 on Build GRCh38
rs774060386 Chr.17: 21300537-21300537 on Build GRCh38
rs776457114 Chr.17: 21300668-21300668 on Build GRCh38
rs767174366 Chr.17: 21300554-21300554 on Build GRCh38
rs753716912 Chr.17: 21300573-21300573 on Build GRCh38
rs763540937 Chr.17: 21300673-21300677 on Build GRCh38
rs73982727 Chr.17: 21300316-21300316 on Build GRCh38
rs764245258 Chr.17: 21300632-21300632 on Build GRCh38
rs747410898 Chr.17: 21300594-21300594 on Build GRCh38
rs1052350209 Chr.17: 21300491-21300491 on Build GRCh38
rs949358606 Chr.17: 21300710-21300710 on Build GRCh38
rs756090989 Chr.17: 21300504-21300504 on Build GRCh38
rs929499130 Chr.17: 21300471-21300471 on Build GRCh38
rs1044972618 Chr.17: 21300725-21300725 on Build GRCh38
rs62057674 Chr.17: 21300686-21300686 on Build GRCh38
rs932086547 Chr.17: 21300425-21300425 on Build GRCh38
rs746731299 Chr.17: 21300654-21300654 on Build GRCh38
rs73311537 Chr.17: 21300399-21300399 on Build GRCh38
rs779932722 Chr.17: 21300505-21300505 on Build GRCh38
rs565034825 Chr.17: 21300380-21300380 on Build GRCh38
rs991584835 Chr.17: 21300403-21300403 on Build GRCh38
rs759527241 Chr.17: 21300624-21300624 on Build GRCh38
rs967193688 Chr.17: 21300334-21300334 on Build GRCh38
rs1021849295 Chr.17: 21300292-21300292 on Build GRCh38
rs777238559 Chr.17: 21300516-21300516 on Build GRCh38
rs770555695 Chr.17: 21300660-21300660 on Build GRCh38
rs767154440 Chr.17: 21300502-21300502 on Build GRCh38
rs766095224 Chr.17: 21300623-21300623 on Build GRCh38
rs761686241 Chr.17: 21300501-21300501 on Build GRCh38
rs768227193 Chr.17: 21300535-21300535 on Build GRCh38
rs750230425 Chr.17: 21300503-21300503 on Build GRCh38
rs765200173 Chr.17: 21300625-21300625 on Build GRCh38
rs919385010 Chr.17: 21300433-21300433 on Build GRCh38
rs530302955 Chr.17: 21300634-21300634 on Build GRCh38
rs79762169 Chr.17: 21300361-21300361 on Build GRCh38
rs75854955 Chr.17: 21300513-21300513 on Build GRCh38
rs773794838 Chr.17: 21300612-21300612 on Build GRCh38
rs71207220 Chr.17: 21300377-21300377 on Build GRCh38
rs73311539 Chr.17: 21300595-21300595 on Build GRCh38
rs771651753 Chr.17: 21300619-21300619 on Build GRCh38
rs73982728 Chr.17: 21300329-21300329 on Build GRCh38
rs36076766 Chr.17: 21300649-21300649 on Build GRCh38
rs780568273 Chr.17: 21300651-21300651 on Build GRCh38
rs752806793 Chr.17: 21300591-21300591 on Build GRCh38
rs757705153 Chr.17: 21300636-21300636 on Build GRCh38
rs759346339 Chr.17: 21300684-21300684 on Build GRCh38
rs746313676 Chr.17: 21300605-21300605 on Build GRCh38
rs1011375302 Chr.17: 21300566-21300566 on Build GRCh38
rs760418674 Chr.17: 21300564-21300564 on Build GRCh38
rs768744839 Chr.17: 21300362-21300363 on Build GRCh38
rs72831559 Chr.17: 21300279-21300279 on Build GRCh38
rs979468173 Chr.17: 21300426-21300426 on Build GRCh38
rs747661001 Chr.17: 21300616-21300616 on Build GRCh38
rs747357793 Chr.17: 21300511-21300511 on Build GRCh38
rs953543926 Chr.17: 21300643-21300643 on Build GRCh38
rs764383139 Chr.17: 21300726-21300726 on Build GRCh38
rs750222581 Chr.17: 21300559-21300559 on Build GRCh38
rs138579071 Chr.17: 21300451-21300451 on Build GRCh38
rs925323798 Chr.17: 21300446-21300446 on Build GRCh38
rs1011010287 Chr.17: 21300288-21300288 on Build GRCh38
rs72831560 Chr.17: 21300289-21300289 on Build GRCh38
rs766322527 Chr.17: 21300568-21300568 on Build GRCh38
rs762578195 Chr.17: 21300522-21300522 on Build GRCh38
rs775449654 Chr.17: 21300695-21300695 on Build GRCh38
rs751688422 Chr.17: 21300703-21300703 on Build GRCh38
rs113621919 Chr.17: 21300443-21300443 on Build GRCh38
rs772332862 Chr.17: 21300509-21300509 on Build GRCh38
rs386796127 Chr.17: 21300361-21300363 on Build GRCh38
rs34105301 Chr.17: 21300581-21300581 on Build GRCh38
rs532510198 Chr.17: 21300406-21300406 on Build GRCh38
rs766582251 Chr.17: 21300710-21300711 on Build GRCh38
rs746225381 Chr.17: 21300645-21300645 on Build GRCh38
rs761441750 Chr.17: 21300538-21300538 on Build GRCh38
rs755054165 Chr.17: 21300576-21300576 on Build GRCh38
rs2305873 Chr.17: 21300629-21300629 on Build GRCh38
rs34458870 Chr.17: 21300652-21300652 on Build GRCh38
rs762521595 Chr.17: 21300498-21300498 on Build GRCh38
rs1023427113 Chr.17: 21300314-21300314 on Build GRCh38
rs200944510 Chr.17: 21300628-21300628 on Build GRCh38
rs781769054 Chr.17: 21300603-21300603 on Build GRCh38
rs915595664 Chr.17: 21300404-21300404 on Build GRCh38
rs976660148 Chr.17: 21300414-21300414 on Build GRCh38

View Full Product Details