Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 21408553-21409056 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNJ12
Entrez Gene ID
3768
Gene Name
potassium voltage-gated channel subfamily J member 12
Gene Aliases
IRK-2, IRK2, KCNJN1, Kir2.2, Kir2.2v, hIRK, hIRK1, hkir2.2x, kcnj12x
UniGene
Hs.200629
Gene Chromosome Location
Chr.17: 21376387-21419870 on Build GRCh38
Transcripts
NM_021012.4 XM_005256625.4 XM_011523831.2

SNP Detail

SNP ID Chromosome Location
rs112839544 Chr.17: 21408583-21408583 on Build GRCh38
rs545057421 Chr.17: 21408957-21408957 on Build GRCh38
rs9911201 Chr.17: 21408901-21408901 on Build GRCh38
rs1657716 Chr.17: 21408604-21408604 on Build GRCh38
rs578067533 Chr.17: 21408823-21408823 on Build GRCh38
rs9908707 Chr.17: 21408599-21408599 on Build GRCh38
rs9911150 Chr.17: 21408573-21408573 on Build GRCh38
rs55869783 Chr.17: 21408977-21408977 on Build GRCh38
rs553187228 Chr.17: 21408722-21408722 on Build GRCh38
rs12944347 Chr.17: 21409005-21409005 on Build GRCh38
rs1714903 Chr.17: 21408628-21408628 on Build GRCh38
rs9911228 Chr.17: 21408954-21408954 on Build GRCh38
rs377548268 Chr.17: 21408689-21408689 on Build GRCh38
rs1714901 Chr.17: 21408933-21408933 on Build GRCh38
rs386796201 Chr.17: 21408603-21408605 on Build GRCh38
rs573839352 Chr.17: 21408603-21408603 on Build GRCh38
rs111874890 Chr.17: 21408734-21408734 on Build GRCh38
rs1714904 Chr.17: 21408620-21408620 on Build GRCh38
rs1714902 Chr.17: 21408921-21408921 on Build GRCh38
rs555701892 Chr.17: 21408592-21408592 on Build GRCh38
rs12946212 Chr.17: 21408988-21408988 on Build GRCh38

View Full Product Details