Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 21417140-21417635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNJ12
Entrez Gene ID
3768
Gene Name
potassium voltage-gated channel subfamily J member 12
Gene Aliases
IRK-2, IRK2, KCNJN1, Kir2.2, Kir2.2v, hIRK, hIRK1, hkir2.2x, kcnj12x
UniGene
Hs.200629
Gene Chromosome Location
Chr.17: 21376387-21419870 on Build GRCh38
Transcripts
NM_021012.4 XM_005256625.4 XM_011523831.2

SNP Detail

SNP ID Chromosome Location
rs76498462 Chr.17: 21417514-21417514 on Build GRCh38
rs80177580 Chr.17: 21417266-21417266 on Build GRCh38
rs34388140 Chr.17: 21417540-21417540 on Build GRCh38
rs386796208 Chr.17: 21417283-21417285 on Build GRCh38
rs72842113 Chr.17: 21417387-21417387 on Build GRCh38
rs77686314 Chr.17: 21417322-21417322 on Build GRCh38
rs73981234 Chr.17: 21417486-21417486 on Build GRCh38
rs551866283 Chr.17: 21417180-21417180 on Build GRCh38
rs73313941 Chr.17: 21417187-21417187 on Build GRCh38
rs113091387 Chr.17: 21417412-21417412 on Build GRCh38
rs72846675 Chr.17: 21417283-21417283 on Build GRCh38
rs570101403 Chr.17: 21417194-21417194 on Build GRCh38
rs75614171 Chr.17: 21417347-21417347 on Build GRCh38
rs67230821 Chr.17: 21417542-21417542 on Build GRCh38
rs67680166 Chr.17: 21417217-21417217 on Build GRCh38
rs78847182 Chr.17: 21417274-21417274 on Build GRCh38
rs78556043 Chr.17: 21417369-21417369 on Build GRCh38
rs553904312 Chr.17: 21417471-21417471 on Build GRCh38
rs73981232 Chr.17: 21417346-21417346 on Build GRCh38
rs572150064 Chr.17: 21417598-21417598 on Build GRCh38
rs556160072 Chr.17: 21417207-21417207 on Build GRCh38
rs72842109 Chr.17: 21417284-21417284 on Build GRCh38
rs72842114 Chr.17: 21417570-21417570 on Build GRCh38
rs537492715 Chr.17: 21417202-21417202 on Build GRCh38
rs35676232 Chr.17: 21417584-21417584 on Build GRCh38
rs76059352 Chr.17: 21417543-21417543 on Build GRCh38
rs72846673 Chr.17: 21417243-21417243 on Build GRCh38
rs781926868 Chr.17: 21417512-21417512 on Build GRCh38
rs80028685 Chr.17: 21417578-21417578 on Build GRCh38
rs75211783 Chr.17: 21417293-21417293 on Build GRCh38
rs72842111 Chr.17: 21417363-21417363 on Build GRCh38
rs76852423 Chr.17: 21417555-21417555 on Build GRCh38
rs574707037 Chr.17: 21417250-21417250 on Build GRCh38
rs73313944 Chr.17: 21417604-21417604 on Build GRCh38
rs76283231 Chr.17: 21417378-21417378 on Build GRCh38
rs72846674 Chr.17: 21417244-21417244 on Build GRCh38
rs76105604 Chr.17: 21417475-21417475 on Build GRCh38
rs76953980 Chr.17: 21417233-21417233 on Build GRCh38
rs73981233 Chr.17: 21417484-21417484 on Build GRCh38
rs74883261 Chr.17: 21417399-21417399 on Build GRCh38
rs74336563 Chr.17: 21417168-21417168 on Build GRCh38

View Full Product Details