Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 21419019-21419524 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNJ12
Entrez Gene ID
3768
Gene Name
potassium voltage-gated channel subfamily J member 12
Gene Aliases
IRK-2, IRK2, KCNJN1, Kir2.2, Kir2.2v, hIRK, hIRK1, hkir2.2x, kcnj12x
UniGene
Hs.200629
Gene Chromosome Location
Chr.17: 21376387-21419870 on Build GRCh38
Transcripts
NM_021012.4 XM_005256625.4 XM_011523831.2

SNP Detail

SNP ID Chromosome Location
rs573977980 Chr.17: 21419167-21419167 on Build GRCh38
rs77808408 Chr.17: 21419409-21419409 on Build GRCh38
rs782661721 Chr.17: 21419059-21419059 on Build GRCh38
rs534734443 Chr.17: 21419223-21419223 on Build GRCh38
rs555623336 Chr.17: 21419080-21419080 on Build GRCh38
rs189091626 Chr.17: 21419322-21419322 on Build GRCh38
rs553233747 Chr.17: 21419285-21419285 on Build GRCh38
rs577879119 Chr.17: 21419338-21419338 on Build GRCh38
rs782556669 Chr.17: 21419297-21419298 on Build GRCh38
rs575441163 Chr.17: 21419427-21419427 on Build GRCh38
rs782382335 Chr.17: 21419056-21419056 on Build GRCh38
rs532343937 Chr.17: 21419284-21419284 on Build GRCh38
rs782215071 Chr.17: 21419296-21419296 on Build GRCh38
rs141189728 Chr.17: 21419503-21419507 on Build GRCh38
rs147919595 Chr.17: 21419057-21419057 on Build GRCh38
rs141861667 Chr.17: 21419370-21419370 on Build GRCh38
rs375045378 Chr.17: 21419166-21419166 on Build GRCh38
rs761131915 Chr.17: 21419091-21419091 on Build GRCh38
rs111484179 Chr.17: 21419424-21419424 on Build GRCh38
rs563493032 Chr.17: 21419406-21419406 on Build GRCh38
rs376592390 Chr.17: 21419444-21419444 on Build GRCh38
rs3078445 Chr.17: 21419295-21419296 on Build GRCh38

View Full Product Details