Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45266456-45266960 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP3K14
Entrez Gene ID
9020
Gene Name
mitogen-activated protein kinase kinase kinase 14
Gene Aliases
FTDCR1B, HS, HSNIK, NIK
UniGene
Hs.404183
Gene Chromosome Location
Chr.17: 45263119-45317064 on Build GRCh38
Transcripts
NM_003954.4 XM_011525441.2 NR_024434.2 NR_024435.2 NR_110324.1 NR_110325.1 NR_110326.1

Gene Symbol
MAP3K14-AS1
Entrez Gene ID
100133991
Gene Name
MAP3K14 antisense RNA 1
Gene Aliases
-
UniGene
Hs.668927
Gene Chromosome Location
Chr.17: 45247925-45268630 on Build GRCh38
Transcripts
NM_003954.4 XM_011525441.2 NR_024434.2 NR_024435.2 NR_110324.1 NR_110325.1 NR_110326.1

SNP Detail

SNP ID Chromosome Location
rs368705783 Chr.17: 45266504-45266504 on Build GRCh38
rs552095058 Chr.17: 45266737-45266737 on Build GRCh38
rs150176691 Chr.17: 45266787-45266787 on Build GRCh38
rs765627329 Chr.17: 45266655-45266655 on Build GRCh38
rs776686902 Chr.17: 45266506-45266506 on Build GRCh38
rs751075145 Chr.17: 45266550-45266550 on Build GRCh38
rs1013880449 Chr.17: 45266938-45266938 on Build GRCh38
rs761580058 Chr.17: 45266728-45266728 on Build GRCh38
rs757262472 Chr.17: 45266693-45266693 on Build GRCh38
rs1002347225 Chr.17: 45266842-45266842 on Build GRCh38
rs372258853 Chr.17: 45266656-45266656 on Build GRCh38
rs779472760 Chr.17: 45266566-45266566 on Build GRCh38
rs1046510709 Chr.17: 45266834-45266834 on Build GRCh38
rs773453246 Chr.17: 45266503-45266503 on Build GRCh38
rs759245022 Chr.17: 45266510-45266510 on Build GRCh38
rs1031157147 Chr.17: 45266612-45266612 on Build GRCh38
rs1052546962 Chr.17: 45266929-45266929 on Build GRCh38
rs758693952 Chr.17: 45266573-45266573 on Build GRCh38
rs187767470 Chr.17: 45266733-45266733 on Build GRCh38
rs762514470 Chr.17: 45266851-45266851 on Build GRCh38
rs898686534 Chr.17: 45266830-45266830 on Build GRCh38
rs992429664 Chr.17: 45266621-45266621 on Build GRCh38
rs755535038 Chr.17: 45266654-45266654 on Build GRCh38
rs762617329 Chr.17: 45266529-45266529 on Build GRCh38
rs773846334 Chr.17: 45266723-45266723 on Build GRCh38
rs769727971 Chr.17: 45266703-45266703 on Build GRCh38
rs761227285 Chr.17: 45266649-45266649 on Build GRCh38
rs775101143 Chr.17: 45266523-45266523 on Build GRCh38
rs763752267 Chr.17: 45266633-45266633 on Build GRCh38
rs778135122 Chr.17: 45266678-45266678 on Build GRCh38
rs560703065 Chr.17: 45266766-45266766 on Build GRCh38
rs772260089 Chr.17: 45266502-45266502 on Build GRCh38
rs947763554 Chr.17: 45266791-45266791 on Build GRCh38
rs984045484 Chr.17: 45266776-45266776 on Build GRCh38
rs370090288 Chr.17: 45266695-45266695 on Build GRCh38
rs374327440 Chr.17: 45266553-45266553 on Build GRCh38
rs542346812 Chr.17: 45266626-45266626 on Build GRCh38
rs755271224 Chr.17: 45266562-45266562 on Build GRCh38
rs568914690 Chr.17: 45266784-45266784 on Build GRCh38
rs758889137 Chr.17: 45266666-45266666 on Build GRCh38
rs868160002 Chr.17: 45266817-45266817 on Build GRCh38
rs754437858 Chr.17: 45266559-45266559 on Build GRCh38
rs34836788 Chr.17: 45266691-45266691 on Build GRCh38
rs560799752 Chr.17: 45266647-45266647 on Build GRCh38
rs767096998 Chr.17: 45266651-45266651 on Build GRCh38
rs766785713 Chr.17: 45266558-45266558 on Build GRCh38
rs765120349 Chr.17: 45266512-45266512 on Build GRCh38
rs776443911 Chr.17: 45266603-45266603 on Build GRCh38
rs1011150275 Chr.17: 45266679-45266679 on Build GRCh38
rs867515151 Chr.17: 45266726-45266726 on Build GRCh38
rs112898702 Chr.17: 45266825-45266825 on Build GRCh38
rs748425005 Chr.17: 45266490-45266490 on Build GRCh38
rs747044497 Chr.17: 45266583-45266583 on Build GRCh38
rs754282933 Chr.17: 45266653-45266653 on Build GRCh38
rs571898629 Chr.17: 45266595-45266595 on Build GRCh38
rs369646866 Chr.17: 45266630-45266630 on Build GRCh38
rs11574834 Chr.17: 45266480-45266480 on Build GRCh38
rs935115432 Chr.17: 45266911-45266911 on Build GRCh38
rs563200129 Chr.17: 45266709-45266709 on Build GRCh38
rs775209563 Chr.17: 45266704-45266704 on Build GRCh38
rs371527687 Chr.17: 45266539-45266539 on Build GRCh38
rs777939708 Chr.17: 45266579-45266579 on Build GRCh38
rs960905788 Chr.17: 45266897-45266897 on Build GRCh38
rs376746377 Chr.17: 45266604-45266604 on Build GRCh38
rs376945193 Chr.17: 45266729-45266729 on Build GRCh38
rs371995924 Chr.17: 45266602-45266602 on Build GRCh38
rs775296010 Chr.17: 45266610-45266610 on Build GRCh38
rs770886542 Chr.17: 45266576-45266576 on Build GRCh38
rs988032968 Chr.17: 45266750-45266758 on Build GRCh38
rs879107508 Chr.17: 45266526-45266526 on Build GRCh38
rs2074293 Chr.17: 45266706-45266706 on Build GRCh38
rs745753442 Chr.17: 45266696-45266696 on Build GRCh38
rs561757277 Chr.17: 45266572-45266572 on Build GRCh38
rs770783454 Chr.17: 45266505-45266505 on Build GRCh38
rs771108600 Chr.17: 45266585-45266585 on Build GRCh38
rs56112291 Chr.17: 45266682-45266682 on Build GRCh38

View Full Product Details