Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45453635-45454139 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLEKHM1
Entrez Gene ID
9842
Gene Name
pleckstrin homology and RUN domain containing M1
Gene Aliases
AP162, B2, OPTB6
UniGene
Hs.514242
Gene Chromosome Location
Chr.17: 45434262-45490780 on Build GRCh38
Transcripts
NM_014798.2 NR_027774.1 NR_027782.1 XM_006722201.3 XM_011525523.2 XM_011525524.1 XM_011525525.1 XM_011525526.1 XM_011525528.2 XM_017025451.1 XM_017025452.1 XM_017025453.1 XM_017025454.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
706091 c.2155G>A p.E719K Chr.17: 45453697-45453697 on Build GRCh38
6146905 c.2147G>T p.R716M Chr.17: 45453705-45453705 on Build GRCh38
706090 c.2047G>C p.D683H Chr.17: 45453805-45453805 on Build GRCh38
5190236 c.2044C>T p.P682S Chr.17: 45453808-45453808 on Build GRCh38
980160 c.1816C>T p.R606C Chr.17: 45454036-45454036 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs894128489 Chr.17: 45453940-45453940 on Build GRCh38
rs768886144 Chr.17: 45453718-45453718 on Build GRCh38
rs779480934 Chr.17: 45453821-45453821 on Build GRCh38
rs753581598 Chr.17: 45453657-45453657 on Build GRCh38
rs753232738 Chr.17: 45454051-45454051 on Build GRCh38
rs575202222 Chr.17: 45453674-45453674 on Build GRCh38
rs560791574 Chr.17: 45453766-45453766 on Build GRCh38
rs199719858 Chr.17: 45454019-45454019 on Build GRCh38
rs777311496 Chr.17: 45453684-45453684 on Build GRCh38
rs776439772 Chr.17: 45453769-45453769 on Build GRCh38
rs773494068 Chr.17: 45453754-45453754 on Build GRCh38
rs759279787 Chr.17: 45453770-45453770 on Build GRCh38
rs760063830 Chr.17: 45453955-45453955 on Build GRCh38
rs145772830 Chr.17: 45453908-45453908 on Build GRCh38
rs539498298 Chr.17: 45454117-45454117 on Build GRCh38
rs140938435 Chr.17: 45453824-45453824 on Build GRCh38
rs2684500 Chr.17: 45453837-45453837 on Build GRCh38
rs768647001 Chr.17: 45454086-45454086 on Build GRCh38
rs774687855 Chr.17: 45453728-45453728 on Build GRCh38
rs749844668 Chr.17: 45453816-45453816 on Build GRCh38
rs374996512 Chr.17: 45453857-45453857 on Build GRCh38
rs372997534 Chr.17: 45453750-45453750 on Build GRCh38
rs775203514 Chr.17: 45453922-45453922 on Build GRCh38
rs539028922 Chr.17: 45453699-45453699 on Build GRCh38
rs2684501 Chr.17: 45453931-45453931 on Build GRCh38
rs749222069 Chr.17: 45454081-45454081 on Build GRCh38
rs752322789 Chr.17: 45453772-45453772 on Build GRCh38
rs770863357 Chr.17: 45453841-45453841 on Build GRCh38
rs781495753 Chr.17: 45454066-45454066 on Build GRCh38
rs369056286 Chr.17: 45453860-45453860 on Build GRCh38
rs763610032 Chr.17: 45454053-45454053 on Build GRCh38
rs145187824 Chr.17: 45454055-45454055 on Build GRCh38
rs546482206 Chr.17: 45453970-45453970 on Build GRCh38
rs372119492 Chr.17: 45454074-45454074 on Build GRCh38
rs757359910 Chr.17: 45454062-45454062 on Build GRCh38
rs749275402 Chr.17: 45454018-45454018 on Build GRCh38
rs778518312 Chr.17: 45454087-45454087 on Build GRCh38
rs748537332 Chr.17: 45453894-45453894 on Build GRCh38
rs755279647 Chr.17: 45453820-45453820 on Build GRCh38
rs567962159 Chr.17: 45453979-45453979 on Build GRCh38
rs2684499 Chr.17: 45453807-45453807 on Build GRCh38
rs758806341 Chr.17: 45453968-45453968 on Build GRCh38
rs747998318 Chr.17: 45454110-45454110 on Build GRCh38
rs780394793 Chr.17: 45453709-45453709 on Build GRCh38
rs564118779 Chr.17: 45453907-45453907 on Build GRCh38
rs779995813 Chr.17: 45454016-45454016 on Build GRCh38
rs577637843 Chr.17: 45453777-45453777 on Build GRCh38
rs762756530 Chr.17: 45453930-45453930 on Build GRCh38
rs554864108 Chr.17: 45453828-45453828 on Build GRCh38
rs780921651 Chr.17: 45453871-45453871 on Build GRCh38
rs386797350 Chr.17: 45453802-45453804 on Build GRCh38
rs758646952 Chr.17: 45453896-45453896 on Build GRCh38
rs375411710 Chr.17: 45454033-45454033 on Build GRCh38
rs530953924 Chr.17: 45453891-45453891 on Build GRCh38
rs563592720 Chr.17: 45453887-45453887 on Build GRCh38
rs760295037 Chr.17: 45454036-45454036 on Build GRCh38
rs372492506 Chr.17: 45454075-45454075 on Build GRCh38
rs143050885 Chr.17: 45453990-45453990 on Build GRCh38
rs758103308 Chr.17: 45453773-45453773 on Build GRCh38
rs559224144 Chr.17: 45453712-45453712 on Build GRCh38
rs767106989 Chr.17: 45453871-45453871 on Build GRCh38
rs751956968 Chr.17: 45454056-45454056 on Build GRCh38
rs535213900 Chr.17: 45453658-45453658 on Build GRCh38
rs370244543 Chr.17: 45454021-45454021 on Build GRCh38
rs769723896 Chr.17: 45453917-45453917 on Build GRCh38
rs151036182 Chr.17: 45453673-45453673 on Build GRCh38
rs545282428 Chr.17: 45453748-45453748 on Build GRCh38
rs145770009 Chr.17: 45453902-45453902 on Build GRCh38
rs745788073 Chr.17: 45453909-45453909 on Build GRCh38
rs150208882 Chr.17: 45453859-45453859 on Build GRCh38
rs769667409 Chr.17: 45453996-45453996 on Build GRCh38
rs747081650 Chr.17: 45453836-45453836 on Build GRCh38
rs778560481 Chr.17: 45453670-45453670 on Build GRCh38
rs541727366 Chr.17: 45453799-45453799 on Build GRCh38
rs528404155 Chr.17: 45453941-45453941 on Build GRCh38
rs367791293 Chr.17: 45453853-45453853 on Build GRCh38
rs752706762 Chr.17: 45454061-45454062 on Build GRCh38
rs1033700358 Chr.17: 45454037-45454037 on Build GRCh38
rs748379987 Chr.17: 45453823-45453823 on Build GRCh38
rs753073455 Chr.17: 45453967-45453967 on Build GRCh38
rs772548345 Chr.17: 45454035-45454035 on Build GRCh38
rs1055570814 Chr.17: 45453667-45453667 on Build GRCh38
rs756582126 Chr.17: 45453797-45453797 on Build GRCh38
rs751033146 Chr.17: 45453788-45453788 on Build GRCh38
rs199741015 Chr.17: 45453749-45453749 on Build GRCh38
rs775326791 Chr.17: 45453858-45453858 on Build GRCh38
rs756694397 Chr.17: 45453867-45453867 on Build GRCh38
rs771770966 Chr.17: 45454034-45454034 on Build GRCh38
rs375419286 Chr.17: 45453690-45453690 on Build GRCh38
rs746709929 Chr.17: 45454118-45454118 on Build GRCh38
rs759565210 Chr.17: 45453852-45453852 on Build GRCh38
rs771584552 Chr.17: 45454111-45454111 on Build GRCh38
rs778248742 Chr.17: 45453969-45453969 on Build GRCh38
rs751028545 Chr.17: 45453866-45453866 on Build GRCh38
rs770268311 Chr.17: 45453702-45453702 on Build GRCh38
rs2684498 Chr.17: 45453736-45453736 on Build GRCh38
rs774274190 Chr.17: 45454026-45454026 on Build GRCh38
rs766028431 Chr.17: 45454050-45454050 on Build GRCh38
rs765056104 Chr.17: 45453771-45453771 on Build GRCh38
rs776400345 Chr.17: 45453846-45453846 on Build GRCh38
rs761586441 Chr.17: 45453935-45453935 on Build GRCh38
rs535231948 Chr.17: 45453991-45453991 on Build GRCh38
rs148973891 Chr.17: 45453903-45453903 on Build GRCh38
rs765867877 Chr.17: 45453960-45453960 on Build GRCh38
rs754291355 Chr.17: 45453872-45453872 on Build GRCh38

View Full Product Details