Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50707755-50708249 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD40
Entrez Gene ID
91369
Gene Name
ankyrin repeat domain 40
Gene Aliases
-
UniGene
Hs.463426
Gene Chromosome Location
Chr.17: 50693190-50707909 on Build GRCh38
Transcripts
NM_052855.3

SNP Detail

SNP ID Chromosome Location
rs560227947 Chr.17: 50708104-50708104 on Build GRCh38
rs898744535 Chr.17: 50708210-50708210 on Build GRCh38
rs930844683 Chr.17: 50708053-50708053 on Build GRCh38
rs577539362 Chr.17: 50707972-50707972 on Build GRCh38
rs780307956 Chr.17: 50708066-50708066 on Build GRCh38
rs932853863 Chr.17: 50707866-50707866 on Build GRCh38
rs921693310 Chr.17: 50707852-50707852 on Build GRCh38
rs1003890096 Chr.17: 50707917-50707917 on Build GRCh38
rs974431335 Chr.17: 50708092-50708092 on Build GRCh38
rs957976463 Chr.17: 50708083-50708083 on Build GRCh38
rs529196416 Chr.17: 50708157-50708157 on Build GRCh38
rs931345548 Chr.17: 50708217-50708222 on Build GRCh38
rs940280127 Chr.17: 50708168-50708168 on Build GRCh38
rs562630132 Chr.17: 50708184-50708184 on Build GRCh38
rs756297622 Chr.17: 50707989-50707989 on Build GRCh38
rs749759912 Chr.17: 50708191-50708191 on Build GRCh38
rs907227275 Chr.17: 50708172-50708172 on Build GRCh38
rs1022503058 Chr.17: 50707962-50707962 on Build GRCh38
rs549348253 Chr.17: 50708170-50708170 on Build GRCh38
rs989238456 Chr.17: 50707961-50707961 on Build GRCh38
rs956776789 Chr.17: 50707949-50707949 on Build GRCh38
rs936405709 Chr.17: 50707834-50707834 on Build GRCh38
rs1013864947 Chr.17: 50708005-50708005 on Build GRCh38
rs991999098 Chr.17: 50707789-50707789 on Build GRCh38
rs987390688 Chr.17: 50708156-50708156 on Build GRCh38
rs1032017062 Chr.17: 50708056-50708056 on Build GRCh38
rs915961462 Chr.17: 50708076-50708076 on Build GRCh38
rs8068442 Chr.17: 50707804-50707804 on Build GRCh38
rs879843073 Chr.17: 50708156-50708158 on Build GRCh38
rs953135044 Chr.17: 50707973-50707973 on Build GRCh38
rs750512115 Chr.17: 50707928-50707928 on Build GRCh38
rs944940426 Chr.17: 50708019-50708019 on Build GRCh38
rs974862200 Chr.17: 50707841-50707841 on Build GRCh38
rs544255374 Chr.17: 50707808-50707808 on Build GRCh38
rs959434304 Chr.17: 50707774-50707774 on Build GRCh38
rs940249788 Chr.17: 50707909-50707909 on Build GRCh38
rs919424521 Chr.17: 50708051-50708051 on Build GRCh38
rs995613679 Chr.17: 50707976-50707976 on Build GRCh38
rs533815552 Chr.17: 50708011-50708011 on Build GRCh38
rs891409349 Chr.17: 50707908-50707908 on Build GRCh38
rs1024920011 Chr.17: 50707803-50707803 on Build GRCh38
rs555055614 Chr.17: 50708154-50708154 on Build GRCh38
rs972754423 Chr.17: 50708023-50708023 on Build GRCh38
rs1034214312 Chr.17: 50707991-50707991 on Build GRCh38
rs796361712 Chr.17: 50708126-50708126 on Build GRCh38
rs866395609 Chr.17: 50707996-50707996 on Build GRCh38
rs1051256776 Chr.17: 50707906-50707913 on Build GRCh38
rs777730757 Chr.17: 50708091-50708091 on Build GRCh38
rs138472752 Chr.17: 50707919-50707919 on Build GRCh38
rs879198192 Chr.17: 50708223-50708223 on Build GRCh38

View Full Product Details