Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50867764-50868274 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TOB1
Entrez Gene ID
10140
Gene Name
transducer of ERBB2, 1
Gene Aliases
APRO5, APRO6, PIG49, TOB, TROB, TROB1
UniGene
Hs.703321
Gene Chromosome Location
Chr.17: 50862223-50867978 on Build GRCh38
Transcripts
NM_001243877.1 NR_038458.1

Gene Symbol
TOB1-AS1
Entrez Gene ID
400604
Gene Name
TOB1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.641441
Gene Chromosome Location
Chr.17: 50866679-50868371 on Build GRCh38
Transcripts
NM_001243877.1 NR_038458.1

SNP Detail

SNP ID Chromosome Location
rs986067144 Chr.17: 50868088-50868088 on Build GRCh38
rs758839410 Chr.17: 50868054-50868054 on Build GRCh38
rs1008974996 Chr.17: 50868008-50868008 on Build GRCh38
rs911837552 Chr.17: 50868132-50868132 on Build GRCh38
rs1134637 Chr.17: 50867916-50867916 on Build GRCh38
rs987911762 Chr.17: 50867865-50867865 on Build GRCh38
rs981778571 Chr.17: 50867868-50867868 on Build GRCh38
rs949163371 Chr.17: 50867867-50867867 on Build GRCh38
rs890574991 Chr.17: 50867972-50867972 on Build GRCh38
rs961318569 Chr.17: 50867812-50867812 on Build GRCh38
rs1053982121 Chr.17: 50867931-50867931 on Build GRCh38
rs1023461779 Chr.17: 50868231-50868231 on Build GRCh38
rs934956516 Chr.17: 50867923-50867923 on Build GRCh38
rs894027734 Chr.17: 50867938-50867938 on Build GRCh38
rs952667066 Chr.17: 50867835-50867835 on Build GRCh38
rs972648567 Chr.17: 50867815-50867815 on Build GRCh38
rs56406873 Chr.17: 50868146-50868146 on Build GRCh38
rs949248747 Chr.17: 50868161-50868161 on Build GRCh38
rs201828321 Chr.17: 50868160-50868161 on Build GRCh38
rs576712914 Chr.17: 50868003-50868003 on Build GRCh38
rs911736816 Chr.17: 50867861-50867861 on Build GRCh38
rs543734471 Chr.17: 50868155-50868155 on Build GRCh38
rs972334774 Chr.17: 50868209-50868209 on Build GRCh38
rs538891756 Chr.17: 50867952-50867952 on Build GRCh38
rs991101288 Chr.17: 50868213-50868213 on Build GRCh38
rs953261347 Chr.17: 50868083-50868083 on Build GRCh38
rs1014977739 Chr.17: 50868034-50868034 on Build GRCh38
rs897847193 Chr.17: 50868050-50868050 on Build GRCh38
rs888675933 Chr.17: 50867942-50867942 on Build GRCh38
rs1004161523 Chr.17: 50868005-50868005 on Build GRCh38
rs916341931 Chr.17: 50867857-50867857 on Build GRCh38
rs376986962 Chr.17: 50867856-50867856 on Build GRCh38
rs767085470 Chr.17: 50868146-50868147 on Build GRCh38
rs750835325 Chr.17: 50867795-50867795 on Build GRCh38
rs149089074 Chr.17: 50868057-50868057 on Build GRCh38
rs1050597808 Chr.17: 50867969-50867969 on Build GRCh38
rs923614503 Chr.17: 50867917-50867917 on Build GRCh38
rs201925433 Chr.17: 50867937-50867937 on Build GRCh38
rs540782672 Chr.17: 50868015-50868015 on Build GRCh38
rs944547172 Chr.17: 50868145-50868145 on Build GRCh38
rs1031077202 Chr.17: 50868010-50868012 on Build GRCh38
rs749934376 Chr.17: 50868147-50868147 on Build GRCh38
rs989423385 Chr.17: 50868012-50868012 on Build GRCh38
rs200297041 Chr.17: 50867934-50867934 on Build GRCh38
rs572064472 Chr.17: 50868004-50868004 on Build GRCh38
rs1024192068 Chr.17: 50867982-50867983 on Build GRCh38
rs952643474 Chr.17: 50868207-50868207 on Build GRCh38
rs932169412 Chr.17: 50867964-50867964 on Build GRCh38
rs1026880547 Chr.17: 50867829-50867829 on Build GRCh38
rs994490505 Chr.17: 50867926-50867926 on Build GRCh38
rs574375524 Chr.17: 50868082-50868082 on Build GRCh38
rs994811828 Chr.17: 50868053-50868053 on Build GRCh38
rs796125444 Chr.17: 50868160-50868160 on Build GRCh38
rs796643444 Chr.17: 50868010-50868010 on Build GRCh38

View Full Product Details