Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 58363195-58363698 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF43
Entrez Gene ID
54894
Gene Name
ring finger protein 43
Gene Aliases
RNF124, URCC
UniGene
Hs.584916
Gene Chromosome Location
Chr.17: 58352500-58418895 on Build GRCh38
Transcripts
NM_001305544.1 NM_001305545.1 NM_017763.5 XM_011524955.2 XM_011524956.2 XM_017024800.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6946373 c.407C>T p.A136V Chr.17: 58363569-58363569 on Build GRCh38
6979914 c.575_576insC p.A193fs*6 Chr.17: 58363281-58363282 on Build GRCh38
6932623 c.489delG p.I164fs*9 Chr.17: 58363368-58363368 on Build GRCh38
6981511 c.508G>T p.E170* Chr.17: 58363349-58363349 on Build GRCh38
1287671 c.554G>T p.R185M Chr.17: 58363303-58363303 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779625197 Chr.17: 58363229-58363229 on Build GRCh38
rs368233607 Chr.17: 58363354-58363354 on Build GRCh38
rs1017343030 Chr.17: 58363587-58363587 on Build GRCh38
rs190547268 Chr.17: 58363374-58363374 on Build GRCh38
rs767814408 Chr.17: 58363410-58363410 on Build GRCh38
rs890509181 Chr.17: 58363241-58363241 on Build GRCh38
rs375704415 Chr.17: 58363497-58363497 on Build GRCh38
rs756576921 Chr.17: 58363329-58363329 on Build GRCh38
rs145164323 Chr.17: 58363339-58363339 on Build GRCh38
rs745640921 Chr.17: 58363614-58363614 on Build GRCh38
rs768895407 Chr.17: 58363637-58363637 on Build GRCh38
rs765811705 Chr.17: 58363571-58363571 on Build GRCh38
rs373829009 Chr.17: 58363508-58363508 on Build GRCh38
rs786205215 Chr.17: 58363582-58363582 on Build GRCh38
rs752331470 Chr.17: 58363586-58363586 on Build GRCh38
rs367982531 Chr.17: 58363452-58363452 on Build GRCh38
rs532357871 Chr.17: 58363386-58363386 on Build GRCh38
rs1044896377 Chr.17: 58363402-58363402 on Build GRCh38
rs369636118 Chr.17: 58363597-58363597 on Build GRCh38
rs762838524 Chr.17: 58363390-58363390 on Build GRCh38
rs754183734 Chr.17: 58363346-58363346 on Build GRCh38
rs779094793 Chr.17: 58363446-58363446 on Build GRCh38
rs371817844 Chr.17: 58363496-58363496 on Build GRCh38
rs780946287 Chr.17: 58363489-58363489 on Build GRCh38
rs79125070 Chr.17: 58363245-58363245 on Build GRCh38
rs752845745 Chr.17: 58363327-58363327 on Build GRCh38
rs754553091 Chr.17: 58363479-58363479 on Build GRCh38
rs35219874 Chr.17: 58363596-58363596 on Build GRCh38
rs779337229 Chr.17: 58363352-58363352 on Build GRCh38
rs764098066 Chr.17: 58363513-58363513 on Build GRCh38
rs746582652 Chr.17: 58363353-58363353 on Build GRCh38
rs1024483355 Chr.17: 58363406-58363406 on Build GRCh38
rs1034826397 Chr.17: 58363455-58363455 on Build GRCh38
rs755845342 Chr.17: 58363589-58363589 on Build GRCh38
rs990836027 Chr.17: 58363666-58363666 on Build GRCh38
rs780526844 Chr.17: 58363357-58363357 on Build GRCh38
rs376504970 Chr.17: 58363505-58363505 on Build GRCh38
rs200276342 Chr.17: 58363627-58363627 on Build GRCh38
rs774211426 Chr.17: 58363396-58363396 on Build GRCh38
rs929307033 Chr.17: 58363223-58363223 on Build GRCh38
rs747508936 Chr.17: 58363618-58363618 on Build GRCh38
rs375590378 Chr.17: 58363585-58363585 on Build GRCh38
rs762889537 Chr.17: 58363286-58363286 on Build GRCh38
rs144994226 Chr.17: 58363395-58363395 on Build GRCh38
rs1000142712 Chr.17: 58363298-58363298 on Build GRCh38
rs76384648 Chr.17: 58363281-58363281 on Build GRCh38
rs766694756 Chr.17: 58363296-58363296 on Build GRCh38
rs985880670 Chr.17: 58363583-58363583 on Build GRCh38
rs139557480 Chr.17: 58363282-58363282 on Build GRCh38
rs776216229 Chr.17: 58363278-58363278 on Build GRCh38
rs764576439 Chr.17: 58363420-58363420 on Build GRCh38
rs754272026 Chr.17: 58363422-58363422 on Build GRCh38
rs750730096 Chr.17: 58363578-58363578 on Build GRCh38
rs757770356 Chr.17: 58363427-58363427 on Build GRCh38
rs767440689 Chr.17: 58363324-58363324 on Build GRCh38
rs926765108 Chr.17: 58363481-58363481 on Build GRCh38
rs747646412 Chr.17: 58363493-58363493 on Build GRCh38
rs142178517 Chr.17: 58363533-58363533 on Build GRCh38
rs143954546 Chr.17: 58363507-58363507 on Build GRCh38
rs767980835 Chr.17: 58363252-58363252 on Build GRCh38
rs760796801 Chr.17: 58363419-58363419 on Build GRCh38
rs778962170 Chr.17: 58363599-58363599 on Build GRCh38
rs1032910124 Chr.17: 58363308-58363308 on Build GRCh38
rs757680292 Chr.17: 58363349-58363349 on Build GRCh38
rs747700626 Chr.17: 58363360-58363360 on Build GRCh38
rs1047683627 Chr.17: 58363536-58363536 on Build GRCh38
rs777057577 Chr.17: 58363411-58363411 on Build GRCh38
rs777370273 Chr.17: 58363590-58363590 on Build GRCh38
rs758854182 Chr.17: 58363581-58363581 on Build GRCh38
rs771973438 Chr.17: 58363615-58363615 on Build GRCh38
rs536391017 Chr.17: 58363464-58363464 on Build GRCh38
rs759809905 Chr.17: 58363319-58363319 on Build GRCh38
rs760743639 Chr.17: 58363511-58363511 on Build GRCh38
rs752745971 Chr.17: 58363418-58363418 on Build GRCh38
rs201665096 Chr.17: 58363556-58363556 on Build GRCh38
rs776658772 Chr.17: 58363647-58363647 on Build GRCh38
rs986321432 Chr.17: 58363257-58363257 on Build GRCh38
rs762377266 Chr.17: 58363481-58363484 on Build GRCh38
rs569008851 Chr.17: 58363408-58363408 on Build GRCh38
rs891676521 Chr.17: 58363674-58363674 on Build GRCh38
rs751796204 Chr.17: 58363310-58363310 on Build GRCh38
rs151209912 Chr.17: 58363542-58363542 on Build GRCh38
rs773979099 Chr.17: 58363506-58363506 on Build GRCh38

View Full Product Details