Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 78160563-78161102 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf99
Entrez Gene ID
100141515
Gene Name
chromosome 17 open reading frame 99
Gene Aliases
UNQ464
UniGene
Hs.633034
Gene Chromosome Location
Chr.17: 78146035-78166283 on Build GRCh38
Transcripts
NM_001163075.1 XM_011524152.2 XM_017023997.1 XM_017023998.1 XM_017023999.1 XM_017024000.1 XM_017024001.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5796172 c.71-7C>G p.? Chr.17: 78160948-78160948 on Build GRCh38
4597493 c.79C>T p.P27S Chr.17: 78160963-78160963 on Build GRCh38
3388240 c.169C>A p.P57T Chr.17: 78161053-78161053 on Build GRCh38
5449057 c.187T>C p.C63R Chr.17: 78161071-78161071 on Build GRCh38
3820761 c.108G>A p.L36L Chr.17: 78160992-78160992 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs974217266 Chr.17: 78160895-78160895 on Build GRCh38
rs866328064 Chr.17: 78160784-78160784 on Build GRCh38
rs982795518 Chr.17: 78160836-78160836 on Build GRCh38
rs557535390 Chr.17: 78160638-78160638 on Build GRCh38
rs992370865 Chr.17: 78160669-78160669 on Build GRCh38
rs907108116 Chr.17: 78160719-78160719 on Build GRCh38
rs770775984 Chr.17: 78160940-78160940 on Build GRCh38
rs1045661091 Chr.17: 78160709-78160709 on Build GRCh38
rs542727025 Chr.17: 78160782-78160782 on Build GRCh38
rs955609489 Chr.17: 78161049-78161049 on Build GRCh38
rs754738866 Chr.17: 78160826-78160826 on Build GRCh38
rs982996823 Chr.17: 78161055-78161055 on Build GRCh38
rs1029811813 Chr.17: 78161038-78161038 on Build GRCh38
rs7225706 Chr.17: 78160744-78160744 on Build GRCh38
rs999571249 Chr.17: 78160881-78160881 on Build GRCh38
rs533446524 Chr.17: 78160901-78160901 on Build GRCh38
rs540561366 Chr.17: 78160741-78160741 on Build GRCh38
rs190149894 Chr.17: 78161051-78161051 on Build GRCh38
rs72898209 Chr.17: 78160963-78160963 on Build GRCh38
rs773346375 Chr.17: 78161022-78161022 on Build GRCh38
rs967709620 Chr.17: 78160868-78160868 on Build GRCh38
rs571062918 Chr.17: 78160872-78160872 on Build GRCh38
rs1047076171 Chr.17: 78160626-78160626 on Build GRCh38
rs907149032 Chr.17: 78160962-78160962 on Build GRCh38
rs142662896 Chr.17: 78160611-78160611 on Build GRCh38
rs755641643 Chr.17: 78160888-78160888 on Build GRCh38
rs113638656 Chr.17: 78160712-78160712 on Build GRCh38
rs754833049 Chr.17: 78160779-78160779 on Build GRCh38
rs73375792 Chr.17: 78161012-78161012 on Build GRCh38
rs980212271 Chr.17: 78160996-78160996 on Build GRCh38
rs900266580 Chr.17: 78160792-78160792 on Build GRCh38
rs755791446 Chr.17: 78160986-78160986 on Build GRCh38
rs748921560 Chr.17: 78160917-78160917 on Build GRCh38
rs778508453 Chr.17: 78160984-78160984 on Build GRCh38
rs751730773 Chr.17: 78161052-78161052 on Build GRCh38
rs975701094 Chr.17: 78160590-78160590 on Build GRCh38
rs995923622 Chr.17: 78160824-78160824 on Build GRCh38
rs1038620148 Chr.17: 78160645-78160645 on Build GRCh38
rs139587690 Chr.17: 78160851-78160851 on Build GRCh38
rs777145443 Chr.17: 78160976-78160976 on Build GRCh38
rs920627559 Chr.17: 78160734-78160734 on Build GRCh38
rs758658660 Chr.17: 78160771-78160771 on Build GRCh38
rs149713334 Chr.17: 78161027-78161027 on Build GRCh38
rs944916403 Chr.17: 78161031-78161031 on Build GRCh38
rs894271690 Chr.17: 78160646-78160646 on Build GRCh38
rs768907018 Chr.17: 78160969-78160969 on Build GRCh38
rs751958948 Chr.17: 78160699-78160699 on Build GRCh38
rs7225593 Chr.17: 78160783-78160783 on Build GRCh38
rs908639391 Chr.17: 78160632-78160632 on Build GRCh38
rs748070910 Chr.17: 78160871-78160871 on Build GRCh38
rs997030667 Chr.17: 78161028-78161028 on Build GRCh38
rs967686711 Chr.17: 78160639-78160639 on Build GRCh38
rs759332203 Chr.17: 78161054-78161054 on Build GRCh38
rs62079086 Chr.17: 78160902-78160902 on Build GRCh38
rs922841711 Chr.17: 78160597-78160597 on Build GRCh38
rs762356679 Chr.17: 78161001-78161001 on Build GRCh38
rs948576950 Chr.17: 78160912-78160912 on Build GRCh38
rs531455790 Chr.17: 78160805-78160805 on Build GRCh38
rs62079085 Chr.17: 78160724-78160724 on Build GRCh38
rs185252514 Chr.17: 78160967-78160967 on Build GRCh38
rs964278333 Chr.17: 78160767-78160767 on Build GRCh38
rs960587369 Chr.17: 78160661-78160661 on Build GRCh38
rs908498358 Chr.17: 78160840-78160840 on Build GRCh38
rs1017768617 Chr.17: 78160765-78160765 on Build GRCh38
rs892838300 Chr.17: 78161011-78161011 on Build GRCh38
rs528912120 Chr.17: 78160745-78160745 on Build GRCh38
rs778066473 Chr.17: 78160662-78160662 on Build GRCh38
rs960873528 Chr.17: 78160916-78160916 on Build GRCh38
rs1016250578 Chr.17: 78161059-78161059 on Build GRCh38
rs1043472476 Chr.17: 78161066-78161066 on Build GRCh38
rs1012702877 Chr.17: 78160705-78160705 on Build GRCh38

View Full Product Details