Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 78674309-78674814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYTH1
Entrez Gene ID
9267
Gene Name
cytohesin 1
Gene Aliases
B2-1, CYTOHESIN-1, D17S811E, PSCD1, SEC7
UniGene
Hs.191215
Gene Chromosome Location
Chr.17: 78674047-78782342 on Build GRCh38
Transcripts
NM_001292018.1 NM_001292019.1 NM_004762.3 NM_017456.3 XM_011525475.2 XM_011525476.2 XM_011525477.1 XM_011525478.1 XM_017025336.1 XM_017025338.1

SNP Detail

SNP ID Chromosome Location
rs774159603 Chr.17: 78674332-78674332 on Build GRCh38
rs1017169104 Chr.17: 78674690-78674690 on Build GRCh38
rs979143185 Chr.17: 78674748-78674748 on Build GRCh38
rs555121390 Chr.17: 78674516-78674516 on Build GRCh38
rs561214281 Chr.17: 78674335-78674335 on Build GRCh38
rs190585191 Chr.17: 78674336-78674336 on Build GRCh38
rs1004767727 Chr.17: 78674445-78674445 on Build GRCh38
rs868829274 Chr.17: 78674717-78674717 on Build GRCh38
rs574885062 Chr.17: 78674363-78674363 on Build GRCh38
rs746767335 Chr.17: 78674610-78674610 on Build GRCh38
rs879018371 Chr.17: 78674521-78674521 on Build GRCh38
rs1015256958 Chr.17: 78674500-78674500 on Build GRCh38
rs941732796 Chr.17: 78674542-78674542 on Build GRCh38
rs953130800 Chr.17: 78674577-78674577 on Build GRCh38
rs950375651 Chr.17: 78674787-78674787 on Build GRCh38
rs552037519 Chr.17: 78674623-78674623 on Build GRCh38
rs543364504 Chr.17: 78674470-78674470 on Build GRCh38
rs1035155209 Chr.17: 78674664-78674664 on Build GRCh38
rs186413373 Chr.17: 78674774-78674774 on Build GRCh38
rs962994957 Chr.17: 78674708-78674708 on Build GRCh38
rs563599012 Chr.17: 78674362-78674362 on Build GRCh38
rs1804435 Chr.17: 78674416-78674416 on Build GRCh38
rs771058263 Chr.17: 78674615-78674615 on Build GRCh38
rs557958198 Chr.17: 78674674-78674674 on Build GRCh38
rs375136402 Chr.17: 78674684-78674684 on Build GRCh38
rs796908302 Chr.17: 78674356-78674356 on Build GRCh38
rs973112990 Chr.17: 78674561-78674561 on Build GRCh38
rs181656925 Chr.17: 78674625-78674625 on Build GRCh38
rs940070918 Chr.17: 78674525-78674525 on Build GRCh38
rs541656126 Chr.17: 78674334-78674334 on Build GRCh38
rs563362826 Chr.17: 78674567-78674567 on Build GRCh38
rs893800573 Chr.17: 78674629-78674629 on Build GRCh38
rs879095170 Chr.17: 78674528-78674528 on Build GRCh38
rs1039005750 Chr.17: 78674627-78674627 on Build GRCh38
rs368675508 Chr.17: 78674591-78674591 on Build GRCh38
rs9894449 Chr.17: 78674729-78674729 on Build GRCh38
rs905992744 Chr.17: 78674385-78674385 on Build GRCh38
rs372680667 Chr.17: 78674601-78674601 on Build GRCh38
rs997331716 Chr.17: 78674712-78674712 on Build GRCh38
rs1021069700 Chr.17: 78674343-78674343 on Build GRCh38
rs1126615 Chr.17: 78674667-78674667 on Build GRCh38
rs915575507 Chr.17: 78674517-78674517 on Build GRCh38
rs952153561 Chr.17: 78674436-78674436 on Build GRCh38
rs982279891 Chr.17: 78674352-78674352 on Build GRCh38
rs987667322 Chr.17: 78674722-78674722 on Build GRCh38
rs535440544 Chr.17: 78674384-78674384 on Build GRCh38
rs76413727 Chr.17: 78674713-78674713 on Build GRCh38
rs1014834764 Chr.17: 78674697-78674697 on Build GRCh38
rs965900700 Chr.17: 78674503-78674503 on Build GRCh38
rs796204241 Chr.17: 78674617-78674617 on Build GRCh38
rs1021392751 Chr.17: 78674641-78674641 on Build GRCh38
rs1038953357 Chr.17: 78674761-78674761 on Build GRCh38
rs772043655 Chr.17: 78674563-78674564 on Build GRCh38
rs1045792540 Chr.17: 78674374-78674374 on Build GRCh38
rs929064205 Chr.17: 78674612-78674612 on Build GRCh38
rs1010935724 Chr.17: 78674634-78674634 on Build GRCh38
rs918930059 Chr.17: 78674716-78674716 on Build GRCh38
rs956396998 Chr.17: 78674768-78674768 on Build GRCh38
rs1010989511 Chr.17: 78674337-78674337 on Build GRCh38
rs112855463 Chr.17: 78674656-78674656 on Build GRCh38
rs946629605 Chr.17: 78674773-78674773 on Build GRCh38
rs1001479056 Chr.17: 78674401-78674401 on Build GRCh38
rs902568683 Chr.17: 78674644-78674644 on Build GRCh38
rs540567164 Chr.17: 78674767-78674767 on Build GRCh38
rs967262331 Chr.17: 78674350-78674350 on Build GRCh38
rs950240695 Chr.17: 78674628-78674628 on Build GRCh38
rs943214969 Chr.17: 78674740-78674740 on Build GRCh38
rs1027636832 Chr.17: 78674403-78674403 on Build GRCh38

View Full Product Details