Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 78680098-78680601 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYTH1
Entrez Gene ID
9267
Gene Name
cytohesin 1
Gene Aliases
B2-1, CYTOHESIN-1, D17S811E, PSCD1, SEC7
UniGene
Hs.191215
Gene Chromosome Location
Chr.17: 78674047-78782342 on Build GRCh38
Transcripts
NM_001292018.1 NM_001292019.1 NM_004762.3 NM_017456.3 XM_011525475.2 XM_011525476.2 XM_011525477.1 XM_011525478.1 XM_017025336.1 XM_017025337.1 XM_017025338.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5470774 c.1016G>A p.C339Y Chr.17: 78680292-78680292 on Build GRCh38
5218838 c.1093_1095delAAG p.K365delK Chr.17: 78680213-78680215 on Build GRCh38
7412369 c.1038G>A p.R346R Chr.17: 78680270-78680270 on Build GRCh38
5981302 c.972T>A p.F324L Chr.17: 78680336-78680336 on Build GRCh38
5574551 c.1067G>T p.R356L Chr.17: 78680241-78680241 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763986941 Chr.17: 78680352-78680352 on Build GRCh38
rs781545237 Chr.17: 78680250-78680250 on Build GRCh38
rs769569463 Chr.17: 78680179-78680182 on Build GRCh38
rs566292294 Chr.17: 78680399-78680399 on Build GRCh38
rs772351939 Chr.17: 78680321-78680321 on Build GRCh38
rs757994747 Chr.17: 78680248-78680248 on Build GRCh38
rs752715707 Chr.17: 78680153-78680153 on Build GRCh38
rs532825704 Chr.17: 78680379-78680379 on Build GRCh38
rs546055271 Chr.17: 78680155-78680155 on Build GRCh38
rs1046026417 Chr.17: 78680534-78680534 on Build GRCh38
rs936534349 Chr.17: 78680513-78680513 on Build GRCh38
rs559421292 Chr.17: 78680159-78680159 on Build GRCh38
rs560342849 Chr.17: 78680548-78680548 on Build GRCh38
rs756765110 Chr.17: 78680231-78680231 on Build GRCh38
rs767603361 Chr.17: 78680148-78680148 on Build GRCh38
rs759794492 Chr.17: 78680225-78680225 on Build GRCh38
rs756927294 Chr.17: 78680160-78680160 on Build GRCh38
rs1038645171 Chr.17: 78680533-78680533 on Build GRCh38
rs78126198 Chr.17: 78680445-78680445 on Build GRCh38
rs540407961 Chr.17: 78680162-78680162 on Build GRCh38
rs779199282 Chr.17: 78680515-78680515 on Build GRCh38
rs766005465 Chr.17: 78680381-78680381 on Build GRCh38
rs199612995 Chr.17: 78680276-78680276 on Build GRCh38
rs771074240 Chr.17: 78680284-78680284 on Build GRCh38
rs751372453 Chr.17: 78680141-78680141 on Build GRCh38
rs954625813 Chr.17: 78680480-78680480 on Build GRCh38
rs986454159 Chr.17: 78680495-78680495 on Build GRCh38
rs898807389 Chr.17: 78680546-78680546 on Build GRCh38
rs748509940 Chr.17: 78680267-78680267 on Build GRCh38
rs773179688 Chr.17: 78680363-78680363 on Build GRCh38
rs747379713 Chr.17: 78680167-78680167 on Build GRCh38
rs201592047 Chr.17: 78680387-78680387 on Build GRCh38
rs770347515 Chr.17: 78680271-78680271 on Build GRCh38
rs923996269 Chr.17: 78680377-78680377 on Build GRCh38
rs770222675 Chr.17: 78680181-78680181 on Build GRCh38
rs371372394 Chr.17: 78680396-78680396 on Build GRCh38
rs773234558 Chr.17: 78680212-78680212 on Build GRCh38
rs572982752 Chr.17: 78680172-78680172 on Build GRCh38
rs972088215 Chr.17: 78680403-78680403 on Build GRCh38
rs35043968 Chr.17: 78680456-78680457 on Build GRCh38
rs774601118 Chr.17: 78680222-78680222 on Build GRCh38
rs780779855 Chr.17: 78680207-78680207 on Build GRCh38
rs373268325 Chr.17: 78680354-78680354 on Build GRCh38
rs923207957 Chr.17: 78680157-78680157 on Build GRCh38
rs112201022 Chr.17: 78680189-78680189 on Build GRCh38
rs761611498 Chr.17: 78680361-78680361 on Build GRCh38
rs376879430 Chr.17: 78680326-78680326 on Build GRCh38
rs750104667 Chr.17: 78680245-78680245 on Build GRCh38
rs940707643 Chr.17: 78680133-78680133 on Build GRCh38
rs1047437396 Chr.17: 78680568-78680568 on Build GRCh38
rs771108245 Chr.17: 78680221-78680221 on Build GRCh38
rs762839575 Chr.17: 78680144-78680144 on Build GRCh38
rs759384668 Chr.17: 78680142-78680142 on Build GRCh38
rs778324645 Chr.17: 78680273-78680273 on Build GRCh38
rs1018783100 Chr.17: 78680285-78680285 on Build GRCh38
rs913675694 Chr.17: 78680521-78680521 on Build GRCh38
rs907425708 Chr.17: 78680316-78680316 on Build GRCh38
rs994575525 Chr.17: 78680165-78680165 on Build GRCh38
rs781494277 Chr.17: 78680173-78680173 on Build GRCh38
rs760532115 Chr.17: 78680332-78680332 on Build GRCh38
rs748673444 Chr.17: 78680174-78680174 on Build GRCh38
rs964984712 Chr.17: 78680294-78680294 on Build GRCh38
rs764823263 Chr.17: 78680364-78680364 on Build GRCh38
rs370098507 Chr.17: 78680226-78680226 on Build GRCh38
rs753113654 Chr.17: 78680382-78680382 on Build GRCh38
rs774248624 Chr.17: 78680311-78680311 on Build GRCh38
rs189651789 Chr.17: 78680461-78680461 on Build GRCh38
rs916792879 Chr.17: 78680510-78680510 on Build GRCh38
rs758115651 Chr.17: 78680376-78680376 on Build GRCh38
rs200419184 Chr.17: 78680228-78680228 on Build GRCh38
rs915216311 Chr.17: 78680333-78680333 on Build GRCh38
rs753345906 Chr.17: 78680230-78680230 on Build GRCh38
rs1056485698 Chr.17: 78680558-78680558 on Build GRCh38
rs759853440 Chr.17: 78680320-78680320 on Build GRCh38
rs764088076 Chr.17: 78680229-78680229 on Build GRCh38
rs147739265 Chr.17: 78680215-78680215 on Build GRCh38
rs370214371 Chr.17: 78680347-78680347 on Build GRCh38
rs372581895 Chr.17: 78680234-78680234 on Build GRCh38
rs923277937 Chr.17: 78680421-78680421 on Build GRCh38
rs112713276 Chr.17: 78680116-78680116 on Build GRCh38
rs113992427 Chr.17: 78680188-78680188 on Build GRCh38
rs910350485 Chr.17: 78680497-78680497 on Build GRCh38
rs564089431 Chr.17: 78680365-78680365 on Build GRCh38
rs777961575 Chr.17: 78680392-78680392 on Build GRCh38
rs749875162 Chr.17: 78680372-78680372 on Build GRCh38
rs1034485014 Chr.17: 78680346-78680347 on Build GRCh38

View Full Product Details