Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 78701588-78702088 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYTH1
Entrez Gene ID
9267
Gene Name
cytohesin 1
Gene Aliases
B2-1, CYTOHESIN-1, D17S811E, PSCD1, SEC7
UniGene
Hs.191215
Gene Chromosome Location
Chr.17: 78674047-78782342 on Build GRCh38
Transcripts
NM_001292018.1 NM_001292019.1 NM_004762.3 NM_017456.3 XM_011525475.2 XM_011525476.2 XM_011525477.1 XM_011525478.1 XM_017025336.1 XM_017025337.1 XM_017025338.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1610911 c.394G>A p.E132K Chr.17: 78701714-78701714 on Build GRCh38
1202910 c.424G>A p.V142I Chr.17: 78701684-78701684 on Build GRCh38
6639540 c.437G>A p.R146Q Chr.17: 78701671-78701671 on Build GRCh38
7186811 c.370A>G p.I124V Chr.17: 78701738-78701738 on Build GRCh38
3370945 c.401A>T p.H134L Chr.17: 78701707-78701707 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547098361 Chr.17: 78702033-78702033 on Build GRCh38
rs370331141 Chr.17: 78701628-78701628 on Build GRCh38
rs925637294 Chr.17: 78701838-78701838 on Build GRCh38
rs71365529 Chr.17: 78701857-78701858 on Build GRCh38
rs746050843 Chr.17: 78701694-78701694 on Build GRCh38
rs78994692 Chr.17: 78702064-78702064 on Build GRCh38
rs1056607963 Chr.17: 78702051-78702051 on Build GRCh38
rs377102926 Chr.17: 78701736-78701736 on Build GRCh38
rs1036924697 Chr.17: 78701978-78701978 on Build GRCh38
rs760662251 Chr.17: 78701733-78701733 on Build GRCh38
rs753000249 Chr.17: 78701796-78701796 on Build GRCh38
rs748705281 Chr.17: 78701633-78701633 on Build GRCh38
rs564836852 Chr.17: 78701967-78701967 on Build GRCh38
rs527245127 Chr.17: 78701993-78701993 on Build GRCh38
rs542549266 Chr.17: 78701840-78701840 on Build GRCh38
rs367956043 Chr.17: 78701722-78701722 on Build GRCh38
rs771332106 Chr.17: 78701671-78701671 on Build GRCh38
rs375030515 Chr.17: 78701655-78701655 on Build GRCh38
rs137927194 Chr.17: 78701903-78701903 on Build GRCh38
rs985123394 Chr.17: 78701807-78701807 on Build GRCh38
rs139968035 Chr.17: 78701690-78701690 on Build GRCh38
rs989666432 Chr.17: 78701841-78701848 on Build GRCh38
rs756703094 Chr.17: 78701801-78701801 on Build GRCh38
rs531452527 Chr.17: 78701900-78701900 on Build GRCh38
rs779113395 Chr.17: 78701825-78701825 on Build GRCh38
rs371849041 Chr.17: 78701922-78701922 on Build GRCh38
rs749319505 Chr.17: 78701666-78701666 on Build GRCh38
rs1056219208 Chr.17: 78701776-78701776 on Build GRCh38
rs955363212 Chr.17: 78701886-78701886 on Build GRCh38
rs146045013 Chr.17: 78701685-78701685 on Build GRCh38
rs962687572 Chr.17: 78701848-78701848 on Build GRCh38
rs761289633 Chr.17: 78701778-78701778 on Build GRCh38
rs767931171 Chr.17: 78701785-78701785 on Build GRCh38
rs748439872 Chr.17: 78701858-78701858 on Build GRCh38
rs1012115382 Chr.17: 78701834-78701834 on Build GRCh38
rs748211179 Chr.17: 78701828-78701828 on Build GRCh38
rs756511252 Chr.17: 78701642-78701642 on Build GRCh38
rs964555044 Chr.17: 78701936-78701936 on Build GRCh38
rs573579392 Chr.17: 78701775-78701775 on Build GRCh38
rs923569804 Chr.17: 78702012-78702012 on Build GRCh38
rs894936800 Chr.17: 78701826-78701826 on Build GRCh38
rs540284523 Chr.17: 78701615-78701615 on Build GRCh38
rs888413407 Chr.17: 78701930-78701930 on Build GRCh38
rs377093079 Chr.17: 78701970-78701970 on Build GRCh38
rs1025628670 Chr.17: 78701869-78701869 on Build GRCh38
rs764907799 Chr.17: 78701780-78701780 on Build GRCh38
rs758889113 Chr.17: 78701659-78701661 on Build GRCh38
rs562336882 Chr.17: 78701892-78701892 on Build GRCh38
rs939027616 Chr.17: 78702014-78702014 on Build GRCh38
rs757989032 Chr.17: 78701784-78701784 on Build GRCh38
rs371883688 Chr.17: 78701757-78701757 on Build GRCh38
rs1005556834 Chr.17: 78701949-78701949 on Build GRCh38
rs977661777 Chr.17: 78702006-78702006 on Build GRCh38
rs914866026 Chr.17: 78701650-78701650 on Build GRCh38
rs750106508 Chr.17: 78701781-78701781 on Build GRCh38
rs763212348 Chr.17: 78701855-78701856 on Build GRCh38
rs747195152 Chr.17: 78701972-78701972 on Build GRCh38
rs201290324 Chr.17: 78701840-78701841 on Build GRCh38
rs914174406 Chr.17: 78701851-78701852 on Build GRCh38
rs34784342 Chr.17: 78701721-78701721 on Build GRCh38
rs751968650 Chr.17: 78701620-78701620 on Build GRCh38
rs976586199 Chr.17: 78701704-78701704 on Build GRCh38
rs990355982 Chr.17: 78701964-78701964 on Build GRCh38
rs1047029863 Chr.17: 78701878-78701878 on Build GRCh38
rs1055124635 Chr.17: 78701912-78701912 on Build GRCh38
rs994099323 Chr.17: 78701850-78701850 on Build GRCh38
rs894919447 Chr.17: 78702063-78702063 on Build GRCh38
rs1042289 Chr.17: 78701715-78701715 on Build GRCh38

View Full Product Details