Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 80191239-80191739 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARD14
Entrez Gene ID
79092
Gene Name
caspase recruitment domain family member 14
Gene Aliases
BIMP2, CARMA2, PRP, PSORS2, PSS1
UniGene
Hs.675480
Gene Chromosome Location
Chr.17: 80169992-80209331 on Build GRCh38
Transcripts
NM_001257970.1 NM_024110.4 NM_052819.2 NR_047566.1 XM_011525212.1 XM_011525213.1 XM_011525215.1 XM_011525216.1 XM_011525217.1 XM_011525218.2 XM_017025057.1 XM_017025058.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
708749 c.1116G>C p.Q372H Chr.17: 80191349-80191349 on Build GRCh38
5741174 c.446G>A p.R149H Chr.17: 80191390-80191390 on Build GRCh38
344612 c.1152C>T p.S384S Chr.17: 80191385-80191385 on Build GRCh38
5528141 c.1151C>T p.S384F Chr.17: 80191384-80191384 on Build GRCh38
7174285 c.1171G>A p.E391K Chr.17: 80191404-80191404 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771844235 Chr.17: 80191318-80191318 on Build GRCh38
rs1004291201 Chr.17: 80191591-80191591 on Build GRCh38
rs775079912 Chr.17: 80191290-80191290 on Build GRCh38
rs555307438 Chr.17: 80191574-80191574 on Build GRCh38
rs549747651 Chr.17: 80191333-80191333 on Build GRCh38
rs750807585 Chr.17: 80191518-80191518 on Build GRCh38
rs749499170 Chr.17: 80191311-80191311 on Build GRCh38
rs757171438 Chr.17: 80191640-80191640 on Build GRCh38
rs772930067 Chr.17: 80191468-80191468 on Build GRCh38
rs1056930033 Chr.17: 80191475-80191475 on Build GRCh38
rs978844114 Chr.17: 80191617-80191617 on Build GRCh38
rs577888174 Chr.17: 80191443-80191443 on Build GRCh38
rs769603509 Chr.17: 80191466-80191466 on Build GRCh38
rs539249557 Chr.17: 80191412-80191412 on Build GRCh38
rs762265566 Chr.17: 80191512-80191512 on Build GRCh38
rs1029013014 Chr.17: 80191605-80191605 on Build GRCh38
rs372109073 Chr.17: 80191292-80191292 on Build GRCh38
rs754253068 Chr.17: 80191334-80191334 on Build GRCh38
rs8068452 Chr.17: 80191712-80191712 on Build GRCh38
rs8068433 Chr.17: 80191657-80191657 on Build GRCh38
rs753226666 Chr.17: 80191341-80191341 on Build GRCh38
rs770558404 Chr.17: 80191484-80191484 on Build GRCh38
rs373435882 Chr.17: 80191325-80191325 on Build GRCh38
rs141698692 Chr.17: 80191324-80191324 on Build GRCh38
rs948663357 Chr.17: 80191494-80191494 on Build GRCh38
rs377287985 Chr.17: 80191308-80191308 on Build GRCh38
rs748956995 Chr.17: 80191483-80191483 on Build GRCh38
rs995633166 Chr.17: 80191371-80191371 on Build GRCh38
rs145763239 Chr.17: 80191530-80191530 on Build GRCh38
rs189681150 Chr.17: 80191291-80191291 on Build GRCh38
rs372678467 Chr.17: 80191305-80191305 on Build GRCh38
rs978430125 Chr.17: 80191444-80191444 on Build GRCh38
rs999064367 Chr.17: 80191283-80191283 on Build GRCh38
rs573718530 Chr.17: 80191582-80191582 on Build GRCh38
rs147432650 Chr.17: 80191421-80191421 on Build GRCh38
rs529132871 Chr.17: 80191641-80191641 on Build GRCh38
rs1018965158 Chr.17: 80191671-80191671 on Build GRCh38
rs968960783 Chr.17: 80191611-80191611 on Build GRCh38
rs995930914 Chr.17: 80191556-80191556 on Build GRCh38
rs978994615 Chr.17: 80191548-80191548 on Build GRCh38
rs563908407 Chr.17: 80191634-80191634 on Build GRCh38
rs375882704 Chr.17: 80191332-80191332 on Build GRCh38
rs774539500 Chr.17: 80191279-80191279 on Build GRCh38
rs779208769 Chr.17: 80191361-80191361 on Build GRCh38
rs777801345 Chr.17: 80191493-80191493 on Build GRCh38
rs540202996 Chr.17: 80191600-80191600 on Build GRCh38
rs551983837 Chr.17: 80191293-80191293 on Build GRCh38
rs975689507 Chr.17: 80191603-80191603 on Build GRCh38
rs754322294 Chr.17: 80191352-80191352 on Build GRCh38
rs780034490 Chr.17: 80191384-80191384 on Build GRCh38
rs138683060 Chr.17: 80191672-80191672 on Build GRCh38
rs772222983 Chr.17: 80191379-80191379 on Build GRCh38
rs934391099 Chr.17: 80191589-80191589 on Build GRCh38
rs761260235 Chr.17: 80191415-80191415 on Build GRCh38
rs149287760 Chr.17: 80191411-80191411 on Build GRCh38
rs368517365 Chr.17: 80191295-80191295 on Build GRCh38
rs965651297 Chr.17: 80191423-80191423 on Build GRCh38
rs899053280 Chr.17: 80191537-80191537 on Build GRCh38
rs1028274779 Chr.17: 80191685-80191685 on Build GRCh38
rs549916464 Chr.17: 80191668-80191668 on Build GRCh38
rs1026946480 Chr.17: 80191565-80191565 on Build GRCh38
rs760670766 Chr.17: 80191602-80191602 on Build GRCh38
rs138208838 Chr.17: 80191267-80191267 on Build GRCh38
rs560385337 Chr.17: 80191632-80191632 on Build GRCh38
rs754854916 Chr.17: 80191458-80191458 on Build GRCh38
rs528883443 Chr.17: 80191679-80191679 on Build GRCh38
rs374706201 Chr.17: 80191280-80191280 on Build GRCh38
rs777308288 Chr.17: 80191497-80191497 on Build GRCh38
rs920918867 Chr.17: 80191302-80191302 on Build GRCh38
rs1041135640 Chr.17: 80191716-80191716 on Build GRCh38
rs369150206 Chr.17: 80191404-80191404 on Build GRCh38
rs746925458 Chr.17: 80191389-80191389 on Build GRCh38
rs996577442 Chr.17: 80191682-80191682 on Build GRCh38
rs368798956 Chr.17: 80191309-80191309 on Build GRCh38
rs545266337 Chr.17: 80191631-80191631 on Build GRCh38
rs767874623 Chr.17: 80191340-80191340 on Build GRCh38
rs865854432 Chr.17: 80191659-80191659 on Build GRCh38
rs758536769 Chr.17: 80191451-80191451 on Build GRCh38
rs1044213141 Chr.17: 80191645-80191645 on Build GRCh38
rs559779821 Chr.17: 80191601-80191601 on Build GRCh38
rs747156081 Chr.17: 80191455-80191455 on Build GRCh38
rs139740020 Chr.17: 80191425-80191425 on Build GRCh38
rs754371883 Chr.17: 80191298-80191299 on Build GRCh38
rs756736748 Chr.17: 80191351-80191351 on Build GRCh38
rs757353801 Chr.17: 80191354-80191354 on Build GRCh38
rs564433759 Chr.17: 80191465-80191465 on Build GRCh38
rs1019832214 Chr.17: 80191595-80191595 on Build GRCh38
rs768406506 Chr.17: 80191326-80191326 on Build GRCh38
rs373415766 Chr.17: 80191460-80191460 on Build GRCh38
rs375465348 Chr.17: 80191294-80191294 on Build GRCh38
rs144636413 Chr.17: 80191431-80191431 on Build GRCh38
rs761614278 Chr.17: 80191399-80191399 on Build GRCh38
rs755377689 Chr.17: 80191296-80191296 on Build GRCh38
rs201501775 Chr.17: 80191424-80191424 on Build GRCh38
rs535721220 Chr.17: 80191390-80191390 on Build GRCh38
rs150536049 Chr.17: 80191367-80191367 on Build GRCh38
rs578225656 Chr.17: 80191609-80191609 on Build GRCh38
rs147466598 Chr.17: 80191432-80191432 on Build GRCh38
rs924294720 Chr.17: 80191620-80191620 on Build GRCh38
rs745964131 Chr.17: 80191316-80191316 on Build GRCh38
rs142309150 Chr.17: 80191289-80191289 on Build GRCh38
rs765609276 Chr.17: 80191514-80191514 on Build GRCh38
rs74951924 Chr.17: 80191403-80191403 on Build GRCh38
rs778853453 Chr.17: 80191313-80191313 on Build GRCh38
rs374504738 Chr.17: 80191485-80191485 on Build GRCh38
rs771338077 Chr.17: 80191312-80191312 on Build GRCh38
rs749480979 Chr.17: 80191331-80191331 on Build GRCh38
rs930542492 Chr.17: 80191660-80191660 on Build GRCh38
rs758639381 Chr.17: 80191519-80191519 on Build GRCh38
rs887102547 Chr.17: 80191681-80191681 on Build GRCh38

View Full Product Details