Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 21765688-21766186 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIB1
Entrez Gene ID
57534
Gene Name
mindbomb E3 ubiquitin protein ligase 1
Gene Aliases
DIP-1, DIP1, LVNC7, MIB, ZZANK2, ZZZ6
UniGene
Hs.140903
Gene Chromosome Location
Chr.18: 21740793-21870957 on Build GRCh38
Transcripts
NM_020774.3 XM_017025874.1 XM_017025875.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3524389 c.293G>A p.W98* Chr.18: 21765835-21765835 on Build GRCh38
1680262 c.266G>A p.R89H Chr.18: 21765808-21765808 on Build GRCh38
6046430 c.390G>T p.P130P Chr.18: 21765932-21765932 on Build GRCh38
3388387 c.237G>A p.K79K Chr.18: 21765779-21765779 on Build GRCh38
1215061 c.385A>G p.T129A Chr.18: 21765927-21765927 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs34478849 Chr.18: 21765902-21765902 on Build GRCh38
rs763042215 Chr.18: 21765898-21765899 on Build GRCh38
rs948633120 Chr.18: 21765852-21765852 on Build GRCh38
rs746295442 Chr.18: 21765869-21765869 on Build GRCh38
rs780810168 Chr.18: 21765854-21765854 on Build GRCh38
rs764303494 Chr.18: 21765751-21765751 on Build GRCh38
rs918672443 Chr.18: 21766115-21766115 on Build GRCh38
rs965384598 Chr.18: 21766100-21766100 on Build GRCh38
rs985041236 Chr.18: 21766150-21766150 on Build GRCh38
rs770637596 Chr.18: 21765850-21765850 on Build GRCh38
rs775840663 Chr.18: 21765741-21765741 on Build GRCh38
rs142852766 Chr.18: 21765823-21765823 on Build GRCh38
rs954461170 Chr.18: 21766049-21766049 on Build GRCh38
rs998773897 Chr.18: 21765930-21765930 on Build GRCh38
rs117138335 Chr.18: 21766018-21766018 on Build GRCh38
rs777278397 Chr.18: 21765819-21765819 on Build GRCh38
rs551926425 Chr.18: 21765750-21765750 on Build GRCh38
rs573330313 Chr.18: 21765893-21765893 on Build GRCh38
rs769232281 Chr.18: 21765912-21765912 on Build GRCh38
rs371546190 Chr.18: 21765754-21765754 on Build GRCh38
rs374307694 Chr.18: 21765759-21765759 on Build GRCh38
rs752605883 Chr.18: 21765808-21765808 on Build GRCh38
rs190657514 Chr.18: 21765918-21765918 on Build GRCh38
rs551715027 Chr.18: 21766102-21766102 on Build GRCh38
rs778947880 Chr.18: 21765807-21765807 on Build GRCh38
rs752477868 Chr.18: 21765945-21765945 on Build GRCh38
rs760908907 Chr.18: 21765932-21765932 on Build GRCh38
rs767897972 Chr.18: 21765926-21765926 on Build GRCh38
rs780299943 Chr.18: 21765975-21765975 on Build GRCh38
rs775887137 Chr.18: 21765909-21765909 on Build GRCh38
rs944922433 Chr.18: 21766117-21766117 on Build GRCh38
rs1007168003 Chr.18: 21766057-21766057 on Build GRCh38
rs756739772 Chr.18: 21765969-21765969 on Build GRCh38
rs563710709 Chr.18: 21766014-21766014 on Build GRCh38
rs750577728 Chr.18: 21765931-21765931 on Build GRCh38
rs773394183 Chr.18: 21765856-21765857 on Build GRCh38
rs561166598 Chr.18: 21765915-21765915 on Build GRCh38
rs1019813722 Chr.18: 21766099-21766099 on Build GRCh38
rs780865224 Chr.18: 21765970-21765970 on Build GRCh38
rs891512175 Chr.18: 21765982-21765982 on Build GRCh38
rs762143417 Chr.18: 21765755-21765755 on Build GRCh38
rs3017047 Chr.18: 21765743-21765743 on Build GRCh38
rs368217297 Chr.18: 21765966-21765966 on Build GRCh38
rs758336318 Chr.18: 21765959-21765959 on Build GRCh38
rs748393871 Chr.18: 21765997-21765997 on Build GRCh38
rs751128298 Chr.18: 21765967-21765967 on Build GRCh38
rs778155689 Chr.18: 21765845-21765846 on Build GRCh38
rs749604317 Chr.18: 21765988-21765988 on Build GRCh38
rs909020893 Chr.18: 21766161-21766161 on Build GRCh38
rs770026342 Chr.18: 21765875-21765875 on Build GRCh38
rs185843324 Chr.18: 21765791-21765791 on Build GRCh38
rs375820093 Chr.18: 21765972-21765972 on Build GRCh38
rs541957058 Chr.18: 21765910-21765910 on Build GRCh38
rs768833397 Chr.18: 21765733-21765735 on Build GRCh38
rs748268854 Chr.18: 21765824-21765825 on Build GRCh38
rs558920020 Chr.18: 21766116-21766116 on Build GRCh38
rs753722259 Chr.18: 21765760-21765760 on Build GRCh38
rs952791770 Chr.18: 21766149-21766149 on Build GRCh38
rs531985535 Chr.18: 21766033-21766033 on Build GRCh38
rs151095403 Chr.18: 21765831-21765831 on Build GRCh38
rs3017048 Chr.18: 21765971-21765971 on Build GRCh38
rs111741828 Chr.18: 21765803-21765803 on Build GRCh38
rs1031468546 Chr.18: 21765938-21765938 on Build GRCh38
rs776924209 Chr.18: 21765773-21765774 on Build GRCh38
rs528217517 Chr.18: 21766127-21766127 on Build GRCh38

View Full Product Details