Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43367890-43368388 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELOVL1
Entrez Gene ID
64834
Gene Name
ELOVL fatty acid elongase 1
Gene Aliases
CGI-88, Ssc1
UniGene
Hs.25597
Gene Chromosome Location
Chr.1: 43363397-43368074 on Build GRCh38
Transcripts
NM_001256401.1 NM_001256402.1 NM_022821.3 NR_046117.1

SNP Detail

SNP ID Chromosome Location
rs1023488985 Chr.1: 43368199-43368199 on Build GRCh38
rs1015463270 Chr.1: 43368266-43368266 on Build GRCh38
rs571930880 Chr.1: 43368197-43368197 on Build GRCh38
rs949069568 Chr.1: 43368220-43368220 on Build GRCh38
rs931781002 Chr.1: 43368161-43368161 on Build GRCh38
rs1042674082 Chr.1: 43367918-43367918 on Build GRCh38
rs1056968239 Chr.1: 43367978-43367978 on Build GRCh38
rs867990337 Chr.1: 43368182-43368182 on Build GRCh38
rs75251249 Chr.1: 43368307-43368307 on Build GRCh38
rs903608795 Chr.1: 43367976-43367976 on Build GRCh38
rs1001713724 Chr.1: 43368084-43368084 on Build GRCh38
rs979247565 Chr.1: 43368170-43368170 on Build GRCh38
rs925125508 Chr.1: 43368344-43368344 on Build GRCh38
rs12731821 Chr.1: 43367974-43367974 on Build GRCh38
rs1045982563 Chr.1: 43368237-43368237 on Build GRCh38
rs1043417336 Chr.1: 43367967-43367967 on Build GRCh38
rs1014913179 Chr.1: 43368139-43368139 on Build GRCh38
rs189893059 Chr.1: 43368274-43368274 on Build GRCh38
rs1024595437 Chr.1: 43368028-43368028 on Build GRCh38
rs12730626 Chr.1: 43367949-43367949 on Build GRCh38
rs965979479 Chr.1: 43368303-43368303 on Build GRCh38
rs994844783 Chr.1: 43368214-43368214 on Build GRCh38
rs1028837131 Chr.1: 43368313-43368314 on Build GRCh38
rs963464375 Chr.1: 43368202-43368202 on Build GRCh38
rs1055499362 Chr.1: 43368123-43368124 on Build GRCh38
rs966051643 Chr.1: 43368140-43368140 on Build GRCh38
rs903117273 Chr.1: 43367936-43367936 on Build GRCh38
rs538932049 Chr.1: 43368189-43368189 on Build GRCh38
rs566545829 Chr.1: 43368315-43368315 on Build GRCh38
rs935075690 Chr.1: 43368184-43368184 on Build GRCh38
rs998689904 Chr.1: 43367938-43367938 on Build GRCh38
rs895667130 Chr.1: 43368191-43368191 on Build GRCh38
rs569803051 Chr.1: 43368092-43368092 on Build GRCh38
rs921027029 Chr.1: 43368300-43368300 on Build GRCh38
rs185142956 Chr.1: 43368194-43368194 on Build GRCh38
rs1014499493 Chr.1: 43367990-43367990 on Build GRCh38
rs918989307 Chr.1: 43367973-43367973 on Build GRCh38
rs934962689 Chr.1: 43367977-43367977 on Build GRCh38
rs906128167 Chr.1: 43368242-43368242 on Build GRCh38
rs965273418 Chr.1: 43368264-43368264 on Build GRCh38
rs1026278354 Chr.1: 43368226-43368226 on Build GRCh38
rs953314725 Chr.1: 43368341-43368341 on Build GRCh38
rs536240749 Chr.1: 43368144-43368144 on Build GRCh38
rs931856356 Chr.1: 43367948-43367948 on Build GRCh38
rs534008044 Chr.1: 43368245-43368245 on Build GRCh38
rs952183813 Chr.1: 43368103-43368103 on Build GRCh38
rs535906284 Chr.1: 43367983-43367983 on Build GRCh38
rs550043835 Chr.1: 43368039-43368039 on Build GRCh38
rs879114374 Chr.1: 43368002-43368002 on Build GRCh38
rs950691943 Chr.1: 43368232-43368232 on Build GRCh38
rs929014351 Chr.1: 43368178-43368178 on Build GRCh38
rs921853464 Chr.1: 43368152-43368152 on Build GRCh38
rs912136522 Chr.1: 43368259-43368259 on Build GRCh38
rs974918085 Chr.1: 43368292-43368292 on Build GRCh38

View Full Product Details