Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3000423-3000809 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TLE2
Entrez Gene ID
7089
Gene Name
transducin like enhancer of split 2
Gene Aliases
ESG, ESG2, GRG2
UniGene
Hs.332173
Gene Chromosome Location
Chr.19: 2997638-3047635 on Build GRCh38
Transcripts
NM_001144761.1 NM_001144762.1 NM_001300846.1 NM_003260.4 XM_006722864.2 XM_011528230.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3531996 c.2094G>A p.W698* Chr.19: 3000677-3000677 on Build GRCh38
4534256 c.2068G>A p.G690R Chr.19: 3000703-3000703 on Build GRCh38
5493765 c.2051G>A p.R684Q Chr.19: 3000720-3000720 on Build GRCh38
4733895 c.2119T>C p.F707L Chr.19: 3000652-3000652 on Build GRCh38
4476858 c.2102C>T p.P701L Chr.19: 3000669-3000669 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370836607 Chr.19: 3000704-3000704 on Build GRCh38
rs796744151 Chr.19: 3000534-3000534 on Build GRCh38
rs751695993 Chr.19: 3000630-3000630 on Build GRCh38
rs900530554 Chr.19: 3000743-3000743 on Build GRCh38
rs969675936 Chr.19: 3000569-3000569 on Build GRCh38
rs182457841 Chr.19: 3000502-3000502 on Build GRCh38
rs372773787 Chr.19: 3000729-3000729 on Build GRCh38
rs199797446 Chr.19: 3000760-3000760 on Build GRCh38
rs769449049 Chr.19: 3000698-3000698 on Build GRCh38
rs377009962 Chr.19: 3000757-3000757 on Build GRCh38
rs759754198 Chr.19: 3000721-3000721 on Build GRCh38
rs966514585 Chr.19: 3000782-3000782 on Build GRCh38
rs912946226 Chr.19: 3000747-3000747 on Build GRCh38
rs756192867 Chr.19: 3000661-3000661 on Build GRCh38
rs1054799942 Chr.19: 3000459-3000459 on Build GRCh38
rs771341717 Chr.19: 3000597-3000597 on Build GRCh38
rs563325243 Chr.19: 3000464-3000464 on Build GRCh38
rs113268472 Chr.19: 3000559-3000559 on Build GRCh38
rs556895399 Chr.19: 3000759-3000759 on Build GRCh38
rs1021241346 Chr.19: 3000778-3000778 on Build GRCh38
rs986657922 Chr.19: 3000776-3000776 on Build GRCh38
rs974845118 Chr.19: 3000517-3000517 on Build GRCh38
rs374105223 Chr.19: 3000668-3000668 on Build GRCh38
rs769785780 Chr.19: 3000623-3000623 on Build GRCh38
rs779376099 Chr.19: 3000669-3000669 on Build GRCh38
rs117008739 Chr.19: 3000665-3000665 on Build GRCh38
rs776338277 Chr.19: 3000619-3000619 on Build GRCh38
rs372296346 Chr.19: 3000467-3000467 on Build GRCh38
rs200430664 Chr.19: 3000758-3000758 on Build GRCh38
rs761128385 Chr.19: 3000773-3000773 on Build GRCh38
rs201746954 Chr.19: 3000671-3000671 on Build GRCh38
rs368876823 Chr.19: 3000602-3000602 on Build GRCh38
rs754268778 Chr.19: 3000755-3000755 on Build GRCh38
rs757839360 Chr.19: 3000756-3000756 on Build GRCh38
rs988546040 Chr.19: 3000604-3000604 on Build GRCh38
rs774264336 Chr.19: 3000714-3000714 on Build GRCh38
rs1026582704 Chr.19: 3000468-3000468 on Build GRCh38
rs201967670 Chr.19: 3000637-3000637 on Build GRCh38
rs960689374 Chr.19: 3000556-3000556 on Build GRCh38
rs188353112 Chr.19: 3000545-3000545 on Build GRCh38
rs769393885 Chr.19: 3000767-3000767 on Build GRCh38
rs1006230747 Chr.19: 3000483-3000483 on Build GRCh38
rs35580021 Chr.19: 3000493-3000493 on Build GRCh38
rs766224117 Chr.19: 3000628-3000628 on Build GRCh38
rs937660699 Chr.19: 3000640-3000640 on Build GRCh38
rs768258092 Chr.19: 3000609-3000609 on Build GRCh38
rs768488905 Chr.19: 3000672-3000672 on Build GRCh38
rs992213351 Chr.19: 3000557-3000562 on Build GRCh38
rs1014992820 Chr.19: 3000552-3000552 on Build GRCh38
rs200341404 Chr.19: 3000601-3000601 on Build GRCh38
rs552962403 Chr.19: 3000788-3000788 on Build GRCh38
rs767663886 Chr.19: 3000727-3000727 on Build GRCh38
rs767574323 Chr.19: 3000638-3000638 on Build GRCh38
rs979980934 Chr.19: 3000595-3000595 on Build GRCh38
rs951011621 Chr.19: 3000472-3000472 on Build GRCh38
rs770605624 Chr.19: 3000713-3000713 on Build GRCh38
rs867675312 Chr.19: 3000485-3000485 on Build GRCh38
rs778196891 Chr.19: 3000664-3000664 on Build GRCh38
rs764190410 Chr.19: 3000739-3000739 on Build GRCh38
rs773394671 Chr.19: 3000749-3000750 on Build GRCh38
rs761836627 Chr.19: 3000620-3000620 on Build GRCh38
rs747603021 Chr.19: 3000692-3000692 on Build GRCh38
rs547759793 Chr.19: 3000624-3000624 on Build GRCh38
rs772980855 Chr.19: 3000703-3000703 on Build GRCh38
rs143713547 Chr.19: 3000657-3000657 on Build GRCh38
rs1004939386 Chr.19: 3000544-3000544 on Build GRCh38
rs376590696 Chr.19: 3000683-3000683 on Build GRCh38
rs550714164 Chr.19: 3000785-3000793 on Build GRCh38
rs887592178 Chr.19: 3000647-3000647 on Build GRCh38
rs780941235 Chr.19: 3000766-3000766 on Build GRCh38
rs555779738 Chr.19: 3000738-3000738 on Build GRCh38
rs777006632 Chr.19: 3000610-3000610 on Build GRCh38
rs1052748128 Chr.19: 3000463-3000463 on Build GRCh38

View Full Product Details