Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3733990-3734534 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TJP3
Entrez Gene ID
27134
Gene Name
tight junction protein 3
Gene Aliases
ZO-3, ZO3
UniGene
Hs.25527
Gene Chromosome Location
Chr.19: 3708337-3750813 on Build GRCh38
Transcripts
NM_001267560.1 NM_001267561.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3200129 c.1052G>A p.R351Q Chr.19: 3734402-3734402 on Build GRCh38
5793974 c.1027C>T p.P343S Chr.19: 3734377-3734377 on Build GRCh38
4904914 c.968C>T p.P323L Chr.19: 3734318-3734318 on Build GRCh38
1711613 c.1015C>T p.P339S Chr.19: 3734365-3734365 on Build GRCh38
4883555 c.1064C>A p.A355D Chr.19: 3734414-3734414 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs74325938 Chr.19: 3734033-3734033 on Build GRCh38
rs566933653 Chr.19: 3734319-3734319 on Build GRCh38
rs3746122 Chr.19: 3734177-3734177 on Build GRCh38
rs142477359 Chr.19: 3734389-3734389 on Build GRCh38
rs150849759 Chr.19: 3734121-3734121 on Build GRCh38
rs774489104 Chr.19: 3734068-3734068 on Build GRCh38
rs759346686 Chr.19: 3734167-3734167 on Build GRCh38
rs777157593 Chr.19: 3734483-3734483 on Build GRCh38
rs111289034 Chr.19: 3734322-3734323 on Build GRCh38
rs373607850 Chr.19: 3734332-3734332 on Build GRCh38
rs769068080 Chr.19: 3734478-3734478 on Build GRCh38
rs554862128 Chr.19: 3734417-3734417 on Build GRCh38
rs768038758 Chr.19: 3734373-3734373 on Build GRCh38
rs759560319 Chr.19: 3734360-3734360 on Build GRCh38
rs772080741 Chr.19: 3734424-3734424 on Build GRCh38
rs746985820 Chr.19: 3734341-3734341 on Build GRCh38
rs139279982 Chr.19: 3734166-3734166 on Build GRCh38
rs146845184 Chr.19: 3734401-3734401 on Build GRCh38
rs756908281 Chr.19: 3734257-3734257 on Build GRCh38
rs776839557 Chr.19: 3734434-3734434 on Build GRCh38
rs747982782 Chr.19: 3734407-3734407 on Build GRCh38
rs757983996 Chr.19: 3734337-3734337 on Build GRCh38
rs3746121 Chr.19: 3734301-3734301 on Build GRCh38
rs777714338 Chr.19: 3734458-3734458 on Build GRCh38
rs372481606 Chr.19: 3734335-3734335 on Build GRCh38
rs757111841 Chr.19: 3734467-3734476 on Build GRCh38
rs748006541 Chr.19: 3734352-3734352 on Build GRCh38
rs148792478 Chr.19: 3734355-3734355 on Build GRCh38
rs750108096 Chr.19: 3734451-3734451 on Build GRCh38
rs1011825088 Chr.19: 3734512-3734512 on Build GRCh38
rs1031775267 Chr.19: 3734124-3734124 on Build GRCh38
rs901989779 Chr.19: 3734322-3734322 on Build GRCh38
rs914328945 Chr.19: 3734178-3734178 on Build GRCh38
rs201265630 Chr.19: 3734275-3734275 on Build GRCh38
rs200789109 Chr.19: 3734240-3734240 on Build GRCh38
rs202057643 Chr.19: 3734354-3734354 on Build GRCh38
rs766161289 Chr.19: 3734435-3734435 on Build GRCh38
rs1039090316 Chr.19: 3734491-3734491 on Build GRCh38
rs757955059 Chr.19: 3734390-3734390 on Build GRCh38
rs573009865 Chr.19: 3734075-3734075 on Build GRCh38
rs1036948445 Chr.19: 3734231-3734231 on Build GRCh38
rs760001909 Chr.19: 3734321-3734321 on Build GRCh38
rs749066214 Chr.19: 3734460-3734460 on Build GRCh38
rs979948074 Chr.19: 3734194-3734194 on Build GRCh38
rs746928778 Chr.19: 3734284-3734284 on Build GRCh38
rs772205601 Chr.19: 3734469-3734469 on Build GRCh38
rs573291095 Chr.19: 3734018-3734018 on Build GRCh38
rs969773764 Chr.19: 3734182-3734182 on Build GRCh38
rs552525132 Chr.19: 3734195-3734195 on Build GRCh38
rs148722853 Chr.19: 3734346-3734346 on Build GRCh38
rs530068179 Chr.19: 3734155-3734155 on Build GRCh38
rs780152576 Chr.19: 3734276-3734276 on Build GRCh38
rs199643630 Chr.19: 3734410-3734410 on Build GRCh38
rs547122072 Chr.19: 3734161-3734161 on Build GRCh38
rs755977057 Chr.19: 3734452-3734452 on Build GRCh38
rs773501089 Chr.19: 3734353-3734353 on Build GRCh38
rs777520507 Chr.19: 3734272-3734278 on Build GRCh38
rs145208682 Chr.19: 3734345-3734345 on Build GRCh38
rs918512594 Chr.19: 3734441-3734441 on Build GRCh38
rs994965762 Chr.19: 3734049-3734049 on Build GRCh38
rs902041121 Chr.19: 3734501-3734501 on Build GRCh38
rs762176022 Chr.19: 3734298-3734298 on Build GRCh38
rs771162770 Chr.19: 3734422-3734422 on Build GRCh38
rs149989552 Chr.19: 3734289-3734289 on Build GRCh38
rs184889448 Chr.19: 3734320-3734320 on Build GRCh38
rs747599567 Chr.19: 3734472-3734472 on Build GRCh38
rs531638340 Chr.19: 3734508-3734508 on Build GRCh38
rs761080049 Chr.19: 3734427-3734427 on Build GRCh38
rs763630445 Chr.19: 3734290-3734290 on Build GRCh38
rs774512601 Chr.19: 3734423-3734423 on Build GRCh38
rs950875929 Chr.19: 3734069-3734069 on Build GRCh38
rs763050637 Chr.19: 3734064-3734064 on Build GRCh38
rs761114012 Chr.19: 3734386-3734386 on Build GRCh38
rs540600865 Chr.19: 3734503-3734503 on Build GRCh38
rs372204758 Chr.19: 3734380-3734380 on Build GRCh38
rs775399143 Chr.19: 3734470-3734470 on Build GRCh38
rs755997157 Chr.19: 3734409-3734409 on Build GRCh38
rs377468613 Chr.19: 3734333-3734333 on Build GRCh38
rs200603772 Chr.19: 3734402-3734402 on Build GRCh38
rs1047963020 Chr.19: 3734312-3734312 on Build GRCh38
rs779704406 Chr.19: 3734340-3734340 on Build GRCh38
rs553055182 Chr.19: 3734334-3734334 on Build GRCh38
rs3746123 Chr.19: 3734066-3734066 on Build GRCh38
rs201579834 Chr.19: 3734436-3734436 on Build GRCh38
rs763102307 Chr.19: 3734309-3734309 on Build GRCh38
rs780861582 Chr.19: 3734497-3734497 on Build GRCh38
rs1021273641 Chr.19: 3734368-3734368 on Build GRCh38
rs764559097 Chr.19: 3734429-3734429 on Build GRCh38
rs758106497 Chr.19: 3734227-3734227 on Build GRCh38
rs1018304221 Chr.19: 3734023-3734023 on Build GRCh38
rs773269955 Chr.19: 3734304-3734304 on Build GRCh38
rs746076685 Chr.19: 3734468-3734468 on Build GRCh38
rs779116819 Chr.19: 3734462-3734462 on Build GRCh38
rs978599734 Chr.19: 3734141-3734141 on Build GRCh38
rs1053675150 Chr.19: 3734507-3734507 on Build GRCh38
rs149590234 Chr.19: 3734176-3734176 on Build GRCh38
rs377122533 Chr.19: 3734425-3734425 on Build GRCh38
rs778377219 Chr.19: 3734258-3734258 on Build GRCh38
rs745919605 Chr.19: 3734272-3734272 on Build GRCh38
rs563181950 Chr.19: 3734081-3734081 on Build GRCh38
rs190389546 Chr.19: 3734100-3734100 on Build GRCh38
rs544849058 Chr.19: 3734076-3734076 on Build GRCh38
rs780969672 Chr.19: 3734467-3734467 on Build GRCh38

View Full Product Details