Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12922390-12922891 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FARSA
Entrez Gene ID
2193
Gene Name
phenylalanyl-tRNA synthetase alpha subunit
Gene Aliases
CML33, FARSL, FARSLA, FRSA, PheHA
UniGene
Hs.23111
Gene Chromosome Location
Chr.19: 12922470-12933744 on Build GRCh38
Transcripts
NM_004461.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4623829 c.1486G>A p.D496N Chr.19: 12922789-12922789 on Build GRCh38
7401014 c.1404A>G p.K468K Chr.19: 12922871-12922871 on Build GRCh38
1680715 c.1523C>T p.A508V Chr.19: 12922752-12922752 on Build GRCh38
4564721 c.1390_1391CC>TT p.P464L Chr.19: 12922884-12922885 on Build GRCh38
6660618 c.1480C>T p.R494C Chr.19: 12922795-12922795 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766975205 Chr.19: 12922704-12922704 on Build GRCh38
rs757649775 Chr.19: 12922597-12922597 on Build GRCh38
rs984678253 Chr.19: 12922578-12922578 on Build GRCh38
rs986516375 Chr.19: 12922484-12922484 on Build GRCh38
rs35636827 Chr.19: 12922715-12922716 on Build GRCh38
rs947150434 Chr.19: 12922415-12922415 on Build GRCh38
rs200199231 Chr.19: 12922846-12922846 on Build GRCh38
rs902914426 Chr.19: 12922459-12922459 on Build GRCh38
rs370197964 Chr.19: 12922784-12922784 on Build GRCh38
rs953680607 Chr.19: 12922456-12922456 on Build GRCh38
rs369470191 Chr.19: 12922437-12922437 on Build GRCh38
rs200922311 Chr.19: 12922716-12922716 on Build GRCh38
rs981446666 Chr.19: 12922806-12922806 on Build GRCh38
rs780245454 Chr.19: 12922642-12922642 on Build GRCh38
rs774024421 Chr.19: 12922795-12922795 on Build GRCh38
rs781595705 Chr.19: 12922489-12922489 on Build GRCh38
rs928608976 Chr.19: 12922831-12922831 on Build GRCh38
rs758534022 Chr.19: 12922637-12922637 on Build GRCh38
rs552230909 Chr.19: 12922426-12922426 on Build GRCh38
rs765665257 Chr.19: 12922851-12922851 on Build GRCh38
rs562246223 Chr.19: 12922411-12922411 on Build GRCh38
rs755241350 Chr.19: 12922868-12922868 on Build GRCh38
rs758719242 Chr.19: 12922752-12922752 on Build GRCh38
rs540075798 Chr.19: 12922779-12922779 on Build GRCh38
rs373657447 Chr.19: 12922852-12922852 on Build GRCh38
rs546977235 Chr.19: 12922635-12922635 on Build GRCh38
rs746184933 Chr.19: 12922507-12922507 on Build GRCh38
rs779115123 Chr.19: 12922614-12922614 on Build GRCh38
rs796135269 Chr.19: 12922508-12922508 on Build GRCh38
rs547783887 Chr.19: 12922746-12922746 on Build GRCh38
rs749038897 Chr.19: 12922698-12922698 on Build GRCh38
rs762285339 Chr.19: 12922719-12922719 on Build GRCh38
rs919052779 Chr.19: 12922565-12922565 on Build GRCh38
rs143111515 Chr.19: 12922751-12922751 on Build GRCh38
rs769683055 Chr.19: 12922691-12922691 on Build GRCh38
rs772764793 Chr.19: 12922716-12922716 on Build GRCh38
rs754294337 Chr.19: 12922591-12922591 on Build GRCh38
rs781296962 Chr.19: 12922658-12922658 on Build GRCh38
rs781403199 Chr.19: 12922774-12922774 on Build GRCh38
rs947596010 Chr.19: 12922843-12922843 on Build GRCh38
rs372053316 Chr.19: 12922699-12922699 on Build GRCh38
rs896206519 Chr.19: 12922413-12922413 on Build GRCh38
rs745489435 Chr.19: 12922801-12922801 on Build GRCh38
rs772871450 Chr.19: 12922694-12922694 on Build GRCh38
rs139040383 Chr.19: 12922863-12922863 on Build GRCh38
rs763246656 Chr.19: 12922853-12922853 on Build GRCh38
rs755048855 Chr.19: 12922648-12922648 on Build GRCh38
rs896189724 Chr.19: 12922686-12922687 on Build GRCh38
rs755136781 Chr.19: 12922768-12922768 on Build GRCh38
rs777702304 Chr.19: 12922783-12922783 on Build GRCh38
rs565376622 Chr.19: 12922721-12922721 on Build GRCh38
rs765689131 Chr.19: 12922736-12922736 on Build GRCh38
rs914755287 Chr.19: 12922840-12922840 on Build GRCh38
rs564274774 Chr.19: 12922439-12922439 on Build GRCh38
rs1042257219 Chr.19: 12922854-12922854 on Build GRCh38
rs958980425 Chr.19: 12922835-12922835 on Build GRCh38
rs748054999 Chr.19: 12922678-12922678 on Build GRCh38
rs747087928 Chr.19: 12922644-12922644 on Build GRCh38
rs771618775 Chr.19: 12922802-12922802 on Build GRCh38
rs528447648 Chr.19: 12922590-12922590 on Build GRCh38
rs991739856 Chr.19: 12922839-12922839 on Build GRCh38
rs926514041 Chr.19: 12922586-12922586 on Build GRCh38
rs766521776 Chr.19: 12922756-12922756 on Build GRCh38
rs903371089 Chr.19: 12922438-12922438 on Build GRCh38
rs765973832 Chr.19: 12922549-12922549 on Build GRCh38
rs1020402676 Chr.19: 12922427-12922427 on Build GRCh38
rs771902448 Chr.19: 12922568-12922568 on Build GRCh38
rs751778234 Chr.19: 12922762-12922762 on Build GRCh38
rs751029243 Chr.19: 12922572-12922572 on Build GRCh38
rs770293959 Chr.19: 12922514-12922514 on Build GRCh38
rs201776310 Chr.19: 12922778-12922778 on Build GRCh38
rs1018985479 Chr.19: 12922487-12922487 on Build GRCh38
rs770712368 Chr.19: 12922794-12922794 on Build GRCh38
rs774968212 Chr.19: 12922844-12922844 on Build GRCh38
rs78937739 Chr.19: 12922700-12922700 on Build GRCh38
rs1051333062 Chr.19: 12922554-12922554 on Build GRCh38
rs1033235566 Chr.19: 12922452-12922452 on Build GRCh38

View Full Product Details