Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13100520-13101010 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LYL1
Entrez Gene ID
4066
Gene Name
LYL1, basic helix-loop-helix family member
Gene Aliases
bHLHa18
UniGene
Hs.46446
Gene Chromosome Location
Chr.19: 13099028-13103160 on Build GRCh38
Transcripts
NM_005583.4 XM_005259912.3 XM_017026816.1 XM_017026817.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3756356 c.165G>A p.R55R Chr.19: 13101007-13101007 on Build GRCh38
5493846 c.162delC p.R55fs*8 Chr.19: 13101010-13101010 on Build GRCh38
3528979 c.356G>A p.G119E Chr.19: 13100728-13100728 on Build GRCh38
5416997 c.186G>C p.V62V Chr.19: 13100986-13100986 on Build GRCh38
4131232 c.409G>A p.E137K Chr.19: 13100675-13100675 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374421432 Chr.19: 13100788-13100788 on Build GRCh38
rs935693218 Chr.19: 13100961-13100961 on Build GRCh38
rs758293848 Chr.19: 13100644-13100644 on Build GRCh38
rs868638075 Chr.19: 13100976-13100976 on Build GRCh38
rs991271474 Chr.19: 13100564-13100564 on Build GRCh38
rs762291322 Chr.19: 13100679-13100679 on Build GRCh38
rs764985367 Chr.19: 13100629-13100629 on Build GRCh38
rs905574983 Chr.19: 13100953-13100954 on Build GRCh38
rs547998490 Chr.19: 13100813-13100813 on Build GRCh38
rs755158367 Chr.19: 13100827-13100827 on Build GRCh38
rs765389282 Chr.19: 13100956-13100956 on Build GRCh38
rs768374257 Chr.19: 13100888-13100888 on Build GRCh38
rs752397641 Chr.19: 13100632-13100632 on Build GRCh38
rs142687463 Chr.19: 13100694-13100694 on Build GRCh38
rs761632281 Chr.19: 13100908-13100908 on Build GRCh38
rs977704590 Chr.19: 13100944-13100944 on Build GRCh38
rs778281539 Chr.19: 13100590-13100590 on Build GRCh38
rs759121048 Chr.19: 13100692-13100692 on Build GRCh38
rs150306817 Chr.19: 13100662-13100662 on Build GRCh38
rs367615562 Chr.19: 13100882-13100882 on Build GRCh38
rs183066929 Chr.19: 13100543-13100543 on Build GRCh38
rs995294708 Chr.19: 13100817-13100817 on Build GRCh38
rs778078690 Chr.19: 13100857-13100857 on Build GRCh38
rs370424830 Chr.19: 13100761-13100761 on Build GRCh38
rs759637924 Chr.19: 13100946-13100946 on Build GRCh38
rs748467042 Chr.19: 13100835-13100835 on Build GRCh38
rs767520242 Chr.19: 13100711-13100711 on Build GRCh38
rs950007821 Chr.19: 13100619-13100619 on Build GRCh38
rs137873305 Chr.19: 13100667-13100667 on Build GRCh38
rs748848233 Chr.19: 13100887-13100887 on Build GRCh38
rs1017004407 Chr.19: 13100891-13100891 on Build GRCh38
rs112549109 Chr.19: 13100707-13100707 on Build GRCh38
rs767299703 Chr.19: 13100807-13100807 on Build GRCh38
rs552041622 Chr.19: 13100964-13100964 on Build GRCh38
rs760636016 Chr.19: 13100921-13100921 on Build GRCh38
rs559161932 Chr.19: 13100797-13100797 on Build GRCh38
rs769390448 Chr.19: 13100958-13100958 on Build GRCh38
rs781409416 Chr.19: 13100651-13100651 on Build GRCh38
rs755248484 Chr.19: 13100747-13100747 on Build GRCh38
rs541680498 Chr.19: 13100775-13100775 on Build GRCh38
rs775977439 Chr.19: 13100686-13100686 on Build GRCh38
rs1055515202 Chr.19: 13100683-13100683 on Build GRCh38
rs769354072 Chr.19: 13100777-13100777 on Build GRCh38
rs1018331115 Chr.19: 13100556-13100556 on Build GRCh38
rs199858890 Chr.19: 13100922-13100922 on Build GRCh38
rs971346575 Chr.19: 13100939-13100939 on Build GRCh38
rs758835291 Chr.19: 13100583-13100583 on Build GRCh38
rs763978229 Chr.19: 13100702-13100702 on Build GRCh38
rs779141564 Chr.19: 13100829-13100829 on Build GRCh38
rs747548528 Chr.19: 13100871-13100871 on Build GRCh38
rs758617479 Chr.19: 13100978-13100978 on Build GRCh38
rs142613662 Chr.19: 13100756-13100756 on Build GRCh38
rs928261945 Chr.19: 13100649-13100649 on Build GRCh38
rs774160036 Chr.19: 13100896-13100896 on Build GRCh38
rs373904382 Chr.19: 13100919-13100919 on Build GRCh38
rs748548623 Chr.19: 13100760-13100760 on Build GRCh38
rs542777258 Chr.19: 13100582-13100582 on Build GRCh38
rs370306488 Chr.19: 13100790-13100790 on Build GRCh38
rs771439967 Chr.19: 13100770-13100771 on Build GRCh38
rs766465464 Chr.19: 13100816-13100816 on Build GRCh38
rs144851024 Chr.19: 13100701-13100701 on Build GRCh38
rs1025930295 Chr.19: 13100851-13100851 on Build GRCh38
rs983319135 Chr.19: 13100658-13100658 on Build GRCh38
rs771400791 Chr.19: 13100880-13100880 on Build GRCh38
rs761722110 Chr.19: 13100801-13100801 on Build GRCh38
rs778187698 Chr.19: 13100990-13100990 on Build GRCh38
rs753870330 Chr.19: 13100929-13100929 on Build GRCh38
rs772611340 Chr.19: 13100685-13100685 on Build GRCh38
rs554915108 Chr.19: 13100678-13100678 on Build GRCh38
rs747521091 Chr.19: 13100646-13100647 on Build GRCh38
rs756230314 Chr.19: 13100728-13100728 on Build GRCh38
rs867035781 Chr.19: 13100867-13100867 on Build GRCh38
rs749503930 Chr.19: 13100743-13100743 on Build GRCh38
rs757239284 Chr.19: 13100708-13100708 on Build GRCh38
rs750420950 Chr.19: 13100717-13100717 on Build GRCh38
rs758806848 Chr.19: 13100836-13100836 on Build GRCh38
rs1025645189 Chr.19: 13100804-13100804 on Build GRCh38
rs769011957 Chr.19: 13100671-13100671 on Build GRCh38
rs1005607110 Chr.19: 13100832-13100832 on Build GRCh38
rs780073041 Chr.19: 13100732-13100732 on Build GRCh38
rs186712016 Chr.19: 13100798-13100798 on Build GRCh38
rs951262279 Chr.19: 13100878-13100878 on Build GRCh38
rs929909002 Chr.19: 13100625-13100625 on Build GRCh38
rs746009179 Chr.19: 13100654-13100654 on Build GRCh38
rs374235806 Chr.19: 13100818-13100818 on Build GRCh38
rs201509972 Chr.19: 13100740-13100740 on Build GRCh38
rs751548940 Chr.19: 13100648-13100648 on Build GRCh38
rs200927941 Chr.19: 13100914-13100914 on Build GRCh38

View Full Product Details