Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15807483-15807991 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR10H1
Entrez Gene ID
26539
Gene Name
olfactory receptor family 10 subfamily H member 1
Gene Aliases
-
UniGene
Hs.533925
Gene Chromosome Location
Chr.19: 15806957-15808759 on Build GRCh38
Transcripts
NM_013940.2 XM_011527911.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6035739 c.349G>A p.V117I Chr.19: 15807689-15807689 on Build GRCh38
3529895 c.550C>T p.L184L Chr.19: 15807488-15807488 on Build GRCh38
992350 c.239G>A p.R80H Chr.19: 15807799-15807799 on Build GRCh38
992348 c.370G>A p.V124M Chr.19: 15807668-15807668 on Build GRCh38
4382372 c.208G>A p.E70K Chr.19: 15807830-15807830 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778560322 Chr.19: 15807832-15807832 on Build GRCh38
rs146037488 Chr.19: 15807622-15807622 on Build GRCh38
rs763778171 Chr.19: 15807606-15807606 on Build GRCh38
rs908901500 Chr.19: 15807784-15807784 on Build GRCh38
rs61739573 Chr.19: 15807645-15807645 on Build GRCh38
rs372619420 Chr.19: 15807555-15807555 on Build GRCh38
rs775804063 Chr.19: 15807558-15807558 on Build GRCh38
rs1859299 Chr.19: 15807624-15807624 on Build GRCh38
rs375042545 Chr.19: 15807522-15807522 on Build GRCh38
rs1859298 Chr.19: 15807513-15807513 on Build GRCh38
rs533663011 Chr.19: 15807742-15807742 on Build GRCh38
rs757551867 Chr.19: 15807611-15807611 on Build GRCh38
rs767857628 Chr.19: 15807678-15807680 on Build GRCh38
rs764184293 Chr.19: 15807846-15807846 on Build GRCh38
rs774838264 Chr.19: 15807650-15807650 on Build GRCh38
rs199503067 Chr.19: 15807881-15807881 on Build GRCh38
rs4808382 Chr.19: 15807844-15807844 on Build GRCh38
rs774047283 Chr.19: 15807762-15807762 on Build GRCh38
rs559894657 Chr.19: 15807607-15807607 on Build GRCh38
rs747476863 Chr.19: 15807735-15807735 on Build GRCh38
rs774345060 Chr.19: 15807907-15807907 on Build GRCh38
rs867506124 Chr.19: 15807786-15807786 on Build GRCh38
rs756400126 Chr.19: 15807651-15807651 on Build GRCh38
rs375356636 Chr.19: 15807589-15807589 on Build GRCh38
rs768241536 Chr.19: 15807761-15807761 on Build GRCh38
rs148327766 Chr.19: 15807690-15807690 on Build GRCh38
rs758169205 Chr.19: 15807959-15807959 on Build GRCh38
rs758986001 Chr.19: 15807588-15807588 on Build GRCh38
rs61746479 Chr.19: 15807799-15807799 on Build GRCh38
rs752362373 Chr.19: 15807948-15807948 on Build GRCh38
rs908067741 Chr.19: 15807758-15807758 on Build GRCh38
rs200516553 Chr.19: 15807760-15807760 on Build GRCh38
rs754019278 Chr.19: 15807717-15807717 on Build GRCh38
rs749824144 Chr.19: 15807559-15807559 on Build GRCh38
rs368911893 Chr.19: 15807528-15807528 on Build GRCh38
rs983631350 Chr.19: 15807726-15807726 on Build GRCh38
rs755033326 Chr.19: 15807718-15807718 on Build GRCh38
rs143305519 Chr.19: 15807682-15807682 on Build GRCh38
rs866190870 Chr.19: 15807951-15807951 on Build GRCh38
rs779333750 Chr.19: 15807669-15807669 on Build GRCh38
rs762081414 Chr.19: 15807676-15807677 on Build GRCh38
rs139388455 Chr.19: 15807656-15807656 on Build GRCh38
rs762370423 Chr.19: 15807908-15807908 on Build GRCh38
rs201403532 Chr.19: 15807723-15807723 on Build GRCh38
rs776869228 Chr.19: 15807773-15807773 on Build GRCh38
rs766555045 Chr.19: 15807708-15807708 on Build GRCh38
rs747171458 Chr.19: 15807540-15807540 on Build GRCh38
rs772200912 Chr.19: 15807766-15807766 on Build GRCh38
rs143049130 Chr.19: 15807668-15807668 on Build GRCh38
rs766108663 Chr.19: 15807772-15807772 on Build GRCh38
rs745501445 Chr.19: 15807676-15807676 on Build GRCh38
rs764766302 Chr.19: 15807901-15807903 on Build GRCh38
rs142314142 Chr.19: 15807894-15807894 on Build GRCh38
rs768980814 Chr.19: 15807570-15807570 on Build GRCh38
rs573474241 Chr.19: 15807619-15807619 on Build GRCh38
rs61746482 Chr.19: 15807673-15807673 on Build GRCh38
rs148303831 Chr.19: 15807636-15807636 on Build GRCh38
rs752179805 Chr.19: 15807955-15807963 on Build GRCh38
rs8103631 Chr.19: 15807623-15807623 on Build GRCh38
rs760803311 Chr.19: 15807705-15807705 on Build GRCh38
rs879000346 Chr.19: 15807901-15807901 on Build GRCh38
rs753169062 Chr.19: 15807730-15807730 on Build GRCh38
rs893357990 Chr.19: 15807677-15807677 on Build GRCh38
rs370227017 Chr.19: 15807867-15807867 on Build GRCh38
rs61746478 Chr.19: 15807884-15807884 on Build GRCh38
rs752532622 Chr.19: 15807595-15807595 on Build GRCh38
rs140117949 Chr.19: 15807674-15807674 on Build GRCh38
rs376750171 Chr.19: 15807733-15807733 on Build GRCh38
rs369544216 Chr.19: 15807802-15807802 on Build GRCh38
rs552935859 Chr.19: 15807537-15807537 on Build GRCh38
rs781441788 Chr.19: 15807817-15807817 on Build GRCh38
rs748942406 Chr.19: 15807820-15807820 on Build GRCh38
rs369960921 Chr.19: 15807621-15807621 on Build GRCh38
rs199888665 Chr.19: 15807769-15807769 on Build GRCh38
rs751094428 Chr.19: 15807966-15807966 on Build GRCh38
rs146111076 Chr.19: 15807924-15807924 on Build GRCh38
rs750301120 Chr.19: 15807697-15807697 on Build GRCh38
rs199627907 Chr.19: 15807770-15807770 on Build GRCh38
rs982554037 Chr.19: 15807961-15807961 on Build GRCh38
rs781409715 Chr.19: 15807543-15807543 on Build GRCh38
rs28426969 Chr.19: 15807570-15807570 on Build GRCh38
rs4081570 Chr.19: 15807837-15807837 on Build GRCh38
rs545411328 Chr.19: 15807857-15807857 on Build GRCh38
rs896359793 Chr.19: 15807650-15807650 on Build GRCh38
rs145210707 Chr.19: 15807614-15807614 on Build GRCh38
rs774827645 Chr.19: 15807568-15807568 on Build GRCh38
rs771720675 Chr.19: 15807836-15807836 on Build GRCh38
rs61745313 Chr.19: 15807539-15807539 on Build GRCh38
rs768863790 Chr.19: 15807564-15807564 on Build GRCh38
rs766085127 Chr.19: 15807572-15807572 on Build GRCh38
rs750952119 Chr.19: 15807919-15807919 on Build GRCh38
rs758403968 Chr.19: 15807596-15807596 on Build GRCh38
rs757744623 Chr.19: 15807855-15807855 on Build GRCh38
rs752066788 Chr.19: 15807851-15807851 on Build GRCh38
rs752690437 Chr.19: 15807523-15807523 on Build GRCh38
rs747593084 Chr.19: 15807833-15807833 on Build GRCh38
rs780214769 Chr.19: 15807620-15807620 on Build GRCh38
rs759214532 Chr.19: 15807627-15807627 on Build GRCh38
rs759715060 Chr.19: 15807774-15807774 on Build GRCh38
rs754728990 Chr.19: 15807831-15807831 on Build GRCh38
rs757593901 Chr.19: 15807815-15807815 on Build GRCh38
rs746108480 Chr.19: 15807615-15807615 on Build GRCh38
rs769593874 Chr.19: 15807885-15807885 on Build GRCh38
rs527762025 Chr.19: 15807646-15807646 on Build GRCh38
rs141400861 Chr.19: 15807638-15807638 on Build GRCh38
rs957531359 Chr.19: 15807566-15807566 on Build GRCh38
rs996274710 Chr.19: 15807635-15807635 on Build GRCh38
rs1014372039 Chr.19: 15807512-15807512 on Build GRCh38
rs1017859236 Chr.19: 15807941-15807941 on Build GRCh38
rs780935913 Chr.19: 15807968-15807968 on Build GRCh38
rs758904258 Chr.19: 15807800-15807800 on Build GRCh38
rs776337622 Chr.19: 15807839-15807839 on Build GRCh38
rs370618160 Chr.19: 15807967-15807967 on Build GRCh38
rs746014232 Chr.19: 15807552-15807552 on Build GRCh38
rs779702739 Chr.19: 15807759-15807759 on Build GRCh38
rs750812015 Chr.19: 15807859-15807859 on Build GRCh38
rs965763384 Chr.19: 15807592-15807592 on Build GRCh38
rs777321103 Chr.19: 15807965-15807965 on Build GRCh38
rs752023657 Chr.19: 15807814-15807814 on Build GRCh38
rs753402574 Chr.19: 15807510-15807510 on Build GRCh38
rs201182090 Chr.19: 15807868-15807868 on Build GRCh38
rs765276511 Chr.19: 15807794-15807794 on Build GRCh38
rs371354392 Chr.19: 15807734-15807734 on Build GRCh38
rs746844709 Chr.19: 15807728-15807730 on Build GRCh38
rs762460753 Chr.19: 15807689-15807689 on Build GRCh38
rs766879923 Chr.19: 15807947-15807947 on Build GRCh38
rs750634455 Chr.19: 15807647-15807647 on Build GRCh38
rs754903893 Chr.19: 15807515-15807515 on Build GRCh38
rs551484315 Chr.19: 15807640-15807640 on Build GRCh38
rs761130178 Chr.19: 15807762-15807772 on Build GRCh38
rs767390628 Chr.19: 15807692-15807692 on Build GRCh38
rs767843902 Chr.19: 15807909-15807909 on Build GRCh38
rs4081569 Chr.19: 15807580-15807580 on Build GRCh38
rs1008193166 Chr.19: 15807937-15807937 on Build GRCh38
rs373703831 Chr.19: 15807880-15807880 on Build GRCh38
rs1033697254 Chr.19: 15807556-15807556 on Build GRCh38
rs143282709 Chr.19: 15807625-15807625 on Build GRCh38
rs187451101 Chr.19: 15807639-15807639 on Build GRCh38
rs757940466 Chr.19: 15807870-15807870 on Build GRCh38

View Full Product Details