Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16831405-16831910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIN3B
Entrez Gene ID
23309
Gene Name
SIN3 transcription regulator family member B
Gene Aliases
-
UniGene
Hs.13999
Gene Chromosome Location
Chr.19: 16829387-16880355 on Build GRCh38
Transcripts
NM_001297595.1 NM_015260.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3932919 c.252A>G p.R84R Chr.19: 16831518-16831518 on Build GRCh38
1225761 c.364T>C p.S122P Chr.19: 16831630-16831630 on Build GRCh38
4075221 c.303C>T p.N101N Chr.19: 16831569-16831569 on Build GRCh38
3530181 c.315C>T p.P105P Chr.19: 16831581-16831581 on Build GRCh38
1480736 c.331G>A p.D111N Chr.19: 16831597-16831597 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770467693 Chr.19: 16831464-16831464 on Build GRCh38
rs978637440 Chr.19: 16831857-16831857 on Build GRCh38
rs920739323 Chr.19: 16831516-16831516 on Build GRCh38
rs752635517 Chr.19: 16831527-16831527 on Build GRCh38
rs373882538 Chr.19: 16831623-16831623 on Build GRCh38
rs976924792 Chr.19: 16831843-16831843 on Build GRCh38
rs182103445 Chr.19: 16831823-16831823 on Build GRCh38
rs1024652465 Chr.19: 16831822-16831822 on Build GRCh38
rs567366909 Chr.19: 16831859-16831859 on Build GRCh38
rs774058845 Chr.19: 16831684-16831684 on Build GRCh38
rs774966159 Chr.19: 16831668-16831668 on Build GRCh38
rs558546247 Chr.19: 16831479-16831479 on Build GRCh38
rs369627696 Chr.19: 16831690-16831690 on Build GRCh38
rs1011964183 Chr.19: 16831803-16831803 on Build GRCh38
rs774721766 Chr.19: 16831490-16831490 on Build GRCh38
rs547508167 Chr.19: 16831847-16831847 on Build GRCh38
rs756348729 Chr.19: 16831696-16831696 on Build GRCh38
rs761688432 Chr.19: 16831687-16831687 on Build GRCh38
rs760305814 Chr.19: 16831495-16831495 on Build GRCh38
rs746254861 Chr.19: 16831658-16831659 on Build GRCh38
rs1019558515 Chr.19: 16831827-16831827 on Build GRCh38
rs1014255017 Chr.19: 16831480-16831480 on Build GRCh38
rs767045323 Chr.19: 16831722-16831722 on Build GRCh38
rs757486941 Chr.19: 16831456-16831456 on Build GRCh38
rs779213696 Chr.19: 16831584-16831584 on Build GRCh38
rs531647315 Chr.19: 16831780-16831780 on Build GRCh38
rs151301610 Chr.19: 16831845-16831845 on Build GRCh38
rs768911009 Chr.19: 16831485-16831485 on Build GRCh38
rs74740618 Chr.19: 16831831-16831831 on Build GRCh38
rs376267076 Chr.19: 16831669-16831669 on Build GRCh38
rs772899091 Chr.19: 16831451-16831451 on Build GRCh38
rs371285274 Chr.19: 16831541-16831541 on Build GRCh38
rs751363811 Chr.19: 16831538-16831538 on Build GRCh38
rs1039016113 Chr.19: 16831540-16831540 on Build GRCh38
rs756297307 Chr.19: 16831555-16831555 on Build GRCh38
rs192092006 Chr.19: 16831759-16831759 on Build GRCh38
rs368592710 Chr.19: 16831628-16831628 on Build GRCh38
rs762890395 Chr.19: 16831533-16831533 on Build GRCh38
rs920953077 Chr.19: 16831569-16831569 on Build GRCh38
rs199552816 Chr.19: 16831667-16831667 on Build GRCh38
rs375926238 Chr.19: 16831688-16831688 on Build GRCh38
rs764057559 Chr.19: 16831675-16831675 on Build GRCh38
rs781203875 Chr.19: 16831478-16831478 on Build GRCh38
rs764678519 Chr.19: 16831526-16831526 on Build GRCh38
rs372131912 Chr.19: 16831660-16831660 on Build GRCh38
rs377068857 Chr.19: 16831880-16831880 on Build GRCh38
rs933246586 Chr.19: 16831616-16831616 on Build GRCh38
rs1053403200 Chr.19: 16831686-16831686 on Build GRCh38
rs541122549 Chr.19: 16831521-16831521 on Build GRCh38
rs553072824 Chr.19: 16831497-16831497 on Build GRCh38
rs770776436 Chr.19: 16831709-16831709 on Build GRCh38
rs145884472 Chr.19: 16831539-16831539 on Build GRCh38
rs748515617 Chr.19: 16831606-16831606 on Build GRCh38
rs750620312 Chr.19: 16831554-16831554 on Build GRCh38
rs1027863864 Chr.19: 16831716-16831716 on Build GRCh38
rs769196384 Chr.19: 16831570-16831570 on Build GRCh38
rs765488110 Chr.19: 16831882-16831882 on Build GRCh38
rs776160582 Chr.19: 16831520-16831520 on Build GRCh38
rs753951125 Chr.19: 16831697-16831697 on Build GRCh38
rs932316205 Chr.19: 16831854-16831854 on Build GRCh38
rs61739673 Chr.19: 16831631-16831631 on Build GRCh38
rs763851909 Chr.19: 16831536-16831536 on Build GRCh38
rs375440897 Chr.19: 16831459-16831459 on Build GRCh38
rs200751623 Chr.19: 16831557-16831557 on Build GRCh38
rs879140038 Chr.19: 16831777-16831777 on Build GRCh38
rs569703927 Chr.19: 16831811-16831811 on Build GRCh38
rs780724977 Chr.19: 16831470-16831470 on Build GRCh38
rs970051358 Chr.19: 16831703-16831703 on Build GRCh38

View Full Product Details