Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 42248654-42249160 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERF
Entrez Gene ID
2077
Gene Name
ETS2 repressor factor
Gene Aliases
CRS4, PE-2, PE2
UniGene
Hs.655969
Gene Chromosome Location
Chr.19: 42247561-42255164 on Build GRCh38
Transcripts
NM_001301035.1 NM_001308402.1 NM_001312656.1 NM_006494.3 XM_017026468.1 XM_017026469.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3534665 c.1103C>T p.S368F Chr.19: 42249009-42249009 on Build GRCh38
5423530 c.1201_1202delAA p.K401fs*10 Chr.19: 42248910-42248911 on Build GRCh38
1394071 c.1259C>T p.P420L Chr.19: 42248853-42248853 on Build GRCh38
6656049 c.1192G>A p.G398S Chr.19: 42248920-42248920 on Build GRCh38
3534663 c.1275C>T p.I425I Chr.19: 42248837-42248837 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs923175877 Chr.19: 42248999-42248999 on Build GRCh38
rs567417863 Chr.19: 42248926-42248926 on Build GRCh38
rs375042833 Chr.19: 42248696-42248696 on Build GRCh38
rs747844852 Chr.19: 42248943-42248943 on Build GRCh38
rs746369293 Chr.19: 42248927-42248927 on Build GRCh38
rs376218770 Chr.19: 42248954-42248954 on Build GRCh38
rs778754760 Chr.19: 42248760-42248760 on Build GRCh38
rs752492976 Chr.19: 42249011-42249011 on Build GRCh38
rs776121744 Chr.19: 42248837-42248837 on Build GRCh38
rs61735151 Chr.19: 42249131-42249131 on Build GRCh38
rs772094252 Chr.19: 42249062-42249062 on Build GRCh38
rs747311260 Chr.19: 42249037-42249037 on Build GRCh38
rs994349738 Chr.19: 42248875-42248875 on Build GRCh38
rs780415692 Chr.19: 42249036-42249036 on Build GRCh38
rs534018115 Chr.19: 42248901-42248901 on Build GRCh38
rs111793182 Chr.19: 42248920-42248920 on Build GRCh38
rs377467874 Chr.19: 42249081-42249081 on Build GRCh38
rs774556485 Chr.19: 42248820-42248820 on Build GRCh38
rs199836815 Chr.19: 42249032-42249032 on Build GRCh38
rs748367925 Chr.19: 42248858-42248858 on Build GRCh38
rs577264310 Chr.19: 42249017-42249017 on Build GRCh38
rs761007034 Chr.19: 42249014-42249014 on Build GRCh38
rs781590022 Chr.19: 42249114-42249114 on Build GRCh38
rs772786203 Chr.19: 42248960-42248960 on Build GRCh38
rs372067864 Chr.19: 42249023-42249023 on Build GRCh38
rs370719643 Chr.19: 42249064-42249064 on Build GRCh38
rs746151991 Chr.19: 42248873-42248873 on Build GRCh38
rs758877628 Chr.19: 42249035-42249035 on Build GRCh38
rs754070619 Chr.19: 42249074-42249074 on Build GRCh38
rs757176564 Chr.19: 42248997-42249002 on Build GRCh38
rs373251870 Chr.19: 42248889-42248889 on Build GRCh38
rs868252252 Chr.19: 42248692-42248692 on Build GRCh38
rs139842507 Chr.19: 42248868-42248868 on Build GRCh38
rs745620257 Chr.19: 42248698-42248698 on Build GRCh38
rs774221915 Chr.19: 42248969-42248969 on Build GRCh38
rs767470579 Chr.19: 42249000-42249000 on Build GRCh38
rs768011203 Chr.19: 42249044-42249044 on Build GRCh38
rs754788068 Chr.19: 42248906-42248906 on Build GRCh38
rs1063897 Chr.19: 42248970-42248970 on Build GRCh38
rs748943380 Chr.19: 42249122-42249122 on Build GRCh38
rs923763445 Chr.19: 42248946-42248946 on Build GRCh38
rs760396867 Chr.19: 42248825-42248825 on Build GRCh38
rs62119632 Chr.19: 42248704-42248704 on Build GRCh38
rs770438845 Chr.19: 42248800-42248800 on Build GRCh38
rs771292199 Chr.19: 42248866-42248866 on Build GRCh38
rs35493131 Chr.19: 42248882-42248882 on Build GRCh38
rs781509895 Chr.19: 42248856-42248856 on Build GRCh38
rs766642440 Chr.19: 42248740-42248740 on Build GRCh38
rs750423382 Chr.19: 42248849-42248849 on Build GRCh38
rs745600955 Chr.19: 42249009-42249011 on Build GRCh38
rs868009009 Chr.19: 42248953-42248953 on Build GRCh38
rs749286761 Chr.19: 42248921-42248921 on Build GRCh38
rs1049744926 Chr.19: 42249006-42249006 on Build GRCh38
rs367946660 Chr.19: 42248717-42248717 on Build GRCh38
rs761252512 Chr.19: 42248843-42248843 on Build GRCh38
rs915295153 Chr.19: 42248874-42248874 on Build GRCh38
rs372854370 Chr.19: 42249049-42249049 on Build GRCh38
rs186795992 Chr.19: 42249059-42249059 on Build GRCh38
rs979226353 Chr.19: 42248709-42248709 on Build GRCh38
rs772282991 Chr.19: 42248705-42248705 on Build GRCh38
rs757906071 Chr.19: 42248923-42248934 on Build GRCh38
rs897260918 Chr.19: 42249046-42249046 on Build GRCh38
rs758914076 Chr.19: 42249091-42249091 on Build GRCh38
rs750229302 Chr.19: 42248761-42248761 on Build GRCh38
rs768844827 Chr.19: 42248708-42248708 on Build GRCh38
rs753010203 Chr.19: 42248911-42248911 on Build GRCh38
rs376130330 Chr.19: 42249060-42249060 on Build GRCh38
rs755405849 Chr.19: 42249104-42249104 on Build GRCh38
rs529385497 Chr.19: 42248690-42248690 on Build GRCh38
rs747308083 Chr.19: 42248774-42248774 on Build GRCh38
rs752343403 Chr.19: 42248917-42248925 on Build GRCh38
rs533363033 Chr.19: 42248850-42248850 on Build GRCh38
rs562876278 Chr.19: 42248765-42248765 on Build GRCh38
rs76105803 Chr.19: 42249008-42249008 on Build GRCh38
rs141603714 Chr.19: 42248827-42248827 on Build GRCh38
rs773915281 Chr.19: 42248808-42248808 on Build GRCh38
rs763354193 Chr.19: 42248739-42248739 on Build GRCh38
rs372122049 Chr.19: 42248726-42248726 on Build GRCh38
rs757787819 Chr.19: 42248924-42248924 on Build GRCh38
rs901782723 Chr.19: 42248798-42248798 on Build GRCh38
rs200540167 Chr.19: 42248966-42248966 on Build GRCh38
rs759570663 Chr.19: 42249061-42249061 on Build GRCh38
rs764805806 Chr.19: 42248888-42248889 on Build GRCh38
rs960372517 Chr.19: 42248700-42248700 on Build GRCh38
rs749808726 Chr.19: 42248812-42248812 on Build GRCh38
rs373026073 Chr.19: 42248932-42248932 on Build GRCh38
rs538236832 Chr.19: 42249024-42249024 on Build GRCh38
rs763637628 Chr.19: 42248995-42248998 on Build GRCh38
rs990909606 Chr.19: 42248706-42248706 on Build GRCh38
rs752052933 Chr.19: 42249103-42249103 on Build GRCh38
rs773719188 Chr.19: 42248735-42248735 on Build GRCh38
rs755237509 Chr.19: 42248855-42248855 on Build GRCh38
rs376718198 Chr.19: 42248891-42248891 on Build GRCh38
rs138891601 Chr.19: 42248855-42248856 on Build GRCh38
rs756329901 Chr.19: 42248859-42248859 on Build GRCh38
rs777970610 Chr.19: 42248862-42248862 on Build GRCh38
rs756887571 Chr.19: 42249124-42249124 on Build GRCh38
rs913822192 Chr.19: 42249029-42249029 on Build GRCh38
rs760213978 Chr.19: 42248744-42248744 on Build GRCh38
rs758268534 Chr.19: 42248762-42248762 on Build GRCh38
rs768716522 Chr.19: 42248778-42248778 on Build GRCh38
rs781454700 Chr.19: 42248780-42248780 on Build GRCh38
rs762060326 Chr.19: 42249075-42249075 on Build GRCh38
rs150750115 Chr.19: 42248846-42248846 on Build GRCh38
rs751274436 Chr.19: 42248852-42248852 on Build GRCh38
rs144812092 Chr.19: 42249066-42249066 on Build GRCh38
rs780483224 Chr.19: 42249092-42249092 on Build GRCh38
rs762316422 Chr.19: 42248963-42248963 on Build GRCh38
rs778319851 Chr.19: 42249125-42249125 on Build GRCh38
rs745308883 Chr.19: 42249128-42249128 on Build GRCh38
rs1050195236 Chr.19: 42248819-42248819 on Build GRCh38
rs1009543151 Chr.19: 42248996-42248997 on Build GRCh38
rs757215993 Chr.19: 42248755-42248755 on Build GRCh38
rs759487353 Chr.19: 42248988-42248988 on Build GRCh38
rs759202471 Chr.19: 42248867-42248869 on Build GRCh38
rs756407997 Chr.19: 42249082-42249087 on Build GRCh38
rs376964824 Chr.19: 42248745-42248745 on Build GRCh38
rs587777010 Chr.19: 42248842-42248842 on Build GRCh38
rs769453776 Chr.19: 42249047-42249047 on Build GRCh38
rs149405304 Chr.19: 42249055-42249055 on Build GRCh38
rs764764288 Chr.19: 42248890-42248890 on Build GRCh38
rs776319104 Chr.19: 42248888-42248888 on Build GRCh38
rs748175534 Chr.19: 42248790-42248790 on Build GRCh38
rs749850802 Chr.19: 42248863-42248863 on Build GRCh38
rs763967813 Chr.19: 42249071-42249071 on Build GRCh38
rs963794580 Chr.19: 42248978-42248983 on Build GRCh38
rs753965187 Chr.19: 42249018-42249018 on Build GRCh38
rs751388026 Chr.19: 42248905-42248905 on Build GRCh38
rs750465267 Chr.19: 42249086-42249086 on Build GRCh38
rs905310462 Chr.19: 42248899-42248899 on Build GRCh38
rs199960550 Chr.19: 42248997-42248999 on Build GRCh38
rs780261925 Chr.19: 42248707-42248707 on Build GRCh38
rs771368216 Chr.19: 42248816-42248816 on Build GRCh38
rs562281233 Chr.19: 42248680-42248680 on Build GRCh38
rs781154235 Chr.19: 42249008-42249009 on Build GRCh38

View Full Product Details