Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 42249687-42250179 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERF
Entrez Gene ID
2077
Gene Name
ETS2 repressor factor
Gene Aliases
CRS4, PE-2, PE2
UniGene
Hs.655969
Gene Chromosome Location
Chr.19: 42247561-42255164 on Build GRCh38
Transcripts
NM_001301035.1 NM_001308402.1 NM_001312656.1 NM_006494.3 XM_017026468.1 XM_017026469.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4283613 c.408G>A p.P136P Chr.19: 42249704-42249704 on Build GRCh38
5573661 c.258-2A>G p.? Chr.19: 42249944-42249944 on Build GRCh38
5855935 c.410C>T p.S137L Chr.19: 42249702-42249702 on Build GRCh38
5190537 c.301C>T p.R101W Chr.19: 42249899-42249899 on Build GRCh38
4934361 c.417T>G p.G139G Chr.19: 42249695-42249695 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs544806380 Chr.19: 42249990-42249990 on Build GRCh38
rs201132767 Chr.19: 42249713-42249713 on Build GRCh38
rs781452659 Chr.19: 42249729-42249729 on Build GRCh38
rs755202583 Chr.19: 42249725-42249725 on Build GRCh38
rs749799715 Chr.19: 42249749-42249749 on Build GRCh38
rs774456067 Chr.19: 42249980-42249981 on Build GRCh38
rs963272925 Chr.19: 42250079-42250079 on Build GRCh38
rs769239795 Chr.19: 42249899-42249899 on Build GRCh38
rs377095568 Chr.19: 42250151-42250151 on Build GRCh38
rs747830281 Chr.19: 42249889-42249889 on Build GRCh38
rs746165064 Chr.19: 42249846-42249846 on Build GRCh38
rs565631783 Chr.19: 42249927-42249927 on Build GRCh38
rs764099591 Chr.19: 42249965-42249965 on Build GRCh38
rs886041001 Chr.19: 42249934-42249934 on Build GRCh38
rs578157674 Chr.19: 42250050-42250050 on Build GRCh38
rs538318687 Chr.19: 42249778-42249778 on Build GRCh38
rs906306796 Chr.19: 42250114-42250114 on Build GRCh38
rs376308513 Chr.19: 42249768-42249768 on Build GRCh38
rs933816378 Chr.19: 42249718-42249718 on Build GRCh38
rs767793643 Chr.19: 42249796-42249796 on Build GRCh38
rs374830901 Chr.19: 42250058-42250058 on Build GRCh38
rs561227068 Chr.19: 42250120-42250120 on Build GRCh38
rs4039718 Chr.19: 42250125-42250125 on Build GRCh38
rs201871955 Chr.19: 42249980-42249980 on Build GRCh38
rs932312576 Chr.19: 42249712-42249712 on Build GRCh38
rs911968414 Chr.19: 42249998-42249998 on Build GRCh38
rs1003363153 Chr.19: 42250136-42250136 on Build GRCh38
rs556720236 Chr.19: 42250000-42250000 on Build GRCh38
rs756218487 Chr.19: 42249810-42249810 on Build GRCh38
rs372380443 Chr.19: 42249757-42249757 on Build GRCh38
rs951326967 Chr.19: 42250110-42250110 on Build GRCh38
rs772731129 Chr.19: 42249900-42249900 on Build GRCh38
rs1030457542 Chr.19: 42250010-42250010 on Build GRCh38
rs770925496 Chr.19: 42249920-42249920 on Build GRCh38
rs560293119 Chr.19: 42250088-42250088 on Build GRCh38
rs768310421 Chr.19: 42249852-42249852 on Build GRCh38
rs774305612 Chr.19: 42249926-42249926 on Build GRCh38
rs866871043 Chr.19: 42249958-42249958 on Build GRCh38
rs778205341 Chr.19: 42249735-42249735 on Build GRCh38
rs200256396 Chr.19: 42249764-42249764 on Build GRCh38
rs1054646737 Chr.19: 42250055-42250055 on Build GRCh38
rs1045254229 Chr.19: 42249915-42249915 on Build GRCh38
rs144678577 Chr.19: 42249707-42249707 on Build GRCh38
rs151275041 Chr.19: 42249732-42249732 on Build GRCh38
rs759843106 Chr.19: 42249789-42249789 on Build GRCh38
rs917969973 Chr.19: 42250142-42250142 on Build GRCh38
rs556023808 Chr.19: 42249939-42249939 on Build GRCh38
rs945037534 Chr.19: 42250085-42250085 on Build GRCh38
rs992174402 Chr.19: 42250130-42250130 on Build GRCh38
rs531529282 Chr.19: 42249731-42249731 on Build GRCh38
rs904084126 Chr.19: 42249917-42249917 on Build GRCh38
rs936233120 Chr.19: 42250054-42250054 on Build GRCh38
rs1050967534 Chr.19: 42249728-42249728 on Build GRCh38
rs140522983 Chr.19: 42249936-42249936 on Build GRCh38
rs548811826 Chr.19: 42249763-42249763 on Build GRCh38
rs746682592 Chr.19: 42249752-42249752 on Build GRCh38
rs776123538 Chr.19: 42249760-42249760 on Build GRCh38
rs866278705 Chr.19: 42249784-42249784 on Build GRCh38
rs762831348 Chr.19: 42249918-42249918 on Build GRCh38
rs1053408488 Chr.19: 42249723-42249723 on Build GRCh38
rs1052030393 Chr.19: 42249937-42249937 on Build GRCh38
rs1033586611 Chr.19: 42249843-42249843 on Build GRCh38
rs760926921 Chr.19: 42249963-42249963 on Build GRCh38
rs79269780 Chr.19: 42249910-42249910 on Build GRCh38
rs948179552 Chr.19: 42249792-42249792 on Build GRCh38
rs542601690 Chr.19: 42250116-42250116 on Build GRCh38
rs771237144 Chr.19: 42249751-42249751 on Build GRCh38
rs780879505 Chr.19: 42249873-42249873 on Build GRCh38
rs922168754 Chr.19: 42250026-42250026 on Build GRCh38
rs753911446 Chr.19: 42249976-42249976 on Build GRCh38
rs893491491 Chr.19: 42249756-42249756 on Build GRCh38
rs1025643124 Chr.19: 42250095-42250095 on Build GRCh38
rs553368620 Chr.19: 42249786-42249786 on Build GRCh38
rs778844159 Chr.19: 42249716-42249716 on Build GRCh38
rs368794183 Chr.19: 42249819-42249819 on Build GRCh38
rs750915227 Chr.19: 42249994-42249994 on Build GRCh38
rs527637425 Chr.19: 42250094-42250094 on Build GRCh38
rs754348493 Chr.19: 42249828-42249828 on Build GRCh38
rs757730703 Chr.19: 42249838-42249838 on Build GRCh38
rs983523826 Chr.19: 42249993-42249993 on Build GRCh38
rs752957233 Chr.19: 42249802-42249802 on Build GRCh38
rs888891232 Chr.19: 42250013-42250013 on Build GRCh38
rs779276597 Chr.19: 42249839-42249839 on Build GRCh38

View Full Product Details