Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44328387-44328887 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF112
Entrez Gene ID
7771
Gene Name
zinc finger protein 112
Gene Aliases
ZFP112, ZNF228
UniGene
Hs.48589
Gene Chromosome Location
Chr.19: 44326553-44356704 on Build GRCh38
Transcripts
NM_001083335.1 NM_013380.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6322300 c.1504C>A p.H502N Chr.19: 44328671-44328671 on Build GRCh38
4909004 c.1653C>T p.F551F Chr.19: 44328573-44328573 on Build GRCh38
7338129 c.1543C>T p.P515S Chr.19: 44328683-44328683 on Build GRCh38
6827903 c.1696G>T p.G566* Chr.19: 44328530-44328530 on Build GRCh38
997911 c.1566G>T p.Q522H Chr.19: 44328609-44328609 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746134196 Chr.19: 44328477-44328477 on Build GRCh38
rs765934644 Chr.19: 44328516-44328516 on Build GRCh38
rs368998978 Chr.19: 44328738-44328738 on Build GRCh38
rs759175276 Chr.19: 44328641-44328641 on Build GRCh38
rs79226975 Chr.19: 44328484-44328484 on Build GRCh38
rs772832133 Chr.19: 44328600-44328600 on Build GRCh38
rs61733037 Chr.19: 44328540-44328540 on Build GRCh38
rs568068134 Chr.19: 44328530-44328530 on Build GRCh38
rs61733036 Chr.19: 44328432-44328432 on Build GRCh38
rs757666902 Chr.19: 44328551-44328551 on Build GRCh38
rs779444932 Chr.19: 44328553-44328553 on Build GRCh38
rs776336796 Chr.19: 44328779-44328779 on Build GRCh38
rs775100653 Chr.19: 44328761-44328761 on Build GRCh38
rs774321408 Chr.19: 44328617-44328617 on Build GRCh38
rs79702957 Chr.19: 44328629-44328629 on Build GRCh38
rs763704408 Chr.19: 44328778-44328778 on Build GRCh38
rs868811027 Chr.19: 44328437-44328437 on Build GRCh38
rs938944674 Chr.19: 44328672-44328672 on Build GRCh38
rs1053120350 Chr.19: 44328838-44328838 on Build GRCh38
rs775669930 Chr.19: 44328485-44328485 on Build GRCh38
rs775546962 Chr.19: 44328860-44328860 on Build GRCh38
rs769640239 Chr.19: 44328588-44328588 on Build GRCh38
rs199557381 Chr.19: 44328412-44328412 on Build GRCh38
rs767131008 Chr.19: 44328648-44328648 on Build GRCh38
rs749692613 Chr.19: 44328464-44328464 on Build GRCh38
rs756612281 Chr.19: 44328537-44328537 on Build GRCh38
rs757006954 Chr.19: 44328834-44328834 on Build GRCh38
rs532186812 Chr.19: 44328471-44328471 on Build GRCh38
rs189870060 Chr.19: 44328823-44328823 on Build GRCh38
rs374389056 Chr.19: 44328801-44328801 on Build GRCh38
rs757081009 Chr.19: 44328561-44328561 on Build GRCh38
rs200140596 Chr.19: 44328627-44328627 on Build GRCh38
rs770744772 Chr.19: 44328754-44328754 on Build GRCh38
rs138793205 Chr.19: 44328419-44328419 on Build GRCh38
rs553591871 Chr.19: 44328602-44328602 on Build GRCh38
rs769577041 Chr.19: 44328495-44328495 on Build GRCh38
rs779348742 Chr.19: 44328679-44328679 on Build GRCh38
rs746562159 Chr.19: 44328861-44328861 on Build GRCh38
rs201811934 Chr.19: 44328533-44328533 on Build GRCh38
rs750316822 Chr.19: 44328415-44328415 on Build GRCh38
rs748396972 Chr.19: 44328832-44328832 on Build GRCh38
rs747164232 Chr.19: 44328585-44328585 on Build GRCh38
rs754197022 Chr.19: 44328670-44328670 on Build GRCh38
rs200968652 Chr.19: 44328547-44328547 on Build GRCh38
rs559269586 Chr.19: 44328486-44328486 on Build GRCh38
rs745999281 Chr.19: 44328570-44328570 on Build GRCh38
rs768718617 Chr.19: 44328745-44328745 on Build GRCh38
rs760202722 Chr.19: 44328766-44328766 on Build GRCh38
rs202199443 Chr.19: 44328758-44328758 on Build GRCh38
rs370801215 Chr.19: 44328753-44328753 on Build GRCh38
rs760356052 Chr.19: 44328664-44328664 on Build GRCh38
rs184596992 Chr.19: 44328466-44328466 on Build GRCh38
rs762155095 Chr.19: 44328787-44328787 on Build GRCh38
rs146859568 Chr.19: 44328847-44328847 on Build GRCh38
rs919505419 Chr.19: 44328638-44328638 on Build GRCh38
rs374446820 Chr.19: 44328676-44328676 on Build GRCh38
rs904590565 Chr.19: 44328785-44328785 on Build GRCh38
rs367874199 Chr.19: 44328584-44328584 on Build GRCh38
rs766474456 Chr.19: 44328805-44328805 on Build GRCh38
rs2722722 Chr.19: 44328722-44328722 on Build GRCh38
rs758849029 Chr.19: 44328802-44328802 on Build GRCh38
rs879186526 Chr.19: 44328435-44328435 on Build GRCh38
rs754624905 Chr.19: 44328446-44328446 on Build GRCh38
rs547904462 Chr.19: 44328510-44328510 on Build GRCh38
rs555758131 Chr.19: 44328683-44328683 on Build GRCh38
rs148277871 Chr.19: 44328849-44328849 on Build GRCh38
rs2722723 Chr.19: 44328839-44328839 on Build GRCh38
rs755061830 Chr.19: 44328815-44328815 on Build GRCh38
rs770438838 Chr.19: 44328601-44328601 on Build GRCh38
rs768209590 Chr.19: 44328862-44328862 on Build GRCh38
rs770812071 Chr.19: 44328605-44328605 on Build GRCh38
rs756522463 Chr.19: 44328459-44328459 on Build GRCh38
rs949914158 Chr.19: 44328469-44328469 on Build GRCh38
rs772129875 Chr.19: 44328573-44328573 on Build GRCh38
rs545633696 Chr.19: 44328775-44328775 on Build GRCh38
rs781713121 Chr.19: 44328447-44328447 on Build GRCh38
rs139777621 Chr.19: 44328509-44328509 on Build GRCh38
rs774497486 Chr.19: 44328757-44328757 on Build GRCh38
rs536128138 Chr.19: 44328655-44328655 on Build GRCh38
rs757990345 Chr.19: 44328416-44328416 on Build GRCh38
rs377135791 Chr.19: 44328748-44328748 on Build GRCh38
rs768981130 Chr.19: 44328735-44328782 on Build GRCh38
rs748563731 Chr.19: 44328454-44328454 on Build GRCh38
rs141323572 Chr.19: 44328790-44328790 on Build GRCh38
rs386352382 Chr.19: 44328788-44328788 on Build GRCh38
rs751676235 Chr.19: 44328814-44328814 on Build GRCh38
rs758481696 Chr.19: 44328701-44328701 on Build GRCh38
rs142095072 Chr.19: 44328665-44328665 on Build GRCh38
rs544240140 Chr.19: 44328730-44328730 on Build GRCh38
rs774516913 Chr.19: 44328753-44328754 on Build GRCh38
rs2609880 Chr.19: 44328531-44328531 on Build GRCh38
rs1000386427 Chr.19: 44328795-44328795 on Build GRCh38
rs904516894 Chr.19: 44328427-44328427 on Build GRCh38
rs147418906 Chr.19: 44328460-44328460 on Build GRCh38
rs1043069542 Chr.19: 44328423-44328423 on Build GRCh38
rs771730790 Chr.19: 44328760-44328760 on Build GRCh38

View Full Product Details