Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49878065-49878569 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKT1S1
Entrez Gene ID
84335
Gene Name
AKT1 substrate 1
Gene Aliases
Lobe, PRAS40
UniGene
Hs.515542
Gene Chromosome Location
Chr.19: 49869033-49878356 on Build GRCh38
Transcripts
NM_001278160.1 NM_001168222.1 NM_024682.2 XM_011527317.2

Gene Symbol
TBC1D17
Entrez Gene ID
79735
Gene Name
TBC1 domain family member 17
Gene Aliases
-
UniGene
Hs.631587
Gene Chromosome Location
Chr.19: 49877425-49888750 on Build GRCh38
Transcripts
NM_001278160.1 NM_001168222.1 NM_024682.2 XM_011527317.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7429876 c.81A>T p.R27R Chr.19: 49878202-49878202 on Build GRCh38
4171012 c.154G>C p.E52Q Chr.19: 49878531-49878531 on Build GRCh38
7111860 c.166G>T p.D56Y Chr.19: 49878543-49878543 on Build GRCh38
1686794 c.22-2A>G p.? Chr.19: 49878141-49878141 on Build GRCh38
135847 c.160G>A p.A54T Chr.19: 49878537-49878537 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs914365559 Chr.19: 49878544-49878544 on Build GRCh38
rs757397837 Chr.19: 49878534-49878534 on Build GRCh38
rs901907473 Chr.19: 49878438-49878438 on Build GRCh38
rs757598133 Chr.19: 49878281-49878296 on Build GRCh38
rs769982618 Chr.19: 49878149-49878149 on Build GRCh38
rs779044992 Chr.19: 49878537-49878537 on Build GRCh38
rs371372629 Chr.19: 49878251-49878251 on Build GRCh38
rs3745485 Chr.19: 49878104-49878104 on Build GRCh38
rs768002610 Chr.19: 49878506-49878506 on Build GRCh38
rs376664452 Chr.19: 49878277-49878277 on Build GRCh38
rs753942998 Chr.19: 49878527-49878527 on Build GRCh38
rs773064562 Chr.19: 49878496-49878496 on Build GRCh38
rs764547310 Chr.19: 49878526-49878526 on Build GRCh38
rs753257348 Chr.19: 49878236-49878236 on Build GRCh38
rs753384830 Chr.19: 49878207-49878207 on Build GRCh38
rs201003850 Chr.19: 49878492-49878492 on Build GRCh38
rs760294370 Chr.19: 49878109-49878109 on Build GRCh38
rs981080546 Chr.19: 49878232-49878232 on Build GRCh38
rs777787351 Chr.19: 49878152-49878152 on Build GRCh38
rs917600748 Chr.19: 49878177-49878177 on Build GRCh38
rs755451595 Chr.19: 49878219-49878219 on Build GRCh38
rs534507894 Chr.19: 49878528-49878528 on Build GRCh38
rs770579085 Chr.19: 49878158-49878158 on Build GRCh38
rs955732754 Chr.19: 49878512-49878512 on Build GRCh38
rs1038524711 Chr.19: 49878434-49878434 on Build GRCh38
rs761559919 Chr.19: 49878210-49878210 on Build GRCh38
rs758633584 Chr.19: 49878122-49878122 on Build GRCh38
rs781776519 Chr.19: 49878135-49878135 on Build GRCh38
rs753534207 Chr.19: 49878112-49878112 on Build GRCh38
rs760037815 Chr.19: 49878181-49878181 on Build GRCh38
rs201403786 Chr.19: 49878288-49878288 on Build GRCh38
rs1005660949 Chr.19: 49878254-49878254 on Build GRCh38
rs745661160 Chr.19: 49878272-49878272 on Build GRCh38
rs887589853 Chr.19: 49878470-49878470 on Build GRCh38
rs765948933 Chr.19: 49878492-49878492 on Build GRCh38
rs759378385 Chr.19: 49878163-49878163 on Build GRCh38
rs780490760 Chr.19: 49878123-49878123 on Build GRCh38
rs763676011 Chr.19: 49878206-49878206 on Build GRCh38
rs771165808 Chr.19: 49878268-49878268 on Build GRCh38
rs751940103 Chr.19: 49878280-49878281 on Build GRCh38
rs756672542 Chr.19: 49878518-49878518 on Build GRCh38
rs548569579 Chr.19: 49878447-49878447 on Build GRCh38
rs748224616 Chr.19: 49878138-49878138 on Build GRCh38
rs781763439 Chr.19: 49878228-49878228 on Build GRCh38
rs777928063 Chr.19: 49878244-49878244 on Build GRCh38
rs764486896 Chr.19: 49878205-49878206 on Build GRCh38
rs988296465 Chr.19: 49878242-49878242 on Build GRCh38
rs1040404271 Chr.19: 49878332-49878332 on Build GRCh38
rs967892281 Chr.19: 49878118-49878118 on Build GRCh38
rs138066596 Chr.19: 49878178-49878178 on Build GRCh38
rs147215692 Chr.19: 49878509-49878509 on Build GRCh38
rs550963060 Chr.19: 49878292-49878292 on Build GRCh38
rs371820666 Chr.19: 49878461-49878461 on Build GRCh38
rs536215010 Chr.19: 49878446-49878446 on Build GRCh38
rs943318246 Chr.19: 49878298-49878298 on Build GRCh38
rs377615932 Chr.19: 49878453-49878453 on Build GRCh38
rs566921605 Chr.19: 49878471-49878471 on Build GRCh38
rs202194413 Chr.19: 49878157-49878157 on Build GRCh38
rs867258368 Chr.19: 49878195-49878195 on Build GRCh38
rs775543649 Chr.19: 49878435-49878435 on Build GRCh38
rs771447717 Chr.19: 49878214-49878214 on Build GRCh38
rs755508077 Chr.19: 49878134-49878134 on Build GRCh38
rs759075735 Chr.19: 49878186-49878186 on Build GRCh38
rs759488701 Chr.19: 49878096-49878096 on Build GRCh38
rs910841003 Chr.19: 49878280-49878280 on Build GRCh38
rs368512102 Chr.19: 49878212-49878212 on Build GRCh38
rs752183948 Chr.19: 49878105-49878105 on Build GRCh38
rs774159609 Chr.19: 49878160-49878160 on Build GRCh38
rs569551779 Chr.19: 49878420-49878420 on Build GRCh38
rs773941249 Chr.19: 49878175-49878175 on Build GRCh38
rs1029700999 Chr.19: 49878093-49878093 on Build GRCh38
rs761276190 Chr.19: 49878484-49878484 on Build GRCh38
rs728898 Chr.19: 49878217-49878217 on Build GRCh38
rs756086406 Chr.19: 49878239-49878239 on Build GRCh38
rs374543547 Chr.19: 49878110-49878110 on Build GRCh38
rs373760628 Chr.19: 49878545-49878545 on Build GRCh38
rs968148616 Chr.19: 49878523-49878523 on Build GRCh38
rs900090064 Chr.19: 49878445-49878445 on Build GRCh38
rs143676147 Chr.19: 49878500-49878500 on Build GRCh38
rs144737346 Chr.19: 49878168-49878168 on Build GRCh38
rs370189185 Chr.19: 49878473-49878473 on Build GRCh38
rs779166451 Chr.19: 49878271-49878271 on Build GRCh38

View Full Product Details