Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 50791881-50792382 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACPT
Entrez Gene ID
93650
Gene Name
acid phosphatase, testicular
Gene Aliases
-
UniGene
Hs.293394
Gene Chromosome Location
Chr.19: 50790415-50795224 on Build GRCh38
Transcripts
NM_033068.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
999811 c.631A>C p.T211P Chr.19: 50792323-50792323 on Build GRCh38
5772143 c.543C>T p.G181G Chr.19: 50792165-50792165 on Build GRCh38
4630517 c.533C>T p.A178V Chr.19: 50792155-50792155 on Build GRCh38
6354737 c.584T>A p.L195Q Chr.19: 50792276-50792276 on Build GRCh38
1717946 c.541G>A p.G181S Chr.19: 50792163-50792163 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs995655593 Chr.19: 50792229-50792229 on Build GRCh38
rs775216661 Chr.19: 50792206-50792206 on Build GRCh38
rs751444282 Chr.19: 50792104-50792104 on Build GRCh38
rs781492393 Chr.19: 50792138-50792138 on Build GRCh38
rs1043568076 Chr.19: 50792144-50792144 on Build GRCh38
rs140570998 Chr.19: 50792141-50792141 on Build GRCh38
rs779532217 Chr.19: 50792308-50792308 on Build GRCh38
rs146503450 Chr.19: 50792132-50792132 on Build GRCh38
rs554344717 Chr.19: 50792326-50792326 on Build GRCh38
rs756735947 Chr.19: 50792173-50792173 on Build GRCh38
rs774074789 Chr.19: 50792035-50792035 on Build GRCh38
rs376827609 Chr.19: 50792271-50792271 on Build GRCh38
rs777162524 Chr.19: 50792127-50792127 on Build GRCh38
rs754524181 Chr.19: 50792300-50792300 on Build GRCh38
rs759788088 Chr.19: 50792065-50792065 on Build GRCh38
rs867971166 Chr.19: 50792355-50792355 on Build GRCh38
rs533594913 Chr.19: 50792228-50792228 on Build GRCh38
rs182178960 Chr.19: 50792060-50792060 on Build GRCh38
rs368106101 Chr.19: 50792182-50792182 on Build GRCh38
rs772226101 Chr.19: 50792257-50792257 on Build GRCh38
rs761747454 Chr.19: 50792284-50792284 on Build GRCh38
rs766692700 Chr.19: 50792103-50792103 on Build GRCh38
rs778345881 Chr.19: 50792177-50792177 on Build GRCh38
rs376944050 Chr.19: 50792163-50792163 on Build GRCh38
rs767376344 Chr.19: 50792114-50792114 on Build GRCh38
rs199887187 Chr.19: 50792160-50792160 on Build GRCh38
rs890428045 Chr.19: 50792040-50792040 on Build GRCh38
rs765141967 Chr.19: 50792288-50792288 on Build GRCh38
rs377747108 Chr.19: 50792238-50792238 on Build GRCh38
rs200104941 Chr.19: 50792067-50792067 on Build GRCh38
rs765260771 Chr.19: 50792092-50792092 on Build GRCh38
rs769597797 Chr.19: 50792048-50792051 on Build GRCh38
rs781246537 Chr.19: 50792036-50792036 on Build GRCh38
rs769030421 Chr.19: 50792137-50792137 on Build GRCh38
rs372111844 Chr.19: 50792189-50792189 on Build GRCh38
rs574997670 Chr.19: 50792000-50792000 on Build GRCh38
rs544529136 Chr.19: 50791991-50791991 on Build GRCh38
rs752612021 Chr.19: 50792121-50792121 on Build GRCh38
rs968796473 Chr.19: 50792078-50792078 on Build GRCh38
rs771170372 Chr.19: 50792191-50792191 on Build GRCh38
rs1018453708 Chr.19: 50792031-50792031 on Build GRCh38
rs559021778 Chr.19: 50791924-50791924 on Build GRCh38
rs759794933 Chr.19: 50792156-50792156 on Build GRCh38
rs560484483 Chr.19: 50792010-50792010 on Build GRCh38
rs1000273753 Chr.19: 50792195-50792195 on Build GRCh38
rs768698469 Chr.19: 50792357-50792357 on Build GRCh38
rs375560449 Chr.19: 50792146-50792146 on Build GRCh38
rs765760900 Chr.19: 50792299-50792299 on Build GRCh38
rs115449310 Chr.19: 50792279-50792279 on Build GRCh38
rs1031833722 Chr.19: 50792097-50792097 on Build GRCh38
rs774509973 Chr.19: 50792192-50792192 on Build GRCh38
rs963122022 Chr.19: 50792324-50792324 on Build GRCh38
rs903871533 Chr.19: 50791994-50791994 on Build GRCh38
rs762416150 Chr.19: 50792296-50792296 on Build GRCh38
rs1044788241 Chr.19: 50791983-50791983 on Build GRCh38
rs374772444 Chr.19: 50792178-50792178 on Build GRCh38
rs755605074 Chr.19: 50792052-50792052 on Build GRCh38
rs762331870 Chr.19: 50792091-50792091 on Build GRCh38
rs370460139 Chr.19: 50792170-50792170 on Build GRCh38
rs59340026 Chr.19: 50792233-50792233 on Build GRCh38
rs941820857 Chr.19: 50791999-50791999 on Build GRCh38
rs753834120 Chr.19: 50792216-50792216 on Build GRCh38
rs563921414 Chr.19: 50792041-50792041 on Build GRCh38
rs916365287 Chr.19: 50792276-50792276 on Build GRCh38
rs761698337 Chr.19: 50792223-50792223 on Build GRCh38
rs758526234 Chr.19: 50792322-50792322 on Build GRCh38
rs769418453 Chr.19: 50791971-50791971 on Build GRCh38
rs757781033 Chr.19: 50792250-50792250 on Build GRCh38
rs1007985851 Chr.19: 50792042-50792042 on Build GRCh38
rs190472139 Chr.19: 50791992-50791992 on Build GRCh38
rs747713242 Chr.19: 50792044-50792044 on Build GRCh38
rs202062875 Chr.19: 50792270-50792270 on Build GRCh38
rs201115271 Chr.19: 50792142-50792142 on Build GRCh38
rs763241730 Chr.19: 50792188-50792189 on Build GRCh38
rs763095653 Chr.19: 50792102-50792102 on Build GRCh38
rs373226837 Chr.19: 50792075-50792075 on Build GRCh38
rs779193664 Chr.19: 50792254-50792254 on Build GRCh38
rs775106229 Chr.19: 50792136-50792138 on Build GRCh38
rs745940385 Chr.19: 50792255-50792255 on Build GRCh38
rs1056332384 Chr.19: 50791977-50791977 on Build GRCh38
rs559203093 Chr.19: 50792231-50792231 on Build GRCh38
rs373891137 Chr.19: 50792190-50792190 on Build GRCh38
rs201693657 Chr.19: 50792301-50792301 on Build GRCh38
rs757907419 Chr.19: 50792306-50792306 on Build GRCh38
rs753327498 Chr.19: 50792171-50792171 on Build GRCh38
rs760589720 Chr.19: 50792167-50792167 on Build GRCh38
rs373190611 Chr.19: 50792232-50792232 on Build GRCh38
rs545383789 Chr.19: 50792003-50792003 on Build GRCh38
rs776221455 Chr.19: 50792280-50792280 on Build GRCh38
rs897734401 Chr.19: 50792006-50792006 on Build GRCh38
rs939094973 Chr.19: 50791944-50791944 on Build GRCh38
rs1047373271 Chr.19: 50791920-50791920 on Build GRCh38
rs764727401 Chr.19: 50792225-50792225 on Build GRCh38
rs201199714 Chr.19: 50792133-50792133 on Build GRCh38
rs780964560 Chr.19: 50791906-50791906 on Build GRCh38
rs988203580 Chr.19: 50791927-50791927 on Build GRCh38
rs970392273 Chr.19: 50792112-50792112 on Build GRCh38
rs930283201 Chr.19: 50791907-50791907 on Build GRCh38
rs144958224 Chr.19: 50792111-50792111 on Build GRCh38
rs894936679 Chr.19: 50791988-50791988 on Build GRCh38
rs368480133 Chr.19: 50792196-50792196 on Build GRCh38
rs867268298 Chr.19: 50792310-50792310 on Build GRCh38
rs3810283 Chr.19: 50791938-50791938 on Build GRCh38
rs747422053 Chr.19: 50792136-50792136 on Build GRCh38
rs375697800 Chr.19: 50792305-50792305 on Build GRCh38
rs773272778 Chr.19: 50792093-50792093 on Build GRCh38
rs748529676 Chr.19: 50792185-50792185 on Build GRCh38
rs760488832 Chr.19: 50792213-50792213 on Build GRCh38
rs562010990 Chr.19: 50792009-50792009 on Build GRCh38
rs531150593 Chr.19: 50792183-50792183 on Build GRCh38
rs754071127 Chr.19: 50791987-50791987 on Build GRCh38
rs774740893 Chr.19: 50792150-50792150 on Build GRCh38
rs949073382 Chr.19: 50791982-50791982 on Build GRCh38
rs777100668 Chr.19: 50792087-50792087 on Build GRCh38
rs775892526 Chr.19: 50792072-50792072 on Build GRCh38
rs756013460 Chr.19: 50792124-50792124 on Build GRCh38
rs144745203 Chr.19: 50792234-50792234 on Build GRCh38
rs745995702 Chr.19: 50792314-50792314 on Build GRCh38
rs370665864 Chr.19: 50792139-50792139 on Build GRCh38
rs1008722233 Chr.19: 50792015-50792015 on Build GRCh38
rs776464916 Chr.19: 50792359-50792359 on Build GRCh38
rs772410451 Chr.19: 50792198-50792198 on Build GRCh38
rs772772680 Chr.19: 50792292-50792292 on Build GRCh38

View Full Product Details