Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 50802102-50802501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf48
Entrez Gene ID
84798
Gene Name
chromosome 19 open reading frame 48
Gene Aliases
-
UniGene
Hs.256301
Gene Chromosome Location
Chr.19: 50797693-50804853 on Build GRCh38
Transcripts
NM_001290149.1 NM_001290150.1 NM_001290151.1 NM_001290152.1 NM_001290153.1 NM_001290154.1 NM_001290155.1 NM_032712.2 NM_199249.2 NM_199250.2 NR_003069.1

Gene Symbol
SNORD88C
Entrez Gene ID
692204
Gene Name
small nucleolar RNA, C/D box 88C
Gene Aliases
HBII-180C
UniGene
Gene Chromosome Location
Chr.19: 50802325-50802421 on Build GRCh38
Transcripts
NM_001290149.1 NM_001290150.1 NM_001290151.1 NM_001290152.1 NM_001290153.1 NM_001290154.1 NM_001290155.1 NM_032712.2 NM_199249.2 NM_199250.2 NR_003069.1

SNP Detail

SNP ID Chromosome Location
rs192992954 Chr.19: 50802144-50802144 on Build GRCh38
rs181447959 Chr.19: 50802383-50802383 on Build GRCh38
rs931917367 Chr.19: 50802180-50802180 on Build GRCh38
rs547160831 Chr.19: 50802444-50802444 on Build GRCh38
rs765461413 Chr.19: 50802425-50802425 on Build GRCh38
rs145969626 Chr.19: 50802341-50802341 on Build GRCh38
rs975822040 Chr.19: 50802410-50802410 on Build GRCh38
rs921760064 Chr.19: 50802176-50802176 on Build GRCh38
rs562269024 Chr.19: 50802390-50802390 on Build GRCh38
rs775962349 Chr.19: 50802234-50802234 on Build GRCh38
rs975874350 Chr.19: 50802166-50802166 on Build GRCh38
rs914701119 Chr.19: 50802391-50802391 on Build GRCh38
rs748416524 Chr.19: 50802335-50802335 on Build GRCh38
rs768964519 Chr.19: 50802329-50802329 on Build GRCh38
rs779427139 Chr.19: 50802297-50802297 on Build GRCh38
rs754575171 Chr.19: 50802282-50802282 on Build GRCh38
rs746292404 Chr.19: 50802301-50802301 on Build GRCh38
rs765973684 Chr.19: 50802381-50802381 on Build GRCh38
rs757621097 Chr.19: 50802401-50802401 on Build GRCh38
rs773208536 Chr.19: 50802360-50802360 on Build GRCh38
rs994517845 Chr.19: 50802317-50802317 on Build GRCh38
rs990083425 Chr.19: 50802417-50802417 on Build GRCh38
rs946043986 Chr.19: 50802220-50802220 on Build GRCh38
rs969514589 Chr.19: 50802233-50802233 on Build GRCh38
rs545850354 Chr.19: 50802292-50802292 on Build GRCh38
rs776621298 Chr.19: 50802332-50802332 on Build GRCh38
rs1050862622 Chr.19: 50802272-50802272 on Build GRCh38
rs761726255 Chr.19: 50802336-50802336 on Build GRCh38
rs765356632 Chr.19: 50802260-50802260 on Build GRCh38
rs1055926185 Chr.19: 50802303-50802303 on Build GRCh38
rs902199797 Chr.19: 50802229-50802229 on Build GRCh38
rs189400105 Chr.19: 50802225-50802225 on Build GRCh38
rs898432103 Chr.19: 50802316-50802316 on Build GRCh38
rs889547057 Chr.19: 50802456-50802456 on Build GRCh38
rs965861199 Chr.19: 50802385-50802385 on Build GRCh38
rs185286420 Chr.19: 50802208-50802208 on Build GRCh38
rs767179570 Chr.19: 50802384-50802384 on Build GRCh38
rs1025550196 Chr.19: 50802337-50802337 on Build GRCh38
rs907247870 Chr.19: 50802280-50802280 on Build GRCh38
rs758823890 Chr.19: 50802424-50802424 on Build GRCh38
rs779086712 Chr.19: 50802402-50802402 on Build GRCh38
rs772204248 Chr.19: 50802302-50802302 on Build GRCh38
rs762743269 Chr.19: 50802366-50802366 on Build GRCh38
rs370133036 Chr.19: 50802227-50802227 on Build GRCh38
rs914578591 Chr.19: 50802211-50802211 on Build GRCh38
rs2659064 Chr.19: 50802293-50802293 on Build GRCh38
rs775415632 Chr.19: 50802312-50802312 on Build GRCh38
rs781599703 Chr.19: 50802461-50802461 on Build GRCh38
rs1027171104 Chr.19: 50802353-50802353 on Build GRCh38
rs758656751 Chr.19: 50802392-50802392 on Build GRCh38
rs942404438 Chr.19: 50802460-50802460 on Build GRCh38
rs995738842 Chr.19: 50802340-50802340 on Build GRCh38
rs778178217 Chr.19: 50802288-50802288 on Build GRCh38
rs747318343 Chr.19: 50802328-50802328 on Build GRCh38
rs776711593 Chr.19: 50802365-50802365 on Build GRCh38
rs747034310 Chr.19: 50802445-50802445 on Build GRCh38
rs374736624 Chr.19: 50802171-50802171 on Build GRCh38
rs751028820 Chr.19: 50802382-50802382 on Build GRCh38
rs77815840 Chr.19: 50802454-50802454 on Build GRCh38
rs953171211 Chr.19: 50802415-50802415 on Build GRCh38
rs750994719 Chr.19: 50802412-50802412 on Build GRCh38
rs572063442 Chr.19: 50802270-50802270 on Build GRCh38
rs772356184 Chr.19: 50802416-50802416 on Build GRCh38
rs946043477 Chr.19: 50802467-50802467 on Build GRCh38

View Full Product Details