Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 50802362-50802854 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf48
Entrez Gene ID
84798
Gene Name
chromosome 19 open reading frame 48
Gene Aliases
-
UniGene
Hs.256301
Gene Chromosome Location
Chr.19: 50797693-50804853 on Build GRCh38
Transcripts
NM_001290149.1 NM_001290150.1 NM_001290151.1 NM_001290152.1 NM_001290153.1 NM_001290154.1 NM_001290155.1 NM_032712.2 NM_199249.2 NM_199250.2 NR_003069.1

Gene Symbol
SNORD88C
Entrez Gene ID
692204
Gene Name
small nucleolar RNA, C/D box 88C
Gene Aliases
HBII-180C
UniGene
Gene Chromosome Location
Chr.19: 50802325-50802421 on Build GRCh38
Transcripts
NM_001290149.1 NM_001290150.1 NM_001290151.1 NM_001290152.1 NM_001290153.1 NM_001290154.1 NM_001290155.1 NM_032712.2 NM_199249.2 NM_199250.2 NR_003069.1

SNP Detail

SNP ID Chromosome Location
rs938799515 Chr.19: 50802492-50802492 on Build GRCh38
rs778513651 Chr.19: 50802644-50802644 on Build GRCh38
rs181447959 Chr.19: 50802383-50802383 on Build GRCh38
rs527698275 Chr.19: 50802725-50802725 on Build GRCh38
rs767482552 Chr.19: 50802502-50802502 on Build GRCh38
rs901493661 Chr.19: 50802558-50802558 on Build GRCh38
rs566821264 Chr.19: 50802679-50802679 on Build GRCh38
rs907090135 Chr.19: 50802505-50802505 on Build GRCh38
rs908694748 Chr.19: 50802709-50802709 on Build GRCh38
rs547160831 Chr.19: 50802444-50802444 on Build GRCh38
rs765461413 Chr.19: 50802425-50802425 on Build GRCh38
rs745577401 Chr.19: 50802650-50802650 on Build GRCh38
rs975822040 Chr.19: 50802410-50802410 on Build GRCh38
rs562269024 Chr.19: 50802390-50802390 on Build GRCh38
rs986476599 Chr.19: 50802626-50802626 on Build GRCh38
rs914701119 Chr.19: 50802391-50802391 on Build GRCh38
rs773612444 Chr.19: 50802478-50802478 on Build GRCh38
rs765973684 Chr.19: 50802381-50802381 on Build GRCh38
rs757621097 Chr.19: 50802401-50802401 on Build GRCh38
rs928621096 Chr.19: 50802480-50802480 on Build GRCh38
rs1004869524 Chr.19: 50802523-50802523 on Build GRCh38
rs866581847 Chr.19: 50802591-50802591 on Build GRCh38
rs368991199 Chr.19: 50802494-50802494 on Build GRCh38
rs990083425 Chr.19: 50802417-50802417 on Build GRCh38
rs1011736272 Chr.19: 50802758-50802758 on Build GRCh38
rs1021490194 Chr.19: 50802794-50802794 on Build GRCh38
rs916099005 Chr.19: 50802501-50802501 on Build GRCh38
rs753342203 Chr.19: 50802611-50802611 on Build GRCh38
rs967600726 Chr.19: 50802807-50802807 on Build GRCh38
rs759386845 Chr.19: 50802500-50802500 on Build GRCh38
rs889547057 Chr.19: 50802456-50802456 on Build GRCh38
rs1055788524 Chr.19: 50802495-50802495 on Build GRCh38
rs965861199 Chr.19: 50802385-50802385 on Build GRCh38
rs752677134 Chr.19: 50802506-50802506 on Build GRCh38
rs184968769 Chr.19: 50802729-50802729 on Build GRCh38
rs1021698422 Chr.19: 50802580-50802580 on Build GRCh38
rs551389376 Chr.19: 50802621-50802621 on Build GRCh38
rs888908509 Chr.19: 50802755-50802755 on Build GRCh38
rs767179570 Chr.19: 50802384-50802384 on Build GRCh38
rs1048595720 Chr.19: 50802707-50802707 on Build GRCh38
rs758823890 Chr.19: 50802424-50802424 on Build GRCh38
rs779086712 Chr.19: 50802402-50802402 on Build GRCh38
rs887327419 Chr.19: 50802514-50802514 on Build GRCh38
rs780327452 Chr.19: 50802720-50802720 on Build GRCh38
rs2560931 Chr.19: 50802527-50802527 on Build GRCh38
rs898353147 Chr.19: 50802491-50802491 on Build GRCh38
rs997287708 Chr.19: 50802572-50802572 on Build GRCh38
rs910915891 Chr.19: 50802627-50802627 on Build GRCh38
rs1050179830 Chr.19: 50802740-50802740 on Build GRCh38
rs374665706 Chr.19: 50802612-50802612 on Build GRCh38
rs781599703 Chr.19: 50802461-50802461 on Build GRCh38
rs533139921 Chr.19: 50802618-50802618 on Build GRCh38
rs758656751 Chr.19: 50802392-50802392 on Build GRCh38
rs940313926 Chr.19: 50802712-50802712 on Build GRCh38
rs746799470 Chr.19: 50802511-50802511 on Build GRCh38
rs942404438 Chr.19: 50802460-50802460 on Build GRCh38
rs977612374 Chr.19: 50802590-50802590 on Build GRCh38
rs747034310 Chr.19: 50802445-50802445 on Build GRCh38
rs991548061 Chr.19: 50802666-50802666 on Build GRCh38
rs1041673468 Chr.19: 50802713-50802713 on Build GRCh38
rs77815840 Chr.19: 50802454-50802454 on Build GRCh38
rs751028820 Chr.19: 50802382-50802382 on Build GRCh38
rs1046861226 Chr.19: 50802747-50802747 on Build GRCh38
rs953171211 Chr.19: 50802415-50802415 on Build GRCh38
rs750994719 Chr.19: 50802412-50802412 on Build GRCh38
rs931483240 Chr.19: 50802694-50802694 on Build GRCh38
rs763952389 Chr.19: 50802584-50802584 on Build GRCh38
rs953203354 Chr.19: 50802601-50802601 on Build GRCh38
rs977730996 Chr.19: 50802641-50802641 on Build GRCh38
rs772356184 Chr.19: 50802416-50802416 on Build GRCh38
rs946043477 Chr.19: 50802467-50802467 on Build GRCh38

View Full Product Details