Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55350582-55351076 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX6B2
Entrez Gene ID
125965
Gene Name
cytochrome c oxidase subunit 6B2
Gene Aliases
COXVIB2, CT59
UniGene
Hs.550544
Gene Chromosome Location
Chr.19: 55349702-55354814 on Build GRCh38
Transcripts
NM_144613.4

SNP Detail

SNP ID Chromosome Location
rs1047397445 Chr.19: 55350674-55350674 on Build GRCh38
rs564396811 Chr.19: 55351003-55351003 on Build GRCh38
rs968558633 Chr.19: 55350700-55350700 on Build GRCh38
rs552200208 Chr.19: 55350725-55350725 on Build GRCh38
rs546149427 Chr.19: 55350960-55350960 on Build GRCh38
rs998312163 Chr.19: 55350901-55350901 on Build GRCh38
rs75135996 Chr.19: 55350756-55350756 on Build GRCh38
rs977190736 Chr.19: 55350701-55350701 on Build GRCh38
rs919622407 Chr.19: 55350941-55350941 on Build GRCh38
rs549234382 Chr.19: 55350843-55350843 on Build GRCh38
rs558646821 Chr.19: 55350917-55350917 on Build GRCh38
rs954112914 Chr.19: 55350937-55350937 on Build GRCh38
rs769303954 Chr.19: 55350896-55350896 on Build GRCh38
rs534439885 Chr.19: 55350820-55350820 on Build GRCh38
rs867457239 Chr.19: 55350643-55350643 on Build GRCh38
rs938422643 Chr.19: 55350735-55350735 on Build GRCh38
rs901274496 Chr.19: 55350877-55350877 on Build GRCh38
rs560667619 Chr.19: 55350628-55350628 on Build GRCh38
rs1056979544 Chr.19: 55351007-55351007 on Build GRCh38
rs538730035 Chr.19: 55350874-55350874 on Build GRCh38
rs532445566 Chr.19: 55350672-55350672 on Build GRCh38
rs1048678995 Chr.19: 55350786-55350786 on Build GRCh38
rs1031438069 Chr.19: 55351049-55351049 on Build GRCh38
rs73607032 Chr.19: 55350773-55350773 on Build GRCh38
rs965417856 Chr.19: 55350623-55350623 on Build GRCh38
rs1019358706 Chr.19: 55350869-55350869 on Build GRCh38
rs1028299324 Chr.19: 55350647-55350647 on Build GRCh38
rs772394113 Chr.19: 55350645-55350645 on Build GRCh38
rs1011319518 Chr.19: 55350956-55350956 on Build GRCh38
rs547653172 Chr.19: 55350837-55350837 on Build GRCh38
rs914489393 Chr.19: 55350677-55350677 on Build GRCh38
rs568627289 Chr.19: 55350812-55350812 on Build GRCh38
rs138149782 Chr.19: 55350872-55350872 on Build GRCh38
rs575315692 Chr.19: 55351038-55351038 on Build GRCh38
rs968566942 Chr.19: 55350992-55350992 on Build GRCh38
rs930947355 Chr.19: 55350943-55350943 on Build GRCh38
rs1023638476 Chr.19: 55350797-55350797 on Build GRCh38
rs1057215764 Chr.19: 55350737-55350737 on Build GRCh38
rs954173696 Chr.19: 55350661-55350661 on Build GRCh38
rs562533592 Chr.19: 55350729-55350729 on Build GRCh38
rs986928597 Chr.19: 55350838-55350838 on Build GRCh38
rs921941 Chr.19: 55351043-55351043 on Build GRCh38
rs553863555 Chr.19: 55350818-55350820 on Build GRCh38
rs894274192 Chr.19: 55351013-55351013 on Build GRCh38

View Full Product Details