Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 56810777-56811281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PEG3
Entrez Gene ID
5178
Gene Name
paternally expressed 3
Gene Aliases
PW1, ZKSCAN22, ZNF904, ZSCAN24
UniGene
Hs.731875
Gene Chromosome Location
Chr.19: 56810077-56840726 on Build GRCh38
Transcripts
NM_001146184.1 NM_001146185.1 NM_001146186.1 NM_001146187.1 NM_006210.2 NM_001146326.1 NM_001146327.1 NM_015363.4

Gene Symbol
ZIM2
Entrez Gene ID
23619
Gene Name
zinc finger imprinted 2
Gene Aliases
ZNF656
UniGene
Hs.731875
Gene Chromosome Location
Chr.19: 56774547-56840729 on Build GRCh38
Transcripts
NM_001146184.1 NM_001146185.1 NM_001146186.1 NM_001146187.1 NM_006210.2 NM_001146326.1 NM_001146327.1 NM_015363.4

SNP Detail

SNP ID Chromosome Location
rs908014051 Chr.19: 56810976-56810976 on Build GRCh38
rs200173182 Chr.19: 56811139-56811139 on Build GRCh38
rs922906543 Chr.19: 56811142-56811142 on Build GRCh38
rs1047530679 Chr.19: 56810969-56810970 on Build GRCh38
rs879616111 Chr.19: 56810936-56810936 on Build GRCh38
rs1012235138 Chr.19: 56811053-56811053 on Build GRCh38
rs1037955611 Chr.19: 56811008-56811008 on Build GRCh38
rs570232941 Chr.19: 56810838-56810842 on Build GRCh38
rs571048482 Chr.19: 56811251-56811251 on Build GRCh38
rs1008091931 Chr.19: 56810891-56810891 on Build GRCh38
rs34172934 Chr.19: 56811067-56811068 on Build GRCh38
rs398038497 Chr.19: 56811068-56811069 on Build GRCh38
rs541846048 Chr.19: 56811019-56811019 on Build GRCh38
rs937496442 Chr.19: 56811134-56811134 on Build GRCh38
rs923033513 Chr.19: 56810920-56810920 on Build GRCh38
rs28648863 Chr.19: 56810886-56810886 on Build GRCh38
rs766056225 Chr.19: 56811117-56811117 on Build GRCh38
rs1011405159 Chr.19: 56811069-56811069 on Build GRCh38
rs1046853876 Chr.19: 56811057-56811061 on Build GRCh38
rs1019905817 Chr.19: 56810924-56810924 on Build GRCh38
rs920045936 Chr.19: 56811212-56811212 on Build GRCh38
rs376017612 Chr.19: 56811137-56811137 on Build GRCh38
rs887055685 Chr.19: 56810889-56810889 on Build GRCh38
rs751235428 Chr.19: 56811242-56811242 on Build GRCh38
rs955883353 Chr.19: 56810831-56810831 on Build GRCh38
rs542201329 Chr.19: 56811022-56811022 on Build GRCh38
rs368589574 Chr.19: 56810906-56810906 on Build GRCh38
rs562556780 Chr.19: 56810830-56810830 on Build GRCh38
rs967660184 Chr.19: 56811066-56811066 on Build GRCh38
rs914495664 Chr.19: 56810851-56810851 on Build GRCh38
rs138560597 Chr.19: 56810809-56810809 on Build GRCh38
rs987831390 Chr.19: 56811056-56811056 on Build GRCh38
rs981649524 Chr.19: 56811068-56811068 on Build GRCh38
rs974850631 Chr.19: 56810918-56810918 on Build GRCh38
rs371223004 Chr.19: 56811250-56811252 on Build GRCh38
rs1021490733 Chr.19: 56811084-56811084 on Build GRCh38
rs963873137 Chr.19: 56810954-56810954 on Build GRCh38
rs561621956 Chr.19: 56811060-56811060 on Build GRCh38
rs957962327 Chr.19: 56811025-56811025 on Build GRCh38
rs995443051 Chr.19: 56810844-56810844 on Build GRCh38
rs528000961 Chr.19: 56811113-56811113 on Build GRCh38
rs1155461 Chr.19: 56811206-56811206 on Build GRCh38
rs997010751 Chr.19: 56811004-56811004 on Build GRCh38
rs749954743 Chr.19: 56810932-56810932 on Build GRCh38

View Full Product Details