Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57844476-57844971 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF587B
Entrez Gene ID
100293516
Gene Name
zinc finger protein 587B
Gene Aliases
-
UniGene
Hs.658251
Gene Chromosome Location
Chr.19: 57830294-57846238 on Build GRCh38
Transcripts
NM_001204818.1

SNP Detail

SNP ID Chromosome Location
rs1021903990 Chr.19: 57844647-57844647 on Build GRCh38
rs185718739 Chr.19: 57844515-57844515 on Build GRCh38
rs759857521 Chr.19: 57844601-57844601 on Build GRCh38
rs943353237 Chr.19: 57844781-57844781 on Build GRCh38
rs1038662758 Chr.19: 57844762-57844762 on Build GRCh38
rs1029443123 Chr.19: 57844866-57844866 on Build GRCh38
rs147012912 Chr.19: 57844766-57844769 on Build GRCh38
rs752788175 Chr.19: 57844727-57844727 on Build GRCh38
rs541989562 Chr.19: 57844701-57844701 on Build GRCh38
rs556919060 Chr.19: 57844622-57844622 on Build GRCh38
rs774654873 Chr.19: 57844531-57844531 on Build GRCh38
rs557594152 Chr.19: 57844545-57844545 on Build GRCh38
rs541447063 Chr.19: 57844780-57844780 on Build GRCh38
rs939106692 Chr.19: 57844725-57844725 on Build GRCh38
rs1029452893 Chr.19: 57844564-57844564 on Build GRCh38
rs866770135 Chr.19: 57844759-57844759 on Build GRCh38
rs536846005 Chr.19: 57844606-57844606 on Build GRCh38
rs919467151 Chr.19: 57844769-57844769 on Build GRCh38
rs991347392 Chr.19: 57844533-57844533 on Build GRCh38
rs555852060 Chr.19: 57844740-57844740 on Build GRCh38
rs1049372533 Chr.19: 57844774-57844774 on Build GRCh38
rs988796290 Chr.19: 57844628-57844628 on Build GRCh38
rs980430403 Chr.19: 57844924-57844924 on Build GRCh38
rs576468809 Chr.19: 57844694-57844694 on Build GRCh38
rs181477632 Chr.19: 57844514-57844514 on Build GRCh38
rs78892325 Chr.19: 57844892-57844892 on Build GRCh38
rs1022144358 Chr.19: 57844909-57844909 on Build GRCh38
rs969464847 Chr.19: 57844546-57844546 on Build GRCh38
rs192044067 Chr.19: 57844775-57844775 on Build GRCh38
rs866589248 Chr.19: 57844894-57844894 on Build GRCh38
rs571216551 Chr.19: 57844572-57844572 on Build GRCh38
rs927956705 Chr.19: 57844603-57844603 on Build GRCh38
rs373873807 Chr.19: 57844858-57844858 on Build GRCh38
rs886773521 Chr.19: 57844743-57844744 on Build GRCh38
rs992289759 Chr.19: 57844746-57844746 on Build GRCh38
rs1003564312 Chr.19: 57844820-57844820 on Build GRCh38
rs920989065 Chr.19: 57844733-57844733 on Build GRCh38
rs74643639 Chr.19: 57844874-57844874 on Build GRCh38
rs377766429 Chr.19: 57844932-57844932 on Build GRCh38
rs150289558 Chr.19: 57844836-57844836 on Build GRCh38
rs968383201 Chr.19: 57844912-57844912 on Build GRCh38
rs980865690 Chr.19: 57844696-57844696 on Build GRCh38
rs929538611 Chr.19: 57844771-57844771 on Build GRCh38
rs998047893 Chr.19: 57844864-57844864 on Build GRCh38
rs576154475 Chr.19: 57844796-57844796 on Build GRCh38
rs1004116336 Chr.19: 57844502-57844502 on Build GRCh38
rs1033767624 Chr.19: 57844930-57844930 on Build GRCh38
rs956618512 Chr.19: 57844593-57844593 on Build GRCh38
rs563763771 Chr.19: 57844939-57844939 on Build GRCh38
rs189039818 Chr.19: 57844499-57844499 on Build GRCh38
rs962855969 Chr.19: 57844530-57844530 on Build GRCh38
rs921417285 Chr.19: 57844920-57844920 on Build GRCh38
rs111493071 Chr.19: 57844897-57844897 on Build GRCh38
rs901936187 Chr.19: 57844527-57844527 on Build GRCh38
rs915528656 Chr.19: 57844670-57844670 on Build GRCh38

View Full Product Details