Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27138785-27139331 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRR30
Entrez Gene ID
339779
Gene Name
proline rich 30
Gene Aliases
C2orf53
UniGene
Hs.136555
Gene Chromosome Location
Chr.2: 27136847-27139464 on Build GRCh38
Transcripts
NM_178553.3

SNP Detail

SNP ID Chromosome Location
rs939497685 Chr.2: 27138843-27138843 on Build GRCh38
rs776445659 Chr.2: 27139050-27139050 on Build GRCh38
rs994690104 Chr.2: 27139160-27139160 on Build GRCh38
rs760380759 Chr.2: 27138983-27138983 on Build GRCh38
rs778255284 Chr.2: 27139246-27139246 on Build GRCh38
rs531811947 Chr.2: 27139197-27139197 on Build GRCh38
rs988392934 Chr.2: 27139126-27139126 on Build GRCh38
rs979584126 Chr.2: 27139294-27139294 on Build GRCh38
rs1018841917 Chr.2: 27138950-27138950 on Build GRCh38
rs951896617 Chr.2: 27139006-27139006 on Build GRCh38
rs1056462382 Chr.2: 27139105-27139105 on Build GRCh38
rs76778733 Chr.2: 27138859-27138859 on Build GRCh38
rs905305918 Chr.2: 27139062-27139062 on Build GRCh38
rs946882271 Chr.2: 27139053-27139053 on Build GRCh38
rs375758200 Chr.2: 27139293-27139293 on Build GRCh38
rs993440574 Chr.2: 27138969-27138969 on Build GRCh38
rs549943660 Chr.2: 27139200-27139200 on Build GRCh38
rs141761343 Chr.2: 27138909-27138909 on Build GRCh38
rs561013747 Chr.2: 27139153-27139153 on Build GRCh38
rs76897518 Chr.2: 27139061-27139061 on Build GRCh38
rs1034962141 Chr.2: 27138845-27138845 on Build GRCh38
rs752966382 Chr.2: 27139111-27139111 on Build GRCh38
rs577187396 Chr.2: 27138956-27138956 on Build GRCh38
rs993409547 Chr.2: 27139274-27139274 on Build GRCh38
rs1006478668 Chr.2: 27139302-27139302 on Build GRCh38
rs1007987462 Chr.2: 27138931-27138931 on Build GRCh38
rs773197291 Chr.2: 27138978-27138978 on Build GRCh38
rs946894536 Chr.2: 27139161-27139161 on Build GRCh38
rs966135006 Chr.2: 27139256-27139256 on Build GRCh38
rs946801137 Chr.2: 27138878-27138878 on Build GRCh38
rs774284637 Chr.2: 27138906-27138906 on Build GRCh38
rs943492411 Chr.2: 27139211-27139211 on Build GRCh38
rs914142451 Chr.2: 27139284-27139284 on Build GRCh38
rs914149534 Chr.2: 27139052-27139052 on Build GRCh38
rs973225581 Chr.2: 27138974-27138974 on Build GRCh38
rs148009071 Chr.2: 27139259-27139259 on Build GRCh38
rs896504064 Chr.2: 27139106-27139106 on Build GRCh38
rs911588622 Chr.2: 27139012-27139012 on Build GRCh38
rs1018703013 Chr.2: 27139250-27139250 on Build GRCh38
rs3795961 Chr.2: 27139079-27139079 on Build GRCh38
rs545112908 Chr.2: 27138961-27138961 on Build GRCh38
rs771895593 Chr.2: 27138962-27138962 on Build GRCh38
rs1004216557 Chr.2: 27139135-27139135 on Build GRCh38
rs928219319 Chr.2: 27138810-27138810 on Build GRCh38
rs755130780 Chr.2: 27139023-27139023 on Build GRCh38
rs1027406421 Chr.2: 27138975-27138975 on Build GRCh38
rs944368698 Chr.2: 27139031-27139031 on Build GRCh38
rs969721668 Chr.2: 27139078-27139078 on Build GRCh38
rs968306645 Chr.2: 27139289-27139289 on Build GRCh38
rs1040766701 Chr.2: 27139236-27139236 on Build GRCh38
rs560536293 Chr.2: 27138999-27138999 on Build GRCh38
rs756427167 Chr.2: 27139156-27139156 on Build GRCh38
rs1007338780 Chr.2: 27139219-27139219 on Build GRCh38
rs1002164526 Chr.2: 27138826-27138826 on Build GRCh38
rs766089833 Chr.2: 27139040-27139040 on Build GRCh38
rs536338534 Chr.2: 27138855-27138855 on Build GRCh38

View Full Product Details