Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 68974201-68974695 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GKN1
Entrez Gene ID
56287
Gene Name
gastrokine 1
Gene Aliases
AMP18, BRICD1, CA11, FOV, foveolin
UniGene
Hs.69319
Gene Chromosome Location
Chr.2: 68974573-68980980 on Build GRCh38
Transcripts
NM_019617.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
171925 c.31C>T p.R11C Chr.2: 68974666-68974666 on Build GRCh38
1208246 c.41A>G p.K14R Chr.2: 68974676-68974676 on Build GRCh38
7088634 c.6_7insG p.A3fs*10 Chr.2: 68974641-68974642 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs4575760 Chr.2: 68974369-68974369 on Build GRCh38
rs967993399 Chr.2: 68974472-68974472 on Build GRCh38
rs539820808 Chr.2: 68974368-68974368 on Build GRCh38
rs985490616 Chr.2: 68974272-68974272 on Build GRCh38
rs776796427 Chr.2: 68974601-68974601 on Build GRCh38
rs777548916 Chr.2: 68974594-68974594 on Build GRCh38
rs929513349 Chr.2: 68974282-68974282 on Build GRCh38
rs764076803 Chr.2: 68974632-68974632 on Build GRCh38
rs540168684 Chr.2: 68974425-68974425 on Build GRCh38
rs751865109 Chr.2: 68974637-68974637 on Build GRCh38
rs760154006 Chr.2: 68974613-68974613 on Build GRCh38
rs923497149 Chr.2: 68974280-68974280 on Build GRCh38
rs535639758 Chr.2: 68974285-68974285 on Build GRCh38
rs766199481 Chr.2: 68974642-68974642 on Build GRCh38
rs370749546 Chr.2: 68974436-68974436 on Build GRCh38
rs576233549 Chr.2: 68974551-68974551 on Build GRCh38
rs757966201 Chr.2: 68974659-68974659 on Build GRCh38
rs961487980 Chr.2: 68974465-68974465 on Build GRCh38
rs561465663 Chr.2: 68974573-68974573 on Build GRCh38
rs746632056 Chr.2: 68974596-68974596 on Build GRCh38
rs141746794 Chr.2: 68974666-68974666 on Build GRCh38
rs754746622 Chr.2: 68974647-68974647 on Build GRCh38
rs777093189 Chr.2: 68974661-68974661 on Build GRCh38
rs569869827 Chr.2: 68974284-68974284 on Build GRCh38
rs1026998272 Chr.2: 68974524-68974524 on Build GRCh38
rs533283558 Chr.2: 68974227-68974227 on Build GRCh38
rs892295271 Chr.2: 68974353-68974353 on Build GRCh38
rs979383190 Chr.2: 68974558-68974558 on Build GRCh38
rs574319100 Chr.2: 68974410-68974410 on Build GRCh38
rs386647040 Chr.2: 68974368-68974369 on Build GRCh38
rs1013831620 Chr.2: 68974440-68974440 on Build GRCh38
rs374698018 Chr.2: 68974619-68974619 on Build GRCh38
rs770373405 Chr.2: 68974616-68974616 on Build GRCh38
rs573913250 Chr.2: 68974618-68974618 on Build GRCh38
rs979463370 Chr.2: 68974235-68974235 on Build GRCh38
rs761881352 Chr.2: 68974639-68974639 on Build GRCh38
rs1054785075 Chr.2: 68974374-68974374 on Build GRCh38
rs79761945 Chr.2: 68974263-68974263 on Build GRCh38
rs752627332 Chr.2: 68974657-68974657 on Build GRCh38
rs556576197 Chr.2: 68974523-68974523 on Build GRCh38
rs371699740 Chr.2: 68974605-68974605 on Build GRCh38
rs75564632 Chr.2: 68974359-68974359 on Build GRCh38
rs368541570 Chr.2: 68974622-68974622 on Build GRCh38
rs943480341 Chr.2: 68974249-68974249 on Build GRCh38
rs143488484 Chr.2: 68974593-68974593 on Build GRCh38
rs768486211 Chr.2: 68974598-68974598 on Build GRCh38
rs1040734018 Chr.2: 68974312-68974312 on Build GRCh38
rs568525329 Chr.2: 68974344-68974344 on Build GRCh38
rs146025449 Chr.2: 68974653-68974653 on Build GRCh38
rs985973137 Chr.2: 68974534-68974534 on Build GRCh38
rs995652509 Chr.2: 68974477-68974477 on Build GRCh38
rs115053382 Chr.2: 68974568-68974568 on Build GRCh38
rs753529833 Chr.2: 68974643-68974643 on Build GRCh38
rs766345312 Chr.2: 68974311-68974311 on Build GRCh38
rs1038069054 Chr.2: 68974331-68974331 on Build GRCh38
rs17524213 Chr.2: 68974299-68974299 on Build GRCh38
rs1016270055 Chr.2: 68974446-68974446 on Build GRCh38
rs746280871 Chr.2: 68974671-68974671 on Build GRCh38
rs945168602 Chr.2: 68974634-68974634 on Build GRCh38
rs968024577 Chr.2: 68974229-68974229 on Build GRCh38
rs974095855 Chr.2: 68974237-68974237 on Build GRCh38
rs954115820 Chr.2: 68974529-68974529 on Build GRCh38
rs17524220 Chr.2: 68974354-68974354 on Build GRCh38
rs920015953 Chr.2: 68974258-68974258 on Build GRCh38
rs139153499 Chr.2: 68974667-68974667 on Build GRCh38

View Full Product Details