Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 68980646-68981150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GKN1
Entrez Gene ID
56287
Gene Name
gastrokine 1
Gene Aliases
AMP18, BRICD1, CA11, FOV, foveolin
UniGene
Hs.69319
Gene Chromosome Location
Chr.2: 68974573-68980980 on Build GRCh38
Transcripts
NM_019617.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6439881 c.557G>A p.W186* Chr.2: 68980780-68980780 on Build GRCh38
3963000 c.533C>A p.T178K Chr.2: 68980756-68980756 on Build GRCh38
1306945 c.567C>T p.D189D Chr.2: 68980790-68980790 on Build GRCh38
5531712 c.530G>A p.G177E Chr.2: 68980753-68980753 on Build GRCh38
7001657 c.523_524insT p.Y175fs*>26 Chr.2: 68980746-68980747 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs570810015 Chr.2: 68981069-68981069 on Build GRCh38
rs749908943 Chr.2: 68981014-68981014 on Build GRCh38
rs965257973 Chr.2: 68981048-68981048 on Build GRCh38
rs141743643 Chr.2: 68980766-68980766 on Build GRCh38
rs771420012 Chr.2: 68980786-68980786 on Build GRCh38
rs180826670 Chr.2: 68981079-68981079 on Build GRCh38
rs777358672 Chr.2: 68980852-68980852 on Build GRCh38
rs775865514 Chr.2: 68980710-68980710 on Build GRCh38
rs966858361 Chr.2: 68980879-68980879 on Build GRCh38
rs905028659 Chr.2: 68980808-68980808 on Build GRCh38
rs896618563 Chr.2: 68980761-68980761 on Build GRCh38
rs761269262 Chr.2: 68980681-68980681 on Build GRCh38
rs200701146 Chr.2: 68980765-68980765 on Build GRCh38
rs949661747 Chr.2: 68981100-68981100 on Build GRCh38
rs754540068 Chr.2: 68980757-68980757 on Build GRCh38
rs867175797 Chr.2: 68980845-68980845 on Build GRCh38
rs754871164 Chr.2: 68980838-68980838 on Build GRCh38
rs55669437 Chr.2: 68980713-68980713 on Build GRCh38
rs757898143 Chr.2: 68980851-68980851 on Build GRCh38
rs1030568285 Chr.2: 68981086-68981086 on Build GRCh38
rs777249036 Chr.2: 68980702-68980702 on Build GRCh38
rs775388744 Chr.2: 68980802-68980802 on Build GRCh38
rs147341504 Chr.2: 68980781-68980781 on Build GRCh38
rs556437954 Chr.2: 68981115-68981115 on Build GRCh38
rs76956057 Chr.2: 68980933-68980933 on Build GRCh38
rs13431582 Chr.2: 68980889-68980889 on Build GRCh38
rs968154048 Chr.2: 68980931-68980931 on Build GRCh38
rs1037604522 Chr.2: 68981111-68981111 on Build GRCh38
rs113178059 Chr.2: 68980856-68980856 on Build GRCh38
rs377206306 Chr.2: 68980811-68980811 on Build GRCh38
rs56276500 Chr.2: 68980718-68980718 on Build GRCh38
rs766057682 Chr.2: 68980706-68980706 on Build GRCh38
rs752199284 Chr.2: 68980847-68980847 on Build GRCh38
rs1017746295 Chr.2: 68981013-68981013 on Build GRCh38
rs547502695 Chr.2: 68981017-68981017 on Build GRCh38
rs576221373 Chr.2: 68980756-68980756 on Build GRCh38
rs73934216 Chr.2: 68980855-68980855 on Build GRCh38
rs564424588 Chr.2: 68980833-68980833 on Build GRCh38
rs143595030 Chr.2: 68980772-68980772 on Build GRCh38
rs368638189 Chr.2: 68980827-68980827 on Build GRCh38
rs933628191 Chr.2: 68980709-68980709 on Build GRCh38
rs781633101 Chr.2: 68980865-68980865 on Build GRCh38
rs76063320 Chr.2: 68980707-68980707 on Build GRCh38
rs568420578 Chr.2: 68980987-68980987 on Build GRCh38
rs771618252 Chr.2: 68980684-68980684 on Build GRCh38
rs755914962 Chr.2: 68980746-68980746 on Build GRCh38
rs767834542 Chr.2: 68980828-68980828 on Build GRCh38
rs778329473 Chr.2: 68980762-68980762 on Build GRCh38
rs867068263 Chr.2: 68981102-68981102 on Build GRCh38
rs191187603 Chr.2: 68980739-68980739 on Build GRCh38
rs757664009 Chr.2: 68981123-68981123 on Build GRCh38
rs759880395 Chr.2: 68980740-68980740 on Build GRCh38
rs540638476 Chr.2: 68980787-68980787 on Build GRCh38
rs937680579 Chr.2: 68981001-68981001 on Build GRCh38
rs982436141 Chr.2: 68981106-68981106 on Build GRCh38
rs371896178 Chr.2: 68980843-68980843 on Build GRCh38
rs571029572 Chr.2: 68980691-68980691 on Build GRCh38
rs563513924 Chr.2: 68980888-68980888 on Build GRCh38
rs766342923 Chr.2: 68980751-68980751 on Build GRCh38
rs75590022 Chr.2: 68981004-68981004 on Build GRCh38
rs145631217 Chr.2: 68980799-68980799 on Build GRCh38
rs767374627 Chr.2: 68980724-68980724 on Build GRCh38
rs774774481 Chr.2: 68980691-68980692 on Build GRCh38
rs760528900 Chr.2: 68980813-68980813 on Build GRCh38

View Full Product Details