Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73901197-73901708 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTG2
Entrez Gene ID
72
Gene Name
actin, gamma 2, smooth muscle, enteric
Gene Aliases
ACT, ACTA3, ACTE, ACTL3, ACTSG, VSCM
UniGene
Hs.516105
Gene Chromosome Location
Chr.2: 73892966-73919653 on Build GRCh38
Transcripts
NM_001199893.1 NM_001615.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3659951 c.110G>A p.G37D Chr.2: 73901421-73901421 on Build GRCh38
5025653 c.28G>A p.V10M Chr.2: 73901339-73901339 on Build GRCh38
265959 c.26T>C p.L9P Chr.2: 73901337-73901337 on Build GRCh38
6199144 c.126+5G>A p.? Chr.2: 73901442-73901442 on Build GRCh38
2998858 c.18C>T p.T6T Chr.2: 73901329-73901329 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202149912 Chr.2: 73901304-73901304 on Build GRCh38
rs994899400 Chr.2: 73901586-73901586 on Build GRCh38
rs373948665 Chr.2: 73901631-73901631 on Build GRCh38
rs1653258 Chr.2: 73901234-73901234 on Build GRCh38
rs985644157 Chr.2: 73901650-73901650 on Build GRCh38
rs759125518 Chr.2: 73901470-73901470 on Build GRCh38
rs756929932 Chr.2: 73901261-73901261 on Build GRCh38
rs760259308 Chr.2: 73901335-73901335 on Build GRCh38
rs564571961 Chr.2: 73901674-73901674 on Build GRCh38
rs747241400 Chr.2: 73901392-73901392 on Build GRCh38
rs779522349 Chr.2: 73901377-73901377 on Build GRCh38
rs1027440718 Chr.2: 73901589-73901589 on Build GRCh38
rs757542398 Chr.2: 73901487-73901487 on Build GRCh38
rs761722608 Chr.2: 73901442-73901442 on Build GRCh38
rs965625894 Chr.2: 73901334-73901334 on Build GRCh38
rs750129394 Chr.2: 73901332-73901332 on Build GRCh38
rs760349315 Chr.2: 73901454-73901454 on Build GRCh38
rs1010567857 Chr.2: 73901327-73901327 on Build GRCh38
rs888865878 Chr.2: 73901591-73901591 on Build GRCh38
rs755461337 Chr.2: 73901356-73901356 on Build GRCh38
rs587777386 Chr.2: 73901430-73901430 on Build GRCh38
rs536702128 Chr.2: 73901565-73901565 on Build GRCh38
rs201921654 Chr.2: 73901606-73901606 on Build GRCh38
rs774136655 Chr.2: 73901315-73901315 on Build GRCh38
rs751926642 Chr.2: 73901486-73901486 on Build GRCh38
rs587777385 Chr.2: 73901429-73901429 on Build GRCh38
rs576110822 Chr.2: 73901563-73901563 on Build GRCh38
rs140664533 Chr.2: 73901588-73901589 on Build GRCh38
rs756282497 Chr.2: 73901542-73901587 on Build GRCh38
rs758891000 Chr.2: 73901389-73901389 on Build GRCh38
rs753346451 Chr.2: 73901353-73901353 on Build GRCh38
rs58042852 Chr.2: 73901576-73901581 on Build GRCh38
rs1007281663 Chr.2: 73901602-73901602 on Build GRCh38
rs537853238 Chr.2: 73901314-73901314 on Build GRCh38
rs534471099 Chr.2: 73901399-73901399 on Build GRCh38
rs376018484 Chr.2: 73901558-73901558 on Build GRCh38
rs547720185 Chr.2: 73901539-73901539 on Build GRCh38
rs774409368 Chr.2: 73901434-73901434 on Build GRCh38
rs111608790 Chr.2: 73901534-73901534 on Build GRCh38
rs370710991 Chr.2: 73901338-73901338 on Build GRCh38
rs763390017 Chr.2: 73901456-73901456 on Build GRCh38
rs530365654 Chr.2: 73901267-73901267 on Build GRCh38
rs758909306 Chr.2: 73901481-73901481 on Build GRCh38
rs753409338 Chr.2: 73901464-73901464 on Build GRCh38
rs528525442 Chr.2: 73901675-73901675 on Build GRCh38
rs752140530 Chr.2: 73901678-73901678 on Build GRCh38
rs774477696 Chr.2: 73901556-73901556 on Build GRCh38
rs748078701 Chr.2: 73901506-73901506 on Build GRCh38
rs112261658 Chr.2: 73901582-73901582 on Build GRCh38
rs965904961 Chr.2: 73901621-73901621 on Build GRCh38
rs536483468 Chr.2: 73901554-73901554 on Build GRCh38
rs746012984 Chr.2: 73901416-73901416 on Build GRCh38
rs754508567 Chr.2: 73901514-73901514 on Build GRCh38
rs780047231 Chr.2: 73901503-73901503 on Build GRCh38
rs58947529 Chr.2: 73901581-73901582 on Build GRCh38
rs753224642 Chr.2: 73901385-73901385 on Build GRCh38
rs771954142 Chr.2: 73901508-73901508 on Build GRCh38
rs746077804 Chr.2: 73901495-73901495 on Build GRCh38
rs534506938 Chr.2: 73901671-73901671 on Build GRCh38
rs768591779 Chr.2: 73901312-73901312 on Build GRCh38
rs867058297 Chr.2: 73901584-73901584 on Build GRCh38
rs563700454 Chr.2: 73901648-73901648 on Build GRCh38
rs548470785 Chr.2: 73901274-73901274 on Build GRCh38
rs751325833 Chr.2: 73901391-73901391 on Build GRCh38
rs773010897 Chr.2: 73901512-73901512 on Build GRCh38
rs771864268 Chr.2: 73901443-73901443 on Build GRCh38
rs781764684 Chr.2: 73901485-73901485 on Build GRCh38
rs143078935 Chr.2: 73901540-73901587 on Build GRCh38
rs868158708 Chr.2: 73901575-73901575 on Build GRCh38
rs1034877601 Chr.2: 73901453-73901453 on Build GRCh38
rs778208494 Chr.2: 73901291-73901291 on Build GRCh38
rs769735590 Chr.2: 73901423-73901423 on Build GRCh38
rs897514713 Chr.2: 73901529-73901529 on Build GRCh38
rs199742302 Chr.2: 73901479-73901479 on Build GRCh38
rs540620186 Chr.2: 73901608-73901608 on Build GRCh38
rs532670077 Chr.2: 73901256-73901256 on Build GRCh38
rs879935763 Chr.2: 73901538-73901587 on Build GRCh38
rs776419371 Chr.2: 73901374-73901374 on Build GRCh38
rs1721241 Chr.2: 73901264-73901264 on Build GRCh38
rs148146087 Chr.2: 73901670-73901670 on Build GRCh38
rs764639740 Chr.2: 73901477-73901477 on Build GRCh38
rs766006420 Chr.2: 73901460-73901460 on Build GRCh38
rs1632564 Chr.2: 73901242-73901242 on Build GRCh38
rs761640844 Chr.2: 73901321-73901321 on Build GRCh38
rs201108131 Chr.2: 73901447-73901447 on Build GRCh38
rs560021348 Chr.2: 73901610-73901610 on Build GRCh38
rs369953659 Chr.2: 73901676-73901676 on Build GRCh38
rs979166437 Chr.2: 73901649-73901649 on Build GRCh38
rs372174317 Chr.2: 73901223-73901223 on Build GRCh38
rs764300189 Chr.2: 73901476-73901476 on Build GRCh38
rs575014634 Chr.2: 73901622-73901622 on Build GRCh38
rs781520451 Chr.2: 73901490-73901490 on Build GRCh38
rs538941366 Chr.2: 73901538-73901538 on Build GRCh38
rs1050146 Chr.2: 73901306-73901306 on Build GRCh38
rs1018466274 Chr.2: 73901604-73901604 on Build GRCh38
rs756271325 Chr.2: 73901502-73901502 on Build GRCh38
rs565816688 Chr.2: 73901543-73901543 on Build GRCh38
rs746869758 Chr.2: 73901516-73901516 on Build GRCh38
rs767210199 Chr.2: 73901263-73901263 on Build GRCh38
rs908663671 Chr.2: 73901235-73901235 on Build GRCh38
rs772542600 Chr.2: 73901290-73901290 on Build GRCh38
rs202017875 Chr.2: 73901260-73901260 on Build GRCh38
rs142489416 Chr.2: 73901534-73901558 on Build GRCh38
rs946019028 Chr.2: 73901380-73901380 on Build GRCh38
rs375149400 Chr.2: 73901414-73901414 on Build GRCh38
rs869312168 Chr.2: 73901424-73901424 on Build GRCh38
rs368989094 Chr.2: 73901218-73901219 on Build GRCh38
rs748495115 Chr.2: 73901282-73901282 on Build GRCh38
rs375675581 Chr.2: 73901328-73901328 on Build GRCh38
rs558138424 Chr.2: 73901569-73901569 on Build GRCh38
rs778227043 Chr.2: 73901390-73901390 on Build GRCh38
rs747221259 Chr.2: 73901292-73901292 on Build GRCh38
rs4852320 Chr.2: 73901587-73901587 on Build GRCh38
rs1015568316 Chr.2: 73901513-73901513 on Build GRCh38
rs371230166 Chr.2: 73901458-73901458 on Build GRCh38
rs771147501 Chr.2: 73901396-73901396 on Build GRCh38
rs147083008 Chr.2: 73901243-73901243 on Build GRCh38
rs200192309 Chr.2: 73901281-73901281 on Build GRCh38
rs978896811 Chr.2: 73901413-73901413 on Build GRCh38

View Full Product Details