Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74554667-74555172 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOK1
Entrez Gene ID
1796
Gene Name
docking protein 1
Gene Aliases
P62DOK, pp62
UniGene
Hs.103854
Gene Chromosome Location
Chr.2: 74549020-74557551 on Build GRCh38
Transcripts
NM_001197260.1 NM_001318866.1 NM_001318867.1 NM_001318868.1 NM_001318869.1 NM_001381.4 XM_017003528.1 XM_011533134.2

Gene Symbol
LOXL3
Entrez Gene ID
84695
Gene Name
lysyl oxidase like 3
Gene Aliases
LOXL
UniGene
Hs.469045
Gene Chromosome Location
Chr.2: 74532258-74555709 on Build GRCh38
Transcripts
NM_001197260.1 NM_001318866.1 NM_001318867.1 NM_001318868.1 NM_001318869.1 NM_001381.4 XM_017003528.1 XM_011533134.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6891604 c.48C>T p.R16R Chr.2: 74554802-74554802 on Build GRCh38
2999945 c.28_29insT p.L12fs*33 Chr.2: 74554782-74554783 on Build GRCh38
4679136 c.36G>A p.L12L Chr.2: 74554790-74554790 on Build GRCh38
5817877 c.7G>C p.G3R Chr.2: 74554761-74554761 on Build GRCh38
1409613 c.29delT p.L12fs*26 Chr.2: 74554783-74554783 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758140616 Chr.2: 74554746-74554746 on Build GRCh38
rs762025271 Chr.2: 74554715-74554715 on Build GRCh38
rs1026665420 Chr.2: 74555108-74555108 on Build GRCh38
rs577425194 Chr.2: 74554755-74554755 on Build GRCh38
rs868513708 Chr.2: 74554760-74554760 on Build GRCh38
rs774514795 Chr.2: 74554705-74554705 on Build GRCh38
rs201014730 Chr.2: 74554765-74554765 on Build GRCh38
rs773465066 Chr.2: 74554794-74554794 on Build GRCh38
rs896042079 Chr.2: 74555136-74555136 on Build GRCh38
rs867919536 Chr.2: 74554791-74554791 on Build GRCh38
rs768640940 Chr.2: 74554704-74554704 on Build GRCh38
rs754352565 Chr.2: 74554731-74554731 on Build GRCh38
rs770761102 Chr.2: 74554708-74554722 on Build GRCh38
rs766878361 Chr.2: 74554726-74554726 on Build GRCh38
rs1043937137 Chr.2: 74555091-74555091 on Build GRCh38
rs775106963 Chr.2: 74554862-74554863 on Build GRCh38
rs777159494 Chr.2: 74554807-74554807 on Build GRCh38
rs931664116 Chr.2: 74554699-74554699 on Build GRCh38
rs771241130 Chr.2: 74554732-74554732 on Build GRCh38
rs372307156 Chr.2: 74554806-74554806 on Build GRCh38
rs757521955 Chr.2: 74555141-74555142 on Build GRCh38
rs771264270 Chr.2: 74554894-74554894 on Build GRCh38
rs763812885 Chr.2: 74554827-74554827 on Build GRCh38
rs773833722 Chr.2: 74554753-74554753 on Build GRCh38
rs200362897 Chr.2: 74554748-74554748 on Build GRCh38
rs13416172 Chr.2: 74554744-74554744 on Build GRCh38
rs189420857 Chr.2: 74555040-74555040 on Build GRCh38
rs1037849755 Chr.2: 74554780-74554780 on Build GRCh38
rs868544813 Chr.2: 74554804-74554804 on Build GRCh38
rs1041950821 Chr.2: 74555010-74555010 on Build GRCh38
rs748197144 Chr.2: 74554856-74554856 on Build GRCh38
rs754402002 Chr.2: 74555136-74555139 on Build GRCh38
rs1003244133 Chr.2: 74555029-74555030 on Build GRCh38
rs533910133 Chr.2: 74555079-74555079 on Build GRCh38
rs1033413028 Chr.2: 74554690-74554691 on Build GRCh38
rs747230717 Chr.2: 74554889-74554889 on Build GRCh38
rs369803198 Chr.2: 74555137-74555137 on Build GRCh38
rs942086901 Chr.2: 74555089-74555089 on Build GRCh38
rs781132302 Chr.2: 74554751-74554751 on Build GRCh38
rs757036727 Chr.2: 74554835-74554835 on Build GRCh38
rs767256272 Chr.2: 74555138-74555138 on Build GRCh38
rs764683660 Chr.2: 74555135-74555136 on Build GRCh38
rs767325562 Chr.2: 74554860-74554861 on Build GRCh38
rs1050029349 Chr.2: 74554703-74554703 on Build GRCh38
rs759252474 Chr.2: 74554782-74554783 on Build GRCh38
rs757047878 Chr.2: 74554750-74554750 on Build GRCh38
rs780034806 Chr.2: 74554847-74554847 on Build GRCh38
rs910643946 Chr.2: 74555069-74555069 on Build GRCh38
rs775100757 Chr.2: 74555125-74555125 on Build GRCh38
rs914660353 Chr.2: 74554901-74554901 on Build GRCh38
rs1054573741 Chr.2: 74554853-74554853 on Build GRCh38
rs954900947 Chr.2: 74555003-74555003 on Build GRCh38
rs549635023 Chr.2: 74554763-74554763 on Build GRCh38
rs751366665 Chr.2: 74554747-74554747 on Build GRCh38
rs766283877 Chr.2: 74555146-74555146 on Build GRCh38
rs10208552 Chr.2: 74554828-74554828 on Build GRCh38
rs762574239 Chr.2: 74555130-74555130 on Build GRCh38
rs750315992 Chr.2: 74554842-74554842 on Build GRCh38
rs747377401 Chr.2: 74554797-74554797 on Build GRCh38
rs775129381 Chr.2: 74554824-74554824 on Build GRCh38
rs373347212 Chr.2: 74555116-74555116 on Build GRCh38
rs910621810 Chr.2: 74554768-74554768 on Build GRCh38
rs13008836 Chr.2: 74554982-74554982 on Build GRCh38
rs746305702 Chr.2: 74555107-74555107 on Build GRCh38
rs988060405 Chr.2: 74554987-74554987 on Build GRCh38
rs986119339 Chr.2: 74555022-74555022 on Build GRCh38
rs1032103008 Chr.2: 74554956-74554956 on Build GRCh38
rs956533560 Chr.2: 74554969-74554969 on Build GRCh38
rs988112758 Chr.2: 74554719-74554719 on Build GRCh38
rs771222739 Chr.2: 74554801-74554801 on Build GRCh38
rs761014351 Chr.2: 74554723-74554723 on Build GRCh38
rs1023819497 Chr.2: 74554888-74554888 on Build GRCh38
rs377209805 Chr.2: 74554718-74554718 on Build GRCh38
rs1057386503 Chr.2: 74554830-74554830 on Build GRCh38
rs556382904 Chr.2: 74555147-74555147 on Build GRCh38
rs200468716 Chr.2: 74555136-74555137 on Build GRCh38
rs375038866 Chr.2: 74555121-74555121 on Build GRCh38
rs1030185107 Chr.2: 74554994-74554994 on Build GRCh38
rs200278469 Chr.2: 74554864-74554864 on Build GRCh38
rs776876262 Chr.2: 74554908-74554908 on Build GRCh38
rs755991157 Chr.2: 74554846-74554846 on Build GRCh38
rs911465988 Chr.2: 74554711-74554711 on Build GRCh38
rs751436681 Chr.2: 74555140-74555140 on Build GRCh38
rs182270753 Chr.2: 74555139-74555139 on Build GRCh38
rs759968687 Chr.2: 74554811-74554811 on Build GRCh38
rs1001111136 Chr.2: 74554942-74554942 on Build GRCh38
rs748283135 Chr.2: 74554782-74554782 on Build GRCh38
rs772278891 Chr.2: 74554792-74554792 on Build GRCh38
rs769613274 Chr.2: 74555026-74555026 on Build GRCh38
rs995196504 Chr.2: 74555101-74555101 on Build GRCh38
rs368294305 Chr.2: 74554761-74554761 on Build GRCh38
rs904919353 Chr.2: 74554932-74554932 on Build GRCh38
rs942203972 Chr.2: 74554773-74554773 on Build GRCh38
rs762588482 Chr.2: 74554826-74554826 on Build GRCh38
rs773554847 Chr.2: 74554720-74554720 on Build GRCh38
rs368897775 Chr.2: 74555127-74555127 on Build GRCh38

View Full Product Details