Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 135202744-135203245 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZRANB3
Entrez Gene ID
84083
Gene Name
zinc finger RANBP2-type containing 3
Gene Aliases
4933425L19Rik, AH2
UniGene
Hs.658422
Gene Chromosome Location
Chr.2: 135164703-135531322 on Build GRCh38
Transcripts
NM_001286568.1 NM_001286569.1 NM_032143.3 XM_005263809.1 XM_006712788.1 XM_011511958.2 XM_011511959.2 XM_011511960.1 XM_011511961.2 XM_011511963.1 XM_011511966.2 XM_017005065.1 XM_017005066.1 XM_017005067.1 XM_017005068.1 XM_017005069.1 XM_017005070.1 XM_017005071.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2150178 c.3119T>C p.L1040P Chr.2: 135202854-135202854 on Build GRCh38
6368692 c.3106C>A p.Q1036K Chr.2: 135202861-135202861 on Build GRCh38
4950355 c.3028G>C p.G1010R Chr.2: 135202939-135202939 on Build GRCh38
1006801 c.1509_1510delCT p.C504fs*13 Chr.2: 135202852-135202853 on Build GRCh38
1006803 c.1447G>T p.D483Y Chr.2: 135202915-135202915 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773956606 Chr.2: 135202933-135202933 on Build GRCh38
rs564697474 Chr.2: 135203043-135203043 on Build GRCh38
rs1039812886 Chr.2: 135203093-135203093 on Build GRCh38
rs372611635 Chr.2: 135202893-135202893 on Build GRCh38
rs757610097 Chr.2: 135202874-135202874 on Build GRCh38
rs759327259 Chr.2: 135203022-135203022 on Build GRCh38
rs550993466 Chr.2: 135203129-135203129 on Build GRCh38
rs898938997 Chr.2: 135202972-135202972 on Build GRCh38
rs756121781 Chr.2: 135202950-135202952 on Build GRCh38
rs756811223 Chr.2: 135202990-135202990 on Build GRCh38
rs57097780 Chr.2: 135203046-135203046 on Build GRCh38
rs906502921 Chr.2: 135202827-135202827 on Build GRCh38
rs187035401 Chr.2: 135202853-135202853 on Build GRCh38
rs746258198 Chr.2: 135202885-135202885 on Build GRCh38
rs764958175 Chr.2: 135203009-135203009 on Build GRCh38
rs370569566 Chr.2: 135202969-135202969 on Build GRCh38
rs995911120 Chr.2: 135202978-135202978 on Build GRCh38
rs866644990 Chr.2: 135202891-135202891 on Build GRCh38
rs886352097 Chr.2: 135202858-135202858 on Build GRCh38
rs562168720 Chr.2: 135203075-135203075 on Build GRCh38
rs766139527 Chr.2: 135202788-135202788 on Build GRCh38
rs753482802 Chr.2: 135202871-135202871 on Build GRCh38
rs779108761 Chr.2: 135202884-135202884 on Build GRCh38
rs992738669 Chr.2: 135203015-135203015 on Build GRCh38
rs1016012597 Chr.2: 135202961-135202961 on Build GRCh38
rs377526122 Chr.2: 135202822-135202822 on Build GRCh38
rs759415274 Chr.2: 135202802-135202802 on Build GRCh38
rs569042275 Chr.2: 135203203-135203203 on Build GRCh38
rs939332357 Chr.2: 135202844-135202844 on Build GRCh38
rs747084229 Chr.2: 135202894-135202894 on Build GRCh38
rs149093985 Chr.2: 135202989-135202989 on Build GRCh38
rs968549580 Chr.2: 135202942-135202942 on Build GRCh38
rs759329064 Chr.2: 135202951-135202951 on Build GRCh38
rs948146327 Chr.2: 135203180-135203180 on Build GRCh38
rs182438031 Chr.2: 135202768-135202768 on Build GRCh38
rs751645395 Chr.2: 135202929-135202930 on Build GRCh38
rs915387078 Chr.2: 135203156-135203156 on Build GRCh38
rs764062272 Chr.2: 135202979-135202980 on Build GRCh38
rs767453595 Chr.2: 135202810-135202810 on Build GRCh38
rs766584385 Chr.2: 135202837-135202838 on Build GRCh38
rs755687130 Chr.2: 135202841-135202841 on Build GRCh38
rs767436995 Chr.2: 135202963-135202963 on Build GRCh38
rs375963644 Chr.2: 135202895-135202895 on Build GRCh38
rs201227471 Chr.2: 135202793-135202793 on Build GRCh38
rs748275029 Chr.2: 135202907-135202907 on Build GRCh38
rs775463343 Chr.2: 135202964-135202964 on Build GRCh38
rs758902867 Chr.2: 135203011-135203011 on Build GRCh38
rs909605087 Chr.2: 135203078-135203078 on Build GRCh38
rs941127198 Chr.2: 135203090-135203090 on Build GRCh38
rs892463102 Chr.2: 135202800-135202800 on Build GRCh38
rs763552179 Chr.2: 135202986-135202986 on Build GRCh38
rs770074349 Chr.2: 135202923-135202923 on Build GRCh38
rs558802170 Chr.2: 135202787-135202787 on Build GRCh38
rs16831434 Chr.2: 135202983-135202983 on Build GRCh38
rs76157281 Chr.2: 135203051-135203051 on Build GRCh38
rs529271086 Chr.2: 135203105-135203105 on Build GRCh38

View Full Product Details